Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175736.4 | NP_783863.4 | 17 | 2136 | |
NM_198900.2 | NP_944489.2 | 16 | 1983 | ||
XM_005269218.2 | XP_005269275.1 | 17 | 2115 | ||
XM_011538968.2 | XP_011537270.1 | 17 | 2605 | ||
XM_011538969.2 | XP_011537271.1 | 17 | 2604 | ||
XM_011538970.2 | XP_011537272.1 | 17 | 2604 | ||
XM_011538971.2 | XP_011537273.1 | 17 | 2180 | ||
XM_011538972.2 | XP_011537274.1 | 16 | 2450 | ||
XM_011538973.2 | XP_011537275.2 | 13 | 1595 | ||
XM_011538974.2 | XP_011537276.1 | 18 | 1937 | ||
GenBank mRNA | AB095934.2 | - | 17 | 2136 | |
AK127200.1 | - | 4 | 609 | ||
AK128195.1 | - | 16 | 1963 | ||
AL162062.1 | - | 1 | 93 | ||
BC159100.1 | - | 17 | 2136 | 8860524 |