Gene Transcripts
- Gene Symbol
- ZMYM4
- Entrez Gene ID
- 9202
- Gene Name
- zinc finger MYM-type containing 4
- Gene Aliases
- CDIR, MYM, ZNF198L3, ZNF262
- Chromosome Location
- Chr. 1: 35268694 - 35421944 on Build GRCh38
- UniGene ID
- Hs.269211
- Species
- Homo sapiens
- Gene Symbol
- ZMYM4-AS1
- Entrez Gene ID
- 100861513
- Gene Name
- ZMYM4 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.585397
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BG187713.1 |
- |
2 |
302 |
|
BG208679.1 |
- |
2 |
318 |
|
View Full Product Details