Gene Transcripts

Gene Symbol
ATPIF1
Entrez Gene ID
93974
Gene Name
ATPase inhibitory factor 1
Gene Aliases
ATPI, ATPIP, IP
Chromosome Location
Chr. 1: 28236091 - 28238105 on Build GRCh38
UniGene ID
Hs.744914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016311.4 NP_057395.1 1, 2 180
NM_178190.2 NP_835497.1 1, 2 180
NM_178191.2 NP_835498.1 1, 2 180
GenBank mRNA AB029042.1 - 1, 2 108
AF114836.1 - 266
AK000934.1 - 1, 2 117
AK316600.1 - 1, 2 167
AL050386.1 - 1, 2 110
AL583540.3 - 1, 2 103
BC004955.1 - 1, 2 63 3625379
BC009677.1 - 1, 2 138 3877506
BG036978.1 - 1, 2 152
BG337531.1 - 1, 2 136
BG714388.1 - 1, 2 173
BP271519.1 - 1, 2 176
BT009849.1 - 1, 2 87
CR457097.1 - 1, 2 87
DB030607.1 - 1, 2 180

View Full Product Details