Gene Transcripts

Gene Symbol
ORMDL3
Entrez Gene ID
94103
Gene Name
ORMDL sphingolipid biosynthesis regulator 3
Gene Aliases
Chromosome Location
Chr. 17: 39921041 - 39927804 on Build GRCh38
UniGene ID
Hs.514151
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320801.1 NP_001307730.1 6 1893
NM_001320802.1 NP_001307731.1 4 731
NM_001320803.1 NP_001307732.1 4 445
NM_139280.3 NP_644809.1 4 736
XM_017025345.1 XP_016880834.1 4 589
XM_017025346.1 XP_016880835.1 4 502
XM_017025347.1 XP_016880836.1 4 682
XM_017025348.1 XP_016880837.1 4 428
GenBank mRNA AF373101.1 - 4 472
AF373102.1 - 4 542
AF395708.1 - 4 527
AK074811.1 - 4 533
AK075212.1 - 4 488
AK093063.1 - 6 1888
AX775843.1 - 4 490
AY357943.1 - 3 345
BC017087.1 - 4 502 3850211
BC071833.1 - 4 481 6340437
DB042662.1 - 4 446

View Full Product Details