Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318876.1 | NP_001305805.1 | 6 | 710 | |
NM_203290.3 | NP_976035.1 | 6 | 710 | ||
XM_011515000.2 | XP_011513302.1 | 6 | 651 | ||
GenBank mRNA | AF008442.1 | - | 6 | 636 | |
AF047441.1 | - | 6 | 646 | ||
AK293561.1 | - | 5 | 863 | ||
BC008118.1 | - | 6 | 601 | ||
BC008863.2 | - | 6 | 636 | 4128104 | |
BQ575535.1 | - | 4 | 638 |