Gene Transcripts
- Gene Symbol
- CLCA3P
- Entrez Gene ID
- 9629
- Gene Name
- chloride channel accessory 3, pseudogene
- Gene Aliases
- CLCA3
- Chromosome Location
- Chr. 1: 86634276 - 86655376 on Build GRCh38
- UniGene ID
- Hs.673847
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024604.1 |
|
2 |
233 |
|
GenBank mRNA |
AF043976.1 |
- |
2 |
233 |
|
View Full Product Details