Gene Transcripts

Gene Symbol
BMS1
Entrez Gene ID
9790
Gene Name
BMS1, ribosome biogenesis factor
Gene Aliases
ACC, BMS1L
Chromosome Location
Chr. 10: 42782506 - 42834937 on Build GRCh38
UniGene ID
Hs.10848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014753.3 NP_055568.3 22 3874
XM_005271846.3 XP_005271903.1 22 3749
XM_005271848.3 XP_005271905.1 21 3416
XM_011540402.2 XP_011538704.1 22 3605
XM_011540403.2 XP_011538705.1 14 2408
GenBank mRNA BC043345.1 - 22 3569
BC150252.1 - 21 3543 9007274
D80009.1 - 21 3543

View Full Product Details