Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001173533.1 | NP_001167004.1 | 4 | 686 | |
NM_001173534.1 | NP_001167005.1 | 4 | 677 | ||
NM_001184781.1 | NP_001171710.1 | 5 | 800 | ||
NM_005137.2 | NP_005128.1 | 5 | 809 | ||
NR_033674.1 | 4 | 694 | |||
GenBank mRNA | AK291670.1 | - | 5 | 767 | |
AK295697.1 | - | 4 | 652 | ||
AK296337.1 | - | 4 | 635 | ||
AK303130.1 | - | 5 | 719 | ||
AK304382.1 | - | 4 | 644 | ||
BC020339.1 | - | 5 | 756 | ||
BC032430.1 | - | 5 | 745 | 5243412 | |
BC035425.1 | - | 5 | 756 | ||
BC040500.1 | - | 5 | 804 | 5272271 | |
CR456433.1 | - | 5 | 581 | ||
CR936871.1 | - | 5 | 754 | ||
D79985.1 | - | 5 | 746 | ||
FJ032369.1 | - | 5 | 561 | ||
L77560.1 | - | 5 | 710 | ||
X83545.1 | - | 5 | 710 | ||
X84076.1 | - | 5 | 707 |