Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
KCNE3 | NM_005472.4 | 3 | 2911 | NP_005463.1 |
XM_017017047.1 | XP_016872536.1 | |||
XM_017017048.1 | XP_016872537.1 | |||
XM_017017049.1 | XP_016872538.1 | |||
XM_017017050.1 | XP_016872539.1 | |||
XM_017017051.1 | XP_016872540.1 | |||
XM_017017052.1 | XP_016872541.1 | |||
CK301015.1 | 1 | 176 | ||
CR936625.1 | CAI56768.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1041154 | Chr11:74403756 - 74466864 on Build GRCh38 | Loss | KCNE3 |
esv34979 | Chr11:74167643 - 74822407 on Build GRCh38 | Gain | MIR4696 RNF169 P4HA3 PPME1 POLD3 LOC100287896 C2CD3 MIR548AL LIPT2 CHRDL2 KCNE3 PGM2L1 LOC101928580 |
esv3626840 | Chr11:74371082 - 74464389 on Build GRCh38 | Gain | MIR548AL KCNE3 PGM2L1 |
esv2759837 | Chr11:74131564 - 74846552 on Build GRCh38 | Gain | MIR4696 RNF169 P4HA3 PPME1 POLD3 LOC100287896 C2CD3 MIR548AL XRRA1 LIPT2 CHRDL2 KCNE3 PGM2L1 LOC101928580 |
nsv555443 | Chr11:74398121 - 74469444 on Build GRCh38 | Loss | MIR548AL KCNE3 PGM2L1 |
nsv832209 | Chr11:74444838 - 74618074 on Build GRCh38 | Gain | POLD3 LIPT2 KCNE3 LOC100287896 |
dgv209e214 | Chr11:74225600 - 74828943 on Build GRCh38 | Gain | MIR4696 MIR548AL RNF169 P4HA3 PPME1 POLD3 LIPT2 CHRDL2 KCNE3 LOC100287896 PGM2L1 LOC101928580 |
nsv393 | Chr11:74424817 - 74470372 on Build GRCh38 | Deletion | KCNE3 |
nsv825994 | Chr11:74225622 - 74823410 on Build GRCh38 | Gain | MIR4696 MIR548AL RNF169 P4HA3 PPME1 POLD3 LIPT2 CHRDL2 KCNE3 LOC100287896 PGM2L1 LOC101928580 |