Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72687496-72687895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARAP1
Entrez Gene ID
116985
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1
Gene Aliases
CENTD2
UniGene
Hs.503165
Gene Chromosome Location
Chr.11: 72685069-72752389 on Build GRCh38
Transcripts
NM_001040118.2 NM_001135190.1 NM_015242.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5727095 c.4074G>A p.W1358* Chr.11: 72687735-72687735 on Build GRCh38
4036533 c.3336C>T p.C1112C Chr.11: 72687738-72687738 on Build GRCh38
4036532 c.4071C>T p.C1357C Chr.11: 72687738-72687738 on Build GRCh38
5727096 c.3339G>A p.W1113* Chr.11: 72687735-72687735 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs969466618 Chr.11: 72687539-72687539 on Build GRCh38
rs546111421 Chr.11: 72687614-72687614 on Build GRCh38
rs995973249 Chr.11: 72687679-72687679 on Build GRCh38
rs763991166 Chr.11: 72687775-72687775 on Build GRCh38
rs1031183364 Chr.11: 72687768-72687768 on Build GRCh38
rs762384227 Chr.11: 72687532-72687533 on Build GRCh38
rs371527949 Chr.11: 72687531-72687531 on Build GRCh38
rs769669938 Chr.11: 72687677-72687678 on Build GRCh38
rs751361625 Chr.11: 72687780-72687780 on Build GRCh38
rs753376805 Chr.11: 72687517-72687517 on Build GRCh38
rs765902691 Chr.11: 72687583-72687583 on Build GRCh38
rs201803171 Chr.11: 72687518-72687518 on Build GRCh38
rs549862951 Chr.11: 72687720-72687720 on Build GRCh38
rs146840475 Chr.11: 72687717-72687717 on Build GRCh38
rs867823597 Chr.11: 72687831-72687831 on Build GRCh38
rs372286747 Chr.11: 72687781-72687781 on Build GRCh38
rs774439609 Chr.11: 72687678-72687678 on Build GRCh38
rs938523856 Chr.11: 72687581-72687581 on Build GRCh38
rs766051917 Chr.11: 72687726-72687726 on Build GRCh38
rs866354979 Chr.11: 72687692-72687692 on Build GRCh38
rs927185806 Chr.11: 72687577-72687577 on Build GRCh38
rs752631781 Chr.11: 72687544-72687544 on Build GRCh38
rs565231057 Chr.11: 72687584-72687584 on Build GRCh38
rs907813320 Chr.11: 72687602-72687602 on Build GRCh38
rs748618032 Chr.11: 72687681-72687681 on Build GRCh38
rs367895167 Chr.11: 72687630-72687630 on Build GRCh38
rs763413928 Chr.11: 72687802-72687802 on Build GRCh38
rs758439617 Chr.11: 72687554-72687554 on Build GRCh38
rs1002142227 Chr.11: 72687805-72687805 on Build GRCh38
rs956980095 Chr.11: 72687792-72687792 on Build GRCh38
rs980337547 Chr.11: 72687561-72687561 on Build GRCh38
rs564308923 Chr.11: 72687637-72687637 on Build GRCh38
rs768666873 Chr.11: 72687677-72687677 on Build GRCh38
rs761780393 Chr.11: 72687552-72687566 on Build GRCh38
rs377103103 Chr.11: 72687784-72687784 on Build GRCh38
rs776082058 Chr.11: 72687735-72687735 on Build GRCh38
rs377486073 Chr.11: 72687762-72687762 on Build GRCh38
rs765997504 Chr.11: 72687516-72687516 on Build GRCh38
rs11541355 Chr.11: 72687732-72687732 on Build GRCh38
rs972747081 Chr.11: 72687541-72687541 on Build GRCh38
rs918659426 Chr.11: 72687576-72687576 on Build GRCh38
rs367690655 Chr.11: 72687519-72687519 on Build GRCh38
rs911019235 Chr.11: 72687625-72687625 on Build GRCh38
rs370370408 Chr.11: 72687636-72687636 on Build GRCh38
rs764798099 Chr.11: 72687759-72687759 on Build GRCh38
rs750812323 Chr.11: 72687661-72687661 on Build GRCh38
rs376069265 Chr.11: 72687647-72687647 on Build GRCh38
rs368198851 Chr.11: 72687802-72687802 on Build GRCh38
rs756342724 Chr.11: 72687658-72687658 on Build GRCh38
rs1048976329 Chr.11: 72687592-72687592 on Build GRCh38
rs765694677 Chr.11: 72687588-72687588 on Build GRCh38
rs561588357 Chr.11: 72687722-72687722 on Build GRCh38
rs531376905 Chr.11: 72687665-72687665 on Build GRCh38
rs772611277 Chr.11: 72687688-72687688 on Build GRCh38
rs749424939 Chr.11: 72687787-72687787 on Build GRCh38
rs368139520 Chr.11: 72687742-72687742 on Build GRCh38

View Full Product Details