Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72695437-72695944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARAP1
Entrez Gene ID
116985
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1
Gene Aliases
CENTD2
UniGene
Hs.503165
Gene Chromosome Location
Chr.11: 72685069-72752389 on Build GRCh38
Transcripts
NM_001040118.2 NM_001135190.1 NM_015242.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3986447 c.3496A>G p.S1166G Chr.11: 72695553-72695553 on Build GRCh38
5027444 c.2604G>A p.T868T Chr.11: 72695799-72695799 on Build GRCh38
4489144 c.3438C>T p.L1146L Chr.11: 72695611-72695611 on Build GRCh38
5786378 c.2671G>A p.V891M Chr.11: 72695732-72695732 on Build GRCh38
3687624 c.2642G>T p.R881L Chr.11: 72695761-72695761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35361895 Chr.11: 72695460-72695461 on Build GRCh38
rs556952780 Chr.11: 72695516-72695516 on Build GRCh38
rs757643978 Chr.11: 72695464-72695464 on Build GRCh38
rs994172879 Chr.11: 72695772-72695772 on Build GRCh38
rs374344913 Chr.11: 72695505-72695505 on Build GRCh38
rs768202949 Chr.11: 72695696-72695696 on Build GRCh38
rs758440606 Chr.11: 72695559-72695559 on Build GRCh38
rs772864582 Chr.11: 72695873-72695873 on Build GRCh38
rs774657163 Chr.11: 72695904-72695904 on Build GRCh38
rs767717922 Chr.11: 72695774-72695774 on Build GRCh38
rs760533059 Chr.11: 72695664-72695664 on Build GRCh38
rs767918911 Chr.11: 72695515-72695516 on Build GRCh38
rs371492893 Chr.11: 72695518-72695518 on Build GRCh38
rs753489643 Chr.11: 72695712-72695712 on Build GRCh38
rs376839214 Chr.11: 72695641-72695641 on Build GRCh38
rs764201350 Chr.11: 72695673-72695673 on Build GRCh38
rs140387760 Chr.11: 72695480-72695480 on Build GRCh38
rs771995025 Chr.11: 72695731-72695731 on Build GRCh38
rs1042487904 Chr.11: 72695682-72695682 on Build GRCh38
rs368732834 Chr.11: 72695579-72695579 on Build GRCh38
rs769714640 Chr.11: 72695895-72695895 on Build GRCh38
rs1025205258 Chr.11: 72695765-72695765 on Build GRCh38
rs768814992 Chr.11: 72695762-72695762 on Build GRCh38
rs748893567 Chr.11: 72695483-72695483 on Build GRCh38
rs985517608 Chr.11: 72695583-72695583 on Build GRCh38
rs79040639 Chr.11: 72695718-72695718 on Build GRCh38
rs184640744 Chr.11: 72695672-72695672 on Build GRCh38
rs750910537 Chr.11: 72695779-72695779 on Build GRCh38
rs755715040 Chr.11: 72695600-72695600 on Build GRCh38
rs765555817 Chr.11: 72695545-72695545 on Build GRCh38
rs144234265 Chr.11: 72695799-72695799 on Build GRCh38
rs550326288 Chr.11: 72695569-72695569 on Build GRCh38
rs773764618 Chr.11: 72695640-72695640 on Build GRCh38
rs910348405 Chr.11: 72695588-72695588 on Build GRCh38
rs777625487 Chr.11: 72695732-72695732 on Build GRCh38
rs139124425 Chr.11: 72695771-72695771 on Build GRCh38
rs774848845 Chr.11: 72695481-72695481 on Build GRCh38
rs752068100 Chr.11: 72695568-72695568 on Build GRCh38
rs781710172 Chr.11: 72695470-72695470 on Build GRCh38
rs745714810 Chr.11: 72695893-72695893 on Build GRCh38
rs745482620 Chr.11: 72695471-72695471 on Build GRCh38
rs369948223 Chr.11: 72695714-72695714 on Build GRCh38
rs903526504 Chr.11: 72695711-72695711 on Build GRCh38
rs768054464 Chr.11: 72695502-72695502 on Build GRCh38
rs371419962 Chr.11: 72695665-72695665 on Build GRCh38
rs778607456 Chr.11: 72695819-72695819 on Build GRCh38
rs762394815 Chr.11: 72695683-72695683 on Build GRCh38
rs751843732 Chr.11: 72695463-72695464 on Build GRCh38
rs756442916 Chr.11: 72695704-72695704 on Build GRCh38
rs373421125 Chr.11: 72695761-72695761 on Build GRCh38
rs756618649 Chr.11: 72695864-72695865 on Build GRCh38
rs181489523 Chr.11: 72695885-72695885 on Build GRCh38
rs367984439 Chr.11: 72695832-72695832 on Build GRCh38
rs1018327868 Chr.11: 72695839-72695839 on Build GRCh38
rs201935587 Chr.11: 72695675-72695675 on Build GRCh38
rs761118663 Chr.11: 72695796-72695796 on Build GRCh38
rs914002782 Chr.11: 72695724-72695724 on Build GRCh38
rs770716173 Chr.11: 72695733-72695733 on Build GRCh38
rs747481932 Chr.11: 72695460-72695460 on Build GRCh38
rs754628608 Chr.11: 72695713-72695713 on Build GRCh38
rs774567132 Chr.11: 72695768-72695768 on Build GRCh38
rs757849933 Chr.11: 72695834-72695834 on Build GRCh38
rs759735831 Chr.11: 72695529-72695529 on Build GRCh38
rs985487874 Chr.11: 72695805-72695805 on Build GRCh38
rs764099489 Chr.11: 72695566-72695566 on Build GRCh38
rs574284921 Chr.11: 72695754-72695754 on Build GRCh38
rs976345726 Chr.11: 72695853-72695853 on Build GRCh38
rs368907367 Chr.11: 72695646-72695646 on Build GRCh38
rs753041272 Chr.11: 72695692-72695698 on Build GRCh38
rs772981848 Chr.11: 72695647-72695647 on Build GRCh38
rs768164284 Chr.11: 72695628-72695628 on Build GRCh38
rs781024353 Chr.11: 72695886-72695886 on Build GRCh38
rs1043611030 Chr.11: 72695509-72695509 on Build GRCh38
rs750969452 Chr.11: 72695702-72695702 on Build GRCh38
rs754647744 Chr.11: 72695800-72695800 on Build GRCh38
rs143873269 Chr.11: 72695844-72695844 on Build GRCh38
rs1025660403 Chr.11: 72695461-72695461 on Build GRCh38
rs748904956 Chr.11: 72695625-72695625 on Build GRCh38
rs775729590 Chr.11: 72695667-72695667 on Build GRCh38
rs372334680 Chr.11: 72695601-72695601 on Build GRCh38
rs780306136 Chr.11: 72695707-72695707 on Build GRCh38
rs759732843 Chr.11: 72695516-72695516 on Build GRCh38
rs752718628 Chr.11: 72695553-72695553 on Build GRCh38
rs943225189 Chr.11: 72695639-72695639 on Build GRCh38
rs781671391 Chr.11: 72695570-72695570 on Build GRCh38
rs1026052568 Chr.11: 72695889-72695889 on Build GRCh38
rs746929372 Chr.11: 72695845-72695845 on Build GRCh38

View Full Product Details