Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86945546-86946069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD4
Entrez Gene ID
8322
Gene Name
frizzled class receptor 4
Gene Aliases
CD344, EVR1, FEVR, FZD4S, Fz-4, Fz4, FzE4, GPCR, hFz4
UniGene
Hs.591968
Gene Chromosome Location
Chr.11: 86945675-86955398 on Build GRCh38
Transcripts
NM_012193.3 NR_120591.1 NR_120592.1

Gene Symbol
PRSS23
Entrez Gene ID
11098
Gene Name
protease, serine 23
Gene Aliases
SIG13, SPUVE, ZSIG13
UniGene
Hs.25338
Gene Chromosome Location
Chr.11: 86791059-86952910 on Build GRCh38
Transcripts
NM_012193.3 NR_120591.1 NR_120592.1

SNP Detail

SNP ID Chromosome Location
rs534530796 Chr.11: 86946042-86946042 on Build GRCh38
rs906873759 Chr.11: 86945697-86945697 on Build GRCh38
rs371128840 Chr.11: 86945859-86945860 on Build GRCh38
rs58750277 Chr.11: 86945691-86945691 on Build GRCh38
rs746033036 Chr.11: 86945960-86945960 on Build GRCh38
rs991997917 Chr.11: 86945931-86945931 on Build GRCh38
rs1022452842 Chr.11: 86945791-86945791 on Build GRCh38
rs184657785 Chr.11: 86945689-86945689 on Build GRCh38
rs562688071 Chr.11: 86945833-86945833 on Build GRCh38
rs952198681 Chr.11: 86945915-86945915 on Build GRCh38
rs549067034 Chr.11: 86945796-86945796 on Build GRCh38
rs1052816823 Chr.11: 86946008-86946008 on Build GRCh38
rs371044823 Chr.11: 86945998-86945998 on Build GRCh38
rs917640757 Chr.11: 86945933-86945933 on Build GRCh38
rs984746219 Chr.11: 86945789-86945789 on Build GRCh38
rs189105376 Chr.11: 86945699-86945699 on Build GRCh38
rs1041205066 Chr.11: 86945978-86945978 on Build GRCh38
rs376559208 Chr.11: 86945965-86945965 on Build GRCh38
rs966459391 Chr.11: 86945843-86945843 on Build GRCh38
rs748157221 Chr.11: 86945763-86945763 on Build GRCh38
rs1008311229 Chr.11: 86945973-86945973 on Build GRCh38
rs953592538 Chr.11: 86945798-86945798 on Build GRCh38
rs15135 Chr.11: 86945761-86945761 on Build GRCh38
rs919324557 Chr.11: 86945894-86945894 on Build GRCh38
rs948655774 Chr.11: 86945612-86945612 on Build GRCh38
rs1007774368 Chr.11: 86945799-86945799 on Build GRCh38
rs79807722 Chr.11: 86945774-86945774 on Build GRCh38
rs998583849 Chr.11: 86945726-86945726 on Build GRCh38
rs551786923 Chr.11: 86945952-86945952 on Build GRCh38
rs781001099 Chr.11: 86945877-86945877 on Build GRCh38
rs1045689207 Chr.11: 86945974-86945974 on Build GRCh38
rs181367055 Chr.11: 86945946-86945946 on Build GRCh38
rs924996226 Chr.11: 86945953-86945953 on Build GRCh38
rs972116807 Chr.11: 86945856-86945856 on Build GRCh38
rs886048715 Chr.11: 86945892-86945892 on Build GRCh38
rs915748571 Chr.11: 86945961-86945961 on Build GRCh38
rs1019632481 Chr.11: 86945644-86945644 on Build GRCh38
rs892869123 Chr.11: 86946025-86946025 on Build GRCh38
rs757264981 Chr.11: 86945766-86945766 on Build GRCh38
rs778991563 Chr.11: 86945603-86945603 on Build GRCh38
rs961443611 Chr.11: 86945685-86945685 on Build GRCh38
rs934302736 Chr.11: 86946002-86946002 on Build GRCh38

View Full Product Details