Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 86950865-86951362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FZD4
Entrez Gene ID
8322
Gene Name
frizzled class receptor 4
Gene Aliases
CD344, EVR1, FEVR, FZD4S, Fz-4, Fz4, FzE4, GPCR, hFz4
UniGene
Hs.591968
Gene Chromosome Location
Chr.11: 86945675-86955398 on Build GRCh38
Transcripts
NM_012193.3 NR_120591.1 NR_120592.1

Gene Symbol
PRSS23
Entrez Gene ID
11098
Gene Name
protease, serine 23
Gene Aliases
SIG13, SPUVE, ZSIG13
UniGene
Hs.25338
Gene Chromosome Location
Chr.11: 86791059-86952910 on Build GRCh38
Transcripts
NM_012193.3 NR_120591.1 NR_120592.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1676361 c.1474G>C p.G492R Chr.11: 86951282-86951282 on Build GRCh38
1579414 c.1469C>T p.T490I Chr.11: 86951287-86951287 on Build GRCh38
932768 c.1509G>A p.T503T Chr.11: 86951247-86951247 on Build GRCh38
4817769 c.1563G>C p.K521N Chr.11: 86951193-86951193 on Build GRCh38
244630 c.1518G>C p.K506N Chr.11: 86951238-86951238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755528979 Chr.11: 86951196-86951196 on Build GRCh38
rs749958709 Chr.11: 86951278-86951278 on Build GRCh38
rs781192701 Chr.11: 86951333-86951333 on Build GRCh38
rs528411909 Chr.11: 86950969-86950969 on Build GRCh38
rs61749246 Chr.11: 86951140-86951140 on Build GRCh38
rs1010167604 Chr.11: 86951315-86951315 on Build GRCh38
rs202085996 Chr.11: 86951133-86951133 on Build GRCh38
rs1049305053 Chr.11: 86950987-86950987 on Build GRCh38
rs752990226 Chr.11: 86951297-86951297 on Build GRCh38
rs769786110 Chr.11: 86951239-86951239 on Build GRCh38
rs7940477 Chr.11: 86951072-86951072 on Build GRCh38
rs1009682639 Chr.11: 86951075-86951075 on Build GRCh38
rs187559867 Chr.11: 86950893-86950893 on Build GRCh38
rs777938879 Chr.11: 86951308-86951308 on Build GRCh38
rs80358301 Chr.11: 86951272-86951277 on Build GRCh38
rs80358303 Chr.11: 86951254-86951255 on Build GRCh38
rs747068280 Chr.11: 86951221-86951221 on Build GRCh38
rs771038159 Chr.11: 86951226-86951226 on Build GRCh38
rs3740661 Chr.11: 86950939-86950939 on Build GRCh38
rs372852310 Chr.11: 86951330-86951330 on Build GRCh38
rs563441127 Chr.11: 86950904-86950904 on Build GRCh38
rs772131787 Chr.11: 86951194-86951195 on Build GRCh38
rs756626640 Chr.11: 86951096-86951096 on Build GRCh38
rs367815750 Chr.11: 86951104-86951104 on Build GRCh38
rs201914515 Chr.11: 86951273-86951273 on Build GRCh38
rs770841790 Chr.11: 86951166-86951166 on Build GRCh38
rs199577384 Chr.11: 86951189-86951189 on Build GRCh38
rs761378697 Chr.11: 86951256-86951256 on Build GRCh38
rs530994733 Chr.11: 86951014-86951014 on Build GRCh38
rs3740662 Chr.11: 86950901-86950901 on Build GRCh38
rs80358298 Chr.11: 86951293-86951293 on Build GRCh38
rs763843440 Chr.11: 86951183-86951183 on Build GRCh38
rs530658595 Chr.11: 86950936-86950936 on Build GRCh38
rs747158245 Chr.11: 86951153-86951153 on Build GRCh38
rs1036091905 Chr.11: 86951162-86951162 on Build GRCh38
rs377030377 Chr.11: 86951329-86951330 on Build GRCh38
rs372257164 Chr.11: 86951141-86951141 on Build GRCh38
rs577048598 Chr.11: 86951336-86951336 on Build GRCh38
rs916863095 Chr.11: 86951084-86951084 on Build GRCh38
rs544964220 Chr.11: 86951195-86951195 on Build GRCh38
rs982170218 Chr.11: 86951115-86951115 on Build GRCh38
rs550787317 Chr.11: 86951015-86951015 on Build GRCh38
rs200205053 Chr.11: 86951102-86951102 on Build GRCh38
rs1008586902 Chr.11: 86950943-86950943 on Build GRCh38
rs886048734 Chr.11: 86951215-86951215 on Build GRCh38
rs73531344 Chr.11: 86951248-86951248 on Build GRCh38
rs566786953 Chr.11: 86951175-86951175 on Build GRCh38
rs764902681 Chr.11: 86951176-86951176 on Build GRCh38
rs886048732 Chr.11: 86950988-86950988 on Build GRCh38
rs148448468 Chr.11: 86951186-86951186 on Build GRCh38
rs1050752347 Chr.11: 86951279-86951279 on Build GRCh38
rs377696818 Chr.11: 86951007-86951007 on Build GRCh38
rs532010386 Chr.11: 86950924-86950924 on Build GRCh38
rs78623095 Chr.11: 86951329-86951329 on Build GRCh38
rs780777696 Chr.11: 86951107-86951107 on Build GRCh38
rs886048733 Chr.11: 86951136-86951136 on Build GRCh38
rs778027186 Chr.11: 86951217-86951217 on Build GRCh38
rs897468308 Chr.11: 86950974-86950974 on Build GRCh38
rs192151920 Chr.11: 86951250-86951250 on Build GRCh38
rs146277634 Chr.11: 86951177-86951177 on Build GRCh38
rs80358300 Chr.11: 86951266-86951266 on Build GRCh38
rs745745495 Chr.11: 86951233-86951233 on Build GRCh38
rs142618839 Chr.11: 86951205-86951205 on Build GRCh38
rs775081751 Chr.11: 86951247-86951247 on Build GRCh38
rs80358302 Chr.11: 86951258-86951258 on Build GRCh38
rs150947482 Chr.11: 86951236-86951236 on Build GRCh38
rs371482043 Chr.11: 86951106-86951106 on Build GRCh38
rs780102146 Chr.11: 86951339-86951339 on Build GRCh38
rs368527723 Chr.11: 86951249-86951249 on Build GRCh38
rs952963687 Chr.11: 86951034-86951034 on Build GRCh38
rs758540332 Chr.11: 86951216-86951216 on Build GRCh38
rs994526427 Chr.11: 86950977-86950977 on Build GRCh38
rs370690635 Chr.11: 86951121-86951121 on Build GRCh38
rs745965975 Chr.11: 86951335-86951335 on Build GRCh38
rs1035232321 Chr.11: 86951020-86951020 on Build GRCh38
rs772349259 Chr.11: 86951146-86951146 on Build GRCh38
rs377431110 Chr.11: 86951105-86951105 on Build GRCh38
rs80358304 Chr.11: 86951243-86951243 on Build GRCh38
rs12574787 Chr.11: 86951094-86951094 on Build GRCh38
rs563169065 Chr.11: 86951301-86951301 on Build GRCh38
rs574398264 Chr.11: 86951159-86951159 on Build GRCh38
rs756889974 Chr.11: 86951188-86951188 on Build GRCh38
rs80358299 Chr.11: 86951282-86951282 on Build GRCh38
rs779086471 Chr.11: 86951203-86951203 on Build GRCh38
rs373872905 Chr.11: 86951125-86951125 on Build GRCh38
rs751738021 Chr.11: 86951316-86951316 on Build GRCh38
rs201256460 Chr.11: 86951167-86951167 on Build GRCh38
rs759890542 Chr.11: 86951281-86951281 on Build GRCh38
rs765834466 Chr.11: 86951288-86951288 on Build GRCh38
rs548377194 Chr.11: 86950970-86950970 on Build GRCh38

View Full Product Details