Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108133649-108134153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAT1
Entrez Gene ID
38
Gene Name
acetyl-CoA acetyltransferase 1
Gene Aliases
ACAT, MAT, T2, THIL
UniGene
Hs.232375
Gene Chromosome Location
Chr.11: 108121531-108148168 on Build GRCh38
Transcripts
NM_000019.3 XM_006718835.3 XM_017017681.1 XM_017017682.1 XM_017017683.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1350694 c.166T>C p.L56L Chr.11: 108133865-108133865 on Build GRCh38
6591832 c.235G>A p.A79T Chr.11: 108133934-108133934 on Build GRCh38
1350693 c.162delT p.L56fs*1 Chr.11: 108133861-108133861 on Build GRCh38
5929256 c.209T>A p.I70N Chr.11: 108133908-108133908 on Build GRCh38
6591831 c.174C>T p.S58S Chr.11: 108133873-108133873 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151080188 Chr.11: 108133854-108133854 on Build GRCh38
rs973030476 Chr.11: 108133676-108133676 on Build GRCh38
rs150258264 Chr.11: 108134096-108134096 on Build GRCh38
rs776870798 Chr.11: 108133791-108133791 on Build GRCh38
rs764707694 Chr.11: 108134112-108134113 on Build GRCh38
rs757128075 Chr.11: 108133870-108133870 on Build GRCh38
rs12365364 Chr.11: 108133960-108133960 on Build GRCh38
rs949493567 Chr.11: 108133695-108133695 on Build GRCh38
rs565347258 Chr.11: 108133693-108133693 on Build GRCh38
rs763748117 Chr.11: 108133982-108133982 on Build GRCh38
rs553246561 Chr.11: 108133741-108133741 on Build GRCh38
rs758176568 Chr.11: 108133912-108133912 on Build GRCh38
rs777222104 Chr.11: 108133733-108133734 on Build GRCh38
rs984124680 Chr.11: 108133730-108133730 on Build GRCh38
rs147018814 Chr.11: 108133873-108133873 on Build GRCh38
rs781366330 Chr.11: 108133935-108133935 on Build GRCh38
rs200394877 Chr.11: 108133993-108133993 on Build GRCh38
rs541926186 Chr.11: 108133777-108133777 on Build GRCh38
rs768829887 Chr.11: 108133788-108133788 on Build GRCh38
rs545826042 Chr.11: 108133839-108133839 on Build GRCh38
rs779565865 Chr.11: 108133848-108133848 on Build GRCh38
rs1046898897 Chr.11: 108133824-108133824 on Build GRCh38
rs779758622 Chr.11: 108133917-108133917 on Build GRCh38
rs772848693 Chr.11: 108133928-108133928 on Build GRCh38
rs763375578 Chr.11: 108133837-108133837 on Build GRCh38
rs578074515 Chr.11: 108134083-108134083 on Build GRCh38
rs771278537 Chr.11: 108133961-108133961 on Build GRCh38
rs997243762 Chr.11: 108133944-108133944 on Build GRCh38
rs753349314 Chr.11: 108133856-108133856 on Build GRCh38
rs924416304 Chr.11: 108133755-108133755 on Build GRCh38
rs760268967 Chr.11: 108133851-108133851 on Build GRCh38
rs773917391 Chr.11: 108133828-108133828 on Build GRCh38
rs934509994 Chr.11: 108134095-108134095 on Build GRCh38
rs113132606 Chr.11: 108133701-108133701 on Build GRCh38
rs776321178 Chr.11: 108133983-108133983 on Build GRCh38
rs901575634 Chr.11: 108133919-108133919 on Build GRCh38
rs563829181 Chr.11: 108133795-108133795 on Build GRCh38
rs748228668 Chr.11: 108133937-108133937 on Build GRCh38
rs199918672 Chr.11: 108133954-108133954 on Build GRCh38
rs770069051 Chr.11: 108133948-108133948 on Build GRCh38
rs747201837 Chr.11: 108133924-108133924 on Build GRCh38
rs780318835 Chr.11: 108133773-108133773 on Build GRCh38
rs189471668 Chr.11: 108133761-108133761 on Build GRCh38
rs774611589 Chr.11: 108133962-108133962 on Build GRCh38
rs180774428 Chr.11: 108133977-108133977 on Build GRCh38
rs794727475 Chr.11: 108133902-108133902 on Build GRCh38
rs115763401 Chr.11: 108133838-108133838 on Build GRCh38
rs990815700 Chr.11: 108134004-108134004 on Build GRCh38
rs770440667 Chr.11: 108133805-108133805 on Build GRCh38
rs141054498 Chr.11: 108134011-108134011 on Build GRCh38
rs1042779739 Chr.11: 108133985-108133985 on Build GRCh38
rs7931681 Chr.11: 108134071-108134071 on Build GRCh38
rs970713510 Chr.11: 108134069-108134069 on Build GRCh38
rs891679038 Chr.11: 108133969-108133969 on Build GRCh38
rs771819781 Chr.11: 108133772-108133772 on Build GRCh38
rs572642909 Chr.11: 108133746-108133751 on Build GRCh38

View Full Product Details