Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111784306-111784806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG9
Entrez Gene ID
79796
Gene Name
ALG9, alpha-1,2-mannosyltransferase
Gene Aliases
CDG1L, DIBD1, GIKANIS, LOH11CR1J
UniGene
Hs.745155
Gene Chromosome Location
Chr.11: 111776096-111871581 on Build GRCh38
Transcripts
NM_001077690.1 NM_001077691.1 NM_001077692.1 NM_024740.2 XM_005277723.4 XM_005277724.4 XM_017018314.1 XM_017018315.1 XM_017018316.1 XM_017018317.1 XM_017018319.1 XM_017018320.1 XM_017018321.1 XM_017018322.1 XM_017018325.1 XM_017018326.1

SNP Detail

SNP ID Chromosome Location
rs919642234 Chr.11: 111784535-111784535 on Build GRCh38
rs996108893 Chr.11: 111784455-111784455 on Build GRCh38
rs891998484 Chr.11: 111784427-111784427 on Build GRCh38
rs150703313 Chr.11: 111784718-111784718 on Build GRCh38
rs116003998 Chr.11: 111784726-111784726 on Build GRCh38
rs558800800 Chr.11: 111784638-111784638 on Build GRCh38
rs892055215 Chr.11: 111784588-111784588 on Build GRCh38
rs776627555 Chr.11: 111784758-111784758 on Build GRCh38
rs1044098206 Chr.11: 111784335-111784335 on Build GRCh38
rs574882570 Chr.11: 111784539-111784539 on Build GRCh38
rs186477879 Chr.11: 111784675-111784675 on Build GRCh38
rs117091304 Chr.11: 111784773-111784773 on Build GRCh38
rs782103178 Chr.11: 111784696-111784696 on Build GRCh38
rs541176886 Chr.11: 111784715-111784716 on Build GRCh38
rs553100335 Chr.11: 111784453-111784453 on Build GRCh38
rs556550557 Chr.11: 111784489-111784489 on Build GRCh38
rs938910845 Chr.11: 111784574-111784574 on Build GRCh38
rs972600588 Chr.11: 111784515-111784515 on Build GRCh38
rs886047676 Chr.11: 111784631-111784631 on Build GRCh38
rs772136205 Chr.11: 111784565-111784565 on Build GRCh38
rs541280594 Chr.11: 111784673-111784673 on Build GRCh38
rs1003150731 Chr.11: 111784364-111784364 on Build GRCh38
rs568087425 Chr.11: 111784454-111784454 on Build GRCh38
rs45620134 Chr.11: 111784549-111784549 on Build GRCh38
rs782038696 Chr.11: 111784548-111784548 on Build GRCh38
rs577399597 Chr.11: 111784667-111784667 on Build GRCh38
rs961332088 Chr.11: 111784742-111784742 on Build GRCh38
rs904586525 Chr.11: 111784662-111784662 on Build GRCh38
rs574839296 Chr.11: 111784677-111784677 on Build GRCh38
rs906090588 Chr.11: 111784352-111784352 on Build GRCh38
rs961465692 Chr.11: 111784506-111784506 on Build GRCh38
rs181231005 Chr.11: 111784420-111784420 on Build GRCh38
rs375953398 Chr.11: 111784417-111784417 on Build GRCh38
rs782776196 Chr.11: 111784626-111784626 on Build GRCh38
rs886047675 Chr.11: 111784386-111784387 on Build GRCh38
rs953953913 Chr.11: 111784466-111784466 on Build GRCh38
rs113409957 Chr.11: 111784586-111784586 on Build GRCh38
rs986716229 Chr.11: 111784474-111784474 on Build GRCh38
rs367569393 Chr.11: 111784710-111784710 on Build GRCh38
rs912394074 Chr.11: 111784476-111784476 on Build GRCh38
rs1022094155 Chr.11: 111784451-111784451 on Build GRCh38
rs781949750 Chr.11: 111784446-111784446 on Build GRCh38
rs542301482 Chr.11: 111784697-111784697 on Build GRCh38
rs535132621 Chr.11: 111784457-111784457 on Build GRCh38
rs886047677 Chr.11: 111784717-111784718 on Build GRCh38
rs782378345 Chr.11: 111784445-111784445 on Build GRCh38
rs563719515 Chr.11: 111784717-111784717 on Build GRCh38
rs1043246533 Chr.11: 111784652-111784652 on Build GRCh38
rs926979351 Chr.11: 111784568-111784568 on Build GRCh38

View Full Product Details