Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112088604-112089107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHD
Entrez Gene ID
6392
Gene Name
succinate dehydrogenase complex subunit D
Gene Aliases
CBT1, CII-4, CWS3, PGL, PGL1, QPs3, SDH4, cybS
UniGene
Hs.356270
Gene Chromosome Location
Chr.11: 112086824-112095801 on Build GRCh38
Transcripts
NM_001276503.1 NM_001276504.1 NM_001276506.1 NM_003002.3 NR_077060.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
466328 c.227T>A p.L76H Chr.11: 112088924-112088924 on Build GRCh38
923089 c.205G>A p.E69K Chr.11: 112088902-112088902 on Build GRCh38
4883438 c.283C>A p.L95M Chr.11: 112088980-112088980 on Build GRCh38
1351279 c.200C>T p.T67I Chr.11: 112088897-112088897 on Build GRCh38
6937860 c.190C>T p.L64F Chr.11: 112088887-112088887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11547893 Chr.11: 112088892-112088892 on Build GRCh38
rs104894302 Chr.11: 112089002-112089002 on Build GRCh38
rs765102002 Chr.11: 112088965-112088965 on Build GRCh38
rs1007945786 Chr.11: 112088802-112088802 on Build GRCh38
rs879404309 Chr.11: 112088842-112088842 on Build GRCh38
rs1056959364 Chr.11: 112088705-112088705 on Build GRCh38
rs534062584 Chr.11: 112088726-112088726 on Build GRCh38
rs199517389 Chr.11: 112088952-112088952 on Build GRCh38
rs773195941 Chr.11: 112089062-112089062 on Build GRCh38
rs755047928 Chr.11: 112088906-112088906 on Build GRCh38
rs756533450 Chr.11: 112088987-112088987 on Build GRCh38
rs764869982 Chr.11: 112088874-112088874 on Build GRCh38
rs755475561 Chr.11: 112089029-112089029 on Build GRCh38
rs202198133 Chr.11: 112088902-112088902 on Build GRCh38
rs121908983 Chr.11: 112088974-112088976 on Build GRCh38
rs387906358 Chr.11: 112088888-112088889 on Build GRCh38
rs780226013 Chr.11: 112088891-112088891 on Build GRCh38
rs904090433 Chr.11: 112088761-112088761 on Build GRCh38
rs758499440 Chr.11: 112088881-112088881 on Build GRCh38
rs148634289 Chr.11: 112088925-112088925 on Build GRCh38
rs1010167356 Chr.11: 112088723-112088723 on Build GRCh38
rs764191089 Chr.11: 112088855-112088855 on Build GRCh38
rs1001139895 Chr.11: 112088764-112088764 on Build GRCh38
rs786202403 Chr.11: 112089001-112089001 on Build GRCh38
rs878854591 Chr.11: 112088939-112088939 on Build GRCh38
rs189698193 Chr.11: 112088650-112088650 on Build GRCh38
rs200354034 Chr.11: 112089048-112089048 on Build GRCh38
rs980027847 Chr.11: 112088948-112088948 on Build GRCh38
rs575262156 Chr.11: 112088940-112088940 on Build GRCh38
rs786205436 Chr.11: 112088972-112088972 on Build GRCh38
rs1053063141 Chr.11: 112088706-112088706 on Build GRCh38
rs9919624 Chr.11: 112088838-112088838 on Build GRCh38
rs142135772 Chr.11: 112088975-112088975 on Build GRCh38
rs772241591 Chr.11: 112088773-112088773 on Build GRCh38
rs750688879 Chr.11: 112088877-112088877 on Build GRCh38
rs557454864 Chr.11: 112088996-112088996 on Build GRCh38
rs749657880 Chr.11: 112088994-112088994 on Build GRCh38
rs370165653 Chr.11: 112088945-112088945 on Build GRCh38
rs200486533 Chr.11: 112088915-112088915 on Build GRCh38
rs1027987517 Chr.11: 112088765-112088765 on Build GRCh38
rs767170188 Chr.11: 112088830-112088830 on Build GRCh38
rs199754684 Chr.11: 112088978-112088978 on Build GRCh38
rs61734352 Chr.11: 112089009-112089009 on Build GRCh38
rs779988676 Chr.11: 112088652-112088652 on Build GRCh38
rs761363469 Chr.11: 112089027-112089027 on Build GRCh38
rs202239399 Chr.11: 112088944-112088944 on Build GRCh38
rs769657742 Chr.11: 112089058-112089058 on Build GRCh38
rs537852530 Chr.11: 112088664-112088664 on Build GRCh38
rs569148522 Chr.11: 112088659-112088659 on Build GRCh38
rs9919552 Chr.11: 112088901-112088901 on Build GRCh38
rs749263280 Chr.11: 112088718-112088718 on Build GRCh38
rs766656463 Chr.11: 112088967-112088967 on Build GRCh38
rs953724919 Chr.11: 112088784-112088784 on Build GRCh38
rs919536825 Chr.11: 112088853-112088853 on Build GRCh38
rs938094457 Chr.11: 112088687-112088687 on Build GRCh38
rs139160150 Chr.11: 112088707-112088707 on Build GRCh38
rs745559875 Chr.11: 112088941-112088941 on Build GRCh38
rs779863605 Chr.11: 112089007-112089007 on Build GRCh38
rs200186743 Chr.11: 112088846-112088846 on Build GRCh38
rs878854590 Chr.11: 112088870-112088870 on Build GRCh38
rs80338844 Chr.11: 112088939-112088939 on Build GRCh38
rs748131895 Chr.11: 112088776-112088776 on Build GRCh38
rs11547899 Chr.11: 112088883-112088883 on Build GRCh38
rs781182616 Chr.11: 112088979-112088979 on Build GRCh38
rs786203067 Chr.11: 112088995-112088998 on Build GRCh38
rs551434354 Chr.11: 112088694-112088694 on Build GRCh38
rs1019114366 Chr.11: 112088779-112088779 on Build GRCh38
rs199645781 Chr.11: 112089019-112089019 on Build GRCh38
rs1049039919 Chr.11: 112088675-112088675 on Build GRCh38
rs886038522 Chr.11: 112089031-112089031 on Build GRCh38
rs918007343 Chr.11: 112089081-112089081 on Build GRCh38
rs926650596 Chr.11: 112088685-112088685 on Build GRCh38
rs761615413 Chr.11: 112088961-112088961 on Build GRCh38
rs766754598 Chr.11: 112088848-112088848 on Build GRCh38
rs556828083 Chr.11: 112088911-112088911 on Build GRCh38
rs752036767 Chr.11: 112088973-112088974 on Build GRCh38
rs966219481 Chr.11: 112088836-112088836 on Build GRCh38
rs945509144 Chr.11: 112089083-112089083 on Build GRCh38
rs1022692450 Chr.11: 112088758-112088758 on Build GRCh38
rs200255724 Chr.11: 112088966-112088966 on Build GRCh38
rs961214333 Chr.11: 112088839-112088839 on Build GRCh38
rs777963699 Chr.11: 112088927-112088927 on Build GRCh38
rs200980609 Chr.11: 112088990-112088990 on Build GRCh38
rs201075295 Chr.11: 112088832-112088832 on Build GRCh38
rs753703004 Chr.11: 112088861-112088861 on Build GRCh38
rs80338846 Chr.11: 112088981-112088981 on Build GRCh38
rs748545223 Chr.11: 112088914-112088914 on Build GRCh38
rs952278127 Chr.11: 112088857-112088857 on Build GRCh38
rs80338845 Chr.11: 112088971-112088971 on Build GRCh38
rs201163618 Chr.11: 112089023-112089023 on Build GRCh38
rs757084564 Chr.11: 112088863-112088863 on Build GRCh38
rs149340379 Chr.11: 112088757-112088757 on Build GRCh38

View Full Product Details