Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1294640-1295144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAP3
Entrez Gene ID
116983
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
CENTB5
UniGene
Hs.535257
Gene Chromosome Location
Chr.1: 1292384-1307925 on Build GRCh38
Transcripts
NM_030649.2 XM_005244715.2 XM_005244717.3 XM_011540606.2 XM_011540607.1 XM_011540608.2 XM_011540609.2 XM_017000233.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7304336 c.1860G>A p.S620S Chr.1: 1294770-1294770 on Build GRCh38
4615432 c.1841_1842insG p.S615fs*22 Chr.1: 1294788-1294789 on Build GRCh38
5453882 c.1814-5T>C p.? Chr.1: 1294821-1294821 on Build GRCh38
1194900 c.1906G>A p.E636K Chr.1: 1294724-1294724 on Build GRCh38
5673350 c.1848G>T p.S616S Chr.1: 1294782-1294782 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs186687859 Chr.1: 1295027-1295027 on Build GRCh38
rs1043743667 Chr.1: 1294958-1294958 on Build GRCh38
rs546483206 Chr.1: 1294950-1294950 on Build GRCh38
rs763875739 Chr.1: 1294673-1294683 on Build GRCh38
rs765039470 Chr.1: 1294806-1294806 on Build GRCh38
rs946530559 Chr.1: 1294768-1294779 on Build GRCh38
rs561209942 Chr.1: 1294832-1294832 on Build GRCh38
rs905166204 Chr.1: 1294969-1294969 on Build GRCh38
rs757689533 Chr.1: 1294689-1294689 on Build GRCh38
rs1010903980 Chr.1: 1294730-1294730 on Build GRCh38
rs1050115812 Chr.1: 1294878-1294878 on Build GRCh38
rs759270107 Chr.1: 1294779-1294779 on Build GRCh38
rs896253500 Chr.1: 1295104-1295104 on Build GRCh38
rs565771451 Chr.1: 1294851-1294851 on Build GRCh38
rs1043439364 Chr.1: 1294770-1294770 on Build GRCh38
rs115280881 Chr.1: 1294934-1294934 on Build GRCh38
rs944418863 Chr.1: 1295065-1295065 on Build GRCh38
rs1002320692 Chr.1: 1294986-1294990 on Build GRCh38
rs774277341 Chr.1: 1294734-1294734 on Build GRCh38
rs995461007 Chr.1: 1294939-1294939 on Build GRCh38
rs751128894 Chr.1: 1294677-1294677 on Build GRCh38
rs534355589 Chr.1: 1295081-1295081 on Build GRCh38
rs907784383 Chr.1: 1294823-1294823 on Build GRCh38
rs560608350 Chr.1: 1294981-1294981 on Build GRCh38
rs931768739 Chr.1: 1294854-1294854 on Build GRCh38
rs1011538945 Chr.1: 1294955-1294955 on Build GRCh38
rs896272114 Chr.1: 1294855-1294855 on Build GRCh38
rs912684493 Chr.1: 1294717-1294717 on Build GRCh38
rs979089322 Chr.1: 1295038-1295038 on Build GRCh38
rs762569603 Chr.1: 1294828-1294828 on Build GRCh38
rs386627472 Chr.1: 1295037-1295039 on Build GRCh38
rs1019710071 Chr.1: 1294980-1294980 on Build GRCh38
rs369722813 Chr.1: 1295107-1295107 on Build GRCh38
rs544493485 Chr.1: 1294761-1294761 on Build GRCh38
rs931848916 Chr.1: 1294716-1294716 on Build GRCh38
rs892720109 Chr.1: 1294952-1294952 on Build GRCh38
rs781755835 Chr.1: 1294697-1294697 on Build GRCh38
rs1002258548 Chr.1: 1294750-1294750 on Build GRCh38
rs1272959 Chr.1: 1295039-1295039 on Build GRCh38
rs1015139145 Chr.1: 1295021-1295021 on Build GRCh38
rs1027346324 Chr.1: 1295035-1295035 on Build GRCh38
rs772154663 Chr.1: 1294913-1294913 on Build GRCh38
rs371287468 Chr.1: 1295046-1295046 on Build GRCh38
rs1028321899 Chr.1: 1294945-1294945 on Build GRCh38
rs756795451 Chr.1: 1294866-1294866 on Build GRCh38
rs1052623048 Chr.1: 1294693-1294693 on Build GRCh38
rs373987414 Chr.1: 1294736-1294736 on Build GRCh38
rs1749953 Chr.1: 1295028-1295028 on Build GRCh38
rs987256141 Chr.1: 1294966-1294966 on Build GRCh38
rs760373044 Chr.1: 1295010-1295010 on Build GRCh38
rs12041067 Chr.1: 1295060-1295060 on Build GRCh38
rs1056584989 Chr.1: 1295077-1295077 on Build GRCh38
rs377269757 Chr.1: 1294755-1294755 on Build GRCh38
rs780370981 Chr.1: 1294718-1294718 on Build GRCh38
rs770293758 Chr.1: 1294714-1294714 on Build GRCh38
rs954305192 Chr.1: 1294672-1294672 on Build GRCh38
rs572853937 Chr.1: 1294663-1294663 on Build GRCh38
rs912089788 Chr.1: 1294909-1294909 on Build GRCh38
rs369609493 Chr.1: 1294731-1294731 on Build GRCh38
rs199515600 Chr.1: 1295036-1295037 on Build GRCh38
rs74045415 Chr.1: 1295037-1295037 on Build GRCh38
rs376812946 Chr.1: 1294709-1294709 on Build GRCh38
rs969505947 Chr.1: 1294738-1294738 on Build GRCh38
rs532514802 Chr.1: 1295016-1295016 on Build GRCh38
rs564887554 Chr.1: 1294754-1294754 on Build GRCh38
rs939829958 Chr.1: 1295110-1295110 on Build GRCh38
rs748320969 Chr.1: 1294749-1294749 on Build GRCh38
rs768965821 Chr.1: 1294732-1294732 on Build GRCh38
rs775084747 Chr.1: 1294827-1294827 on Build GRCh38
rs974061363 Chr.1: 1294659-1294659 on Build GRCh38
rs763865321 Chr.1: 1294830-1294830 on Build GRCh38

View Full Product Details