Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1296133-1296621 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAP3
Entrez Gene ID
116983
Gene Name
ArfGAP with coiled-coil, ankyrin repeat and PH domains 3
Gene Aliases
CENTB5
UniGene
Hs.535257
Gene Chromosome Location
Chr.1: 1292384-1307925 on Build GRCh38
Transcripts
NM_030649.2 XM_005244715.2 XM_005244717.3 XM_011540606.2 XM_011540607.1 XM_011540608.2 XM_011540609.2 XM_017000233.1 NR_106784.1

Gene Symbol
MIR6726
Entrez Gene ID
102465434
Gene Name
microRNA 6726
Gene Aliases
hsa-mir-6726
UniGene
Gene Chromosome Location
Chr.1: 1296110-1296170 on Build GRCh38
Transcripts
NM_030649.2 XM_005244715.2 XM_005244717.3 XM_011540606.2 XM_011540607.1 XM_011540608.2 XM_011540609.2 XM_017000233.1 NR_106784.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1181549 c.1227C>A p.H409Q Chr.1: 1296265-1296265 on Build GRCh38
6313877 c.1151C>A p.P384Q Chr.1: 1296611-1296611 on Build GRCh38
5483550 c.1302C>T p.G434G Chr.1: 1296460-1296460 on Build GRCh38
5483551 c.1176C>T p.G392G Chr.1: 1296460-1296460 on Build GRCh38
4465748 c.1273C>G p.L425V Chr.1: 1296219-1296219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751692844 Chr.1: 1296490-1296490 on Build GRCh38
rs761224285 Chr.1: 1296411-1296411 on Build GRCh38
rs930824923 Chr.1: 1296211-1296211 on Build GRCh38
rs776360480 Chr.1: 1296155-1296155 on Build GRCh38
rs570241461 Chr.1: 1296491-1296491 on Build GRCh38
rs866964637 Chr.1: 1296233-1296233 on Build GRCh38
rs771900538 Chr.1: 1296551-1296551 on Build GRCh38
rs759899820 Chr.1: 1296564-1296564 on Build GRCh38
rs533022989 Chr.1: 1296177-1296177 on Build GRCh38
rs1020670103 Chr.1: 1296572-1296572 on Build GRCh38
rs747770222 Chr.1: 1296540-1296540 on Build GRCh38
rs372140455 Chr.1: 1296188-1296188 on Build GRCh38
rs781112703 Chr.1: 1296235-1296235 on Build GRCh38
rs777049311 Chr.1: 1296436-1296436 on Build GRCh38
rs866268389 Chr.1: 1296401-1296401 on Build GRCh38
rs199607738 Chr.1: 1296164-1296164 on Build GRCh38
rs563840123 Chr.1: 1296482-1296482 on Build GRCh38
rs749266601 Chr.1: 1296162-1296162 on Build GRCh38
rs1049278529 Chr.1: 1296212-1296212 on Build GRCh38
rs769656240 Chr.1: 1296242-1296242 on Build GRCh38
rs774159827 Chr.1: 1296379-1296379 on Build GRCh38
rs747611424 Chr.1: 1296396-1296396 on Build GRCh38
rs756557205 Chr.1: 1296201-1296201 on Build GRCh38
rs529560009 Chr.1: 1296556-1296556 on Build GRCh38
rs749100090 Chr.1: 1296325-1296325 on Build GRCh38
rs766737326 Chr.1: 1296283-1296283 on Build GRCh38
rs768287189 Chr.1: 1296332-1296332 on Build GRCh38
rs768250680 Chr.1: 1296252-1296252 on Build GRCh38
rs779856406 Chr.1: 1296498-1296498 on Build GRCh38
rs757297460 Chr.1: 1296594-1296594 on Build GRCh38
rs971429707 Chr.1: 1296349-1296349 on Build GRCh38
rs115482194 Chr.1: 1296403-1296403 on Build GRCh38
rs781059689 Chr.1: 1296303-1296303 on Build GRCh38
rs977958674 Chr.1: 1296398-1296398 on Build GRCh38
rs755461888 Chr.1: 1296508-1296508 on Build GRCh38
rs1749951 Chr.1: 1296276-1296276 on Build GRCh38
rs367638759 Chr.1: 1296185-1296185 on Build GRCh38
rs745605879 Chr.1: 1296240-1296240 on Build GRCh38
rs771736917 Chr.1: 1296402-1296402 on Build GRCh38
rs868453747 Chr.1: 1296434-1296434 on Build GRCh38
rs771026153 Chr.1: 1296415-1296416 on Build GRCh38
rs765389168 Chr.1: 1296446-1296446 on Build GRCh38
rs113099629 Chr.1: 1296531-1296531 on Build GRCh38
rs751641071 Chr.1: 1296206-1296206 on Build GRCh38
rs139600717 Chr.1: 1296207-1296207 on Build GRCh38
rs967183070 Chr.1: 1296302-1296302 on Build GRCh38
rs761408156 Chr.1: 1296268-1296268 on Build GRCh38
rs998200889 Chr.1: 1296571-1296571 on Build GRCh38
rs575866601 Chr.1: 1296247-1296247 on Build GRCh38
rs917199781 Chr.1: 1296339-1296339 on Build GRCh38
rs755219085 Chr.1: 1296288-1296288 on Build GRCh38
rs374708743 Chr.1: 1296176-1296176 on Build GRCh38
rs752707325 Chr.1: 1296295-1296295 on Build GRCh38
rs779071943 Chr.1: 1296294-1296294 on Build GRCh38
rs772986954 Chr.1: 1296165-1296165 on Build GRCh38
rs745771516 Chr.1: 1296306-1296306 on Build GRCh38
rs1041472258 Chr.1: 1296507-1296507 on Build GRCh38
rs762328501 Chr.1: 1296178-1296178 on Build GRCh38
rs764363918 Chr.1: 1296588-1296588 on Build GRCh38
rs1000022315 Chr.1: 1296519-1296519 on Build GRCh38
rs763145875 Chr.1: 1296583-1296583 on Build GRCh38
rs761082675 Chr.1: 1296280-1296280 on Build GRCh38
rs370009609 Chr.1: 1296487-1296487 on Build GRCh38
rs958907731 Chr.1: 1296331-1296331 on Build GRCh38
rs763627196 Chr.1: 1296184-1296184 on Build GRCh38
rs757116358 Chr.1: 1296523-1296523 on Build GRCh38
rs763594739 Chr.1: 1296348-1296348 on Build GRCh38
rs201281051 Chr.1: 1296204-1296204 on Build GRCh38
rs766854185 Chr.1: 1296435-1296435 on Build GRCh38
rs773994190 Chr.1: 1296263-1296263 on Build GRCh38
rs777021496 Chr.1: 1296563-1296563 on Build GRCh38
rs779031901 Chr.1: 1296220-1296220 on Build GRCh38
rs553742772 Chr.1: 1296190-1296190 on Build GRCh38
rs775880236 Chr.1: 1296576-1296576 on Build GRCh38
rs778666650 Chr.1: 1296537-1296537 on Build GRCh38
rs894789044 Chr.1: 1296223-1296223 on Build GRCh38
rs527673390 Chr.1: 1296350-1296350 on Build GRCh38
rs1040631059 Chr.1: 1296483-1296483 on Build GRCh38
rs754207517 Chr.1: 1296285-1296285 on Build GRCh38
rs762644301 Chr.1: 1296251-1296251 on Build GRCh38
rs113414019 Chr.1: 1296364-1296364 on Build GRCh38
rs967828190 Chr.1: 1296397-1296397 on Build GRCh38
rs765624546 Chr.1: 1296565-1296565 on Build GRCh38
rs377421242 Chr.1: 1296170-1296170 on Build GRCh38
rs199873685 Chr.1: 1296321-1296321 on Build GRCh38
rs370373247 Chr.1: 1296172-1296172 on Build GRCh38
rs758488003 Chr.1: 1296301-1296301 on Build GRCh38
rs368868926 Chr.1: 1296589-1296589 on Build GRCh38
rs1020366330 Chr.1: 1296388-1296388 on Build GRCh38
rs750987038 Chr.1: 1296277-1296277 on Build GRCh38
rs1000056560 Chr.1: 1296315-1296315 on Build GRCh38
rs748289109 Chr.1: 1296232-1296232 on Build GRCh38
rs762353431 Chr.1: 1296335-1296335 on Build GRCh38
rs375874447 Chr.1: 1296444-1296444 on Build GRCh38
rs201882356 Chr.1: 1296198-1296198 on Build GRCh38
rs371372231 Chr.1: 1296505-1296505 on Build GRCh38
rs533685225 Chr.1: 1296451-1296451 on Build GRCh38
rs746568332 Chr.1: 1296555-1296555 on Build GRCh38
rs769667618 Chr.1: 1296316-1296316 on Build GRCh38
rs772836801 Chr.1: 1296552-1296552 on Build GRCh38
rs746921654 Chr.1: 1296166-1296166 on Build GRCh38

View Full Product Details