Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130121526-130122030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APLP2
Entrez Gene ID
334
Gene Name
amyloid beta precursor like protein 2
Gene Aliases
APLP-2, APPH, APPL2, CDEBP
UniGene
Hs.370247
Gene Chromosome Location
Chr.11: 130069821-130144811 on Build GRCh38
Transcripts
NM_001142276.1 NM_001142277.1 NM_001142278.1 NM_001243299.1 NM_001642.2 NR_024515.1 NR_024516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7270090 c.702T>C p.D234D Chr.11: 130121799-130121799 on Build GRCh38
139936 c.638C>T p.S213L Chr.11: 130121735-130121735 on Build GRCh38
3718433 c.660T>G p.D220E Chr.11: 130121757-130121757 on Build GRCh38
318728 c.684T>G p.D228E Chr.11: 130121781-130121781 on Build GRCh38
7183393 c.613C>G p.Q205E Chr.11: 130121710-130121710 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs76279939 Chr.11: 130121763-130121763 on Build GRCh38
rs200485579 Chr.11: 130121720-130121720 on Build GRCh38
rs753704296 Chr.11: 130121591-130121591 on Build GRCh38
rs760211992 Chr.11: 130121740-130121745 on Build GRCh38
rs1030849883 Chr.11: 130121964-130121964 on Build GRCh38
rs771288672 Chr.11: 130121840-130121841 on Build GRCh38
rs564914445 Chr.11: 130121969-130121969 on Build GRCh38
rs754360304 Chr.11: 130121808-130121808 on Build GRCh38
rs748662145 Chr.11: 130121623-130121623 on Build GRCh38
rs779885112 Chr.11: 130121758-130121775 on Build GRCh38
rs368717793 Chr.11: 130121857-130121857 on Build GRCh38
rs766107227 Chr.11: 130121563-130121563 on Build GRCh38
rs771665731 Chr.11: 130121840-130121840 on Build GRCh38
rs753252729 Chr.11: 130121872-130121872 on Build GRCh38
rs981203966 Chr.11: 130121847-130121847 on Build GRCh38
rs755122715 Chr.11: 130121608-130121608 on Build GRCh38
rs537248071 Chr.11: 130121658-130121658 on Build GRCh38
rs756921141 Chr.11: 130121740-130121757 on Build GRCh38
rs750228847 Chr.11: 130121752-130121757 on Build GRCh38
rs763790091 Chr.11: 130121589-130121589 on Build GRCh38
rs377280332 Chr.11: 130121567-130121567 on Build GRCh38
rs767371773 Chr.11: 130121577-130121577 on Build GRCh38
rs979308203 Chr.11: 130121664-130121664 on Build GRCh38
rs773462397 Chr.11: 130121799-130121801 on Build GRCh38
rs918416218 Chr.11: 130121565-130121565 on Build GRCh38
rs144449684 Chr.11: 130121839-130121839 on Build GRCh38
rs745511944 Chr.11: 130121726-130121726 on Build GRCh38
rs918034567 Chr.11: 130121795-130121795 on Build GRCh38
rs1063199 Chr.11: 130121740-130121740 on Build GRCh38
rs773090799 Chr.11: 130121859-130121859 on Build GRCh38
rs748289969 Chr.11: 130121700-130121700 on Build GRCh38
rs758525723 Chr.11: 130121593-130121593 on Build GRCh38
rs148916176 Chr.11: 130121668-130121668 on Build GRCh38
rs546811606 Chr.11: 130121912-130121912 on Build GRCh38
rs550926364 Chr.11: 130121982-130121982 on Build GRCh38
rs763528326 Chr.11: 130121740-130121763 on Build GRCh38
rs530537599 Chr.11: 130121715-130121715 on Build GRCh38
rs376609298 Chr.11: 130121848-130121848 on Build GRCh38
rs192318181 Chr.11: 130121880-130121880 on Build GRCh38
rs773429340 Chr.11: 130121635-130121635 on Build GRCh38
rs1047965 Chr.11: 130121764-130121764 on Build GRCh38
rs774062269 Chr.11: 130121649-130121649 on Build GRCh38
rs375773341 Chr.11: 130121659-130121659 on Build GRCh38
rs892653367 Chr.11: 130121688-130121688 on Build GRCh38
rs746873538 Chr.11: 130121758-130121760 on Build GRCh38
rs749689111 Chr.11: 130121640-130121640 on Build GRCh38
rs747045247 Chr.11: 130121606-130121606 on Build GRCh38
rs1025956614 Chr.11: 130121779-130121779 on Build GRCh38
rs767877572 Chr.11: 130121787-130121787 on Build GRCh38
rs756098712 Chr.11: 130121587-130121587 on Build GRCh38
rs762869004 Chr.11: 130121860-130121860 on Build GRCh38
rs779221283 Chr.11: 130121833-130121833 on Build GRCh38
rs867693478 Chr.11: 130121904-130121904 on Build GRCh38
rs184532542 Chr.11: 130121999-130121999 on Build GRCh38
rs765077430 Chr.11: 130121767-130121767 on Build GRCh38
rs768868685 Chr.11: 130121757-130121758 on Build GRCh38
rs567940481 Chr.11: 130121994-130121994 on Build GRCh38
rs752781277 Chr.11: 130121796-130121796 on Build GRCh38
rs986972144 Chr.11: 130121676-130121676 on Build GRCh38
rs369959621 Chr.11: 130121568-130121568 on Build GRCh38
rs140807639 Chr.11: 130121799-130121799 on Build GRCh38
rs752641528 Chr.11: 130121582-130121582 on Build GRCh38
rs781478768 Chr.11: 130121758-130121769 on Build GRCh38
rs1047131436 Chr.11: 130121550-130121550 on Build GRCh38
rs373359703 Chr.11: 130121650-130121650 on Build GRCh38
rs190276777 Chr.11: 130121677-130121677 on Build GRCh38
rs560817140 Chr.11: 130121820-130121820 on Build GRCh38
rs763248768 Chr.11: 130121821-130121821 on Build GRCh38
rs370107772 Chr.11: 130121723-130121723 on Build GRCh38
rs199857117 Chr.11: 130121751-130121751 on Build GRCh38
rs767932698 Chr.11: 130121627-130121627 on Build GRCh38
rs753525962 Chr.11: 130121663-130121663 on Build GRCh38
rs933511016 Chr.11: 130121894-130121894 on Build GRCh38
rs531097106 Chr.11: 130121978-130121978 on Build GRCh38
rs111377808 Chr.11: 130121566-130121566 on Build GRCh38
rs3837393 Chr.11: 130121775-130121780 on Build GRCh38
rs11822318 Chr.11: 130121711-130121711 on Build GRCh38
rs559756595 Chr.11: 130121870-130121870 on Build GRCh38
rs565247278 Chr.11: 130121762-130121762 on Build GRCh38
rs771749591 Chr.11: 130121727-130121727 on Build GRCh38
rs755319091 Chr.11: 130121679-130121679 on Build GRCh38
rs769948788 Chr.11: 130121776-130121781 on Build GRCh38
rs200945950 Chr.11: 130121781-130121781 on Build GRCh38
rs778627183 Chr.11: 130121746-130121757 on Build GRCh38
rs748234634 Chr.11: 130121767-130121769 on Build GRCh38
rs545528084 Chr.11: 130121758-130121758 on Build GRCh38
rs781481027 Chr.11: 130121621-130121621 on Build GRCh38
rs974338699 Chr.11: 130121556-130121556 on Build GRCh38
rs78715550 Chr.11: 130121873-130121873 on Build GRCh38
rs147875682 Chr.11: 130121620-130121620 on Build GRCh38
rs753504944 Chr.11: 130121739-130121740 on Build GRCh38
rs757775115 Chr.11: 130121812-130121812 on Build GRCh38
rs760199708 Chr.11: 130121754-130121754 on Build GRCh38
rs770454146 Chr.11: 130121647-130121647 on Build GRCh38
rs756218456 Chr.11: 130121797-130121797 on Build GRCh38
rs775045761 Chr.11: 130121655-130121655 on Build GRCh38
rs112139576 Chr.11: 130121749-130121749 on Build GRCh38
rs1063201 Chr.11: 130121757-130121757 on Build GRCh38
rs574399703 Chr.11: 130121684-130121684 on Build GRCh38
rs759811297 Chr.11: 130121784-130121784 on Build GRCh38
rs769900114 Chr.11: 130121858-130121858 on Build GRCh38

View Full Product Details