Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 134258334-134258857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD8
Entrez Gene ID
27034
Gene Name
acyl-CoA dehydrogenase family member 8
Gene Aliases
ACAD-8, ARC42
UniGene
Hs.14791
Gene Chromosome Location
Chr.11: 134253532-134265858 on Build GRCh38
Transcripts
NM_014384.2 XM_005271505.3 XM_011542750.2 XM_017017542.1 XM_017017543.1 XM_017017544.1 XM_017017545.1 XM_017017546.1 XM_017017547.1 XM_017017548.1 XM_017017549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6190805 c.169G>A p.D57N Chr.11: 134258534-134258534 on Build GRCh38
1727460 c.413A>C p.N138T Chr.11: 134258547-134258547 on Build GRCh38
5372104 c.134A>G p.H45R Chr.11: 134258562-134258562 on Build GRCh38
925193 c.443C>T p.P148L Chr.11: 134258577-134258577 on Build GRCh38
5142221 c.149C>T p.P50L Chr.11: 134258577-134258577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765021009 Chr.11: 134258475-134258475 on Build GRCh38
rs752761235 Chr.11: 134258517-134258517 on Build GRCh38
rs201273972 Chr.11: 134258600-134258600 on Build GRCh38
rs369088431 Chr.11: 134258792-134258792 on Build GRCh38
rs398124269 Chr.11: 134258582-134258583 on Build GRCh38
rs752810983 Chr.11: 134258584-134258584 on Build GRCh38
rs771887909 Chr.11: 134258539-134258539 on Build GRCh38
rs544414693 Chr.11: 134258733-134258733 on Build GRCh38
rs776363572 Chr.11: 134258553-134258553 on Build GRCh38
rs754229258 Chr.11: 134258533-134258533 on Build GRCh38
rs143408080 Chr.11: 134258542-134258542 on Build GRCh38
rs371449613 Chr.11: 134258543-134258543 on Build GRCh38
rs374317179 Chr.11: 134258518-134258518 on Build GRCh38
rs11223738 Chr.11: 134258419-134258419 on Build GRCh38
rs947147708 Chr.11: 134258796-134258796 on Build GRCh38
rs892670416 Chr.11: 134258360-134258360 on Build GRCh38
rs1043899311 Chr.11: 134258813-134258813 on Build GRCh38
rs570858339 Chr.11: 134258389-134258389 on Build GRCh38
rs997267059 Chr.11: 134258396-134258396 on Build GRCh38
rs866403074 Chr.11: 134258535-134258535 on Build GRCh38
rs749694455 Chr.11: 134258623-134258659 on Build GRCh38
rs112203893 Chr.11: 134258765-134258765 on Build GRCh38
rs779385985 Chr.11: 134258607-134258607 on Build GRCh38
rs772284615 Chr.11: 134258666-134258666 on Build GRCh38
rs368254088 Chr.11: 134258644-134258644 on Build GRCh38
rs779637415 Chr.11: 134258629-134258629 on Build GRCh38
rs1026449115 Chr.11: 134258364-134258364 on Build GRCh38
rs955623789 Chr.11: 134258401-134258401 on Build GRCh38
rs1006005186 Chr.11: 134258754-134258754 on Build GRCh38
rs779263186 Chr.11: 134258648-134258648 on Build GRCh38
rs564294269 Chr.11: 134258829-134258829 on Build GRCh38
rs74753754 Chr.11: 134258397-134258397 on Build GRCh38
rs886048024 Chr.11: 134258566-134258566 on Build GRCh38
rs200055094 Chr.11: 134258501-134258501 on Build GRCh38
rs367857040 Chr.11: 134258534-134258534 on Build GRCh38
rs121908418 Chr.11: 134258589-134258589 on Build GRCh38
rs938362128 Chr.11: 134258752-134258752 on Build GRCh38
rs758107874 Chr.11: 134258617-134258617 on Build GRCh38
rs573859354 Chr.11: 134258448-134258448 on Build GRCh38
rs533311814 Chr.11: 134258382-134258382 on Build GRCh38
rs552523927 Chr.11: 134258666-134258667 on Build GRCh38
rs969749306 Chr.11: 134258803-134258803 on Build GRCh38
rs552267218 Chr.11: 134258385-134258385 on Build GRCh38
rs903301376 Chr.11: 134258700-134258700 on Build GRCh38
rs373878500 Chr.11: 134258793-134258793 on Build GRCh38
rs886048023 Chr.11: 134258511-134258511 on Build GRCh38
rs938545157 Chr.11: 134258506-134258506 on Build GRCh38
rs572820646 Chr.11: 134258578-134258578 on Build GRCh38
rs1026504134 Chr.11: 134258688-134258688 on Build GRCh38
rs770146494 Chr.11: 134258489-134258493 on Build GRCh38
rs562700005 Chr.11: 134258735-134258735 on Build GRCh38
rs1044437581 Chr.11: 134258454-134258454 on Build GRCh38
rs908338967 Chr.11: 134258406-134258406 on Build GRCh38
rs775673393 Chr.11: 134258730-134258733 on Build GRCh38
rs778203609 Chr.11: 134258520-134258520 on Build GRCh38
rs751723313 Chr.11: 134258481-134258481 on Build GRCh38
rs35423297 Chr.11: 134258593-134258594 on Build GRCh38
rs948669930 Chr.11: 134258453-134258454 on Build GRCh38
rs760736043 Chr.11: 134258588-134258588 on Build GRCh38
rs757866565 Chr.11: 134258605-134258605 on Build GRCh38
rs986938589 Chr.11: 134258404-134258404 on Build GRCh38
rs1009786730 Chr.11: 134258369-134258369 on Build GRCh38
rs761661586 Chr.11: 134258473-134258473 on Build GRCh38
rs761751430 Chr.11: 134258563-134258563 on Build GRCh38
rs778104543 Chr.11: 134258545-134258545 on Build GRCh38
rs140515698 Chr.11: 134258538-134258538 on Build GRCh38
rs1008690789 Chr.11: 134258564-134258564 on Build GRCh38
rs11306396 Chr.11: 134258793-134258793 on Build GRCh38
rs928253576 Chr.11: 134258736-134258736 on Build GRCh38
rs886163031 Chr.11: 134258689-134258689 on Build GRCh38
rs370050595 Chr.11: 134258502-134258502 on Build GRCh38
rs759776882 Chr.11: 134258486-134258486 on Build GRCh38
rs150399643 Chr.11: 134258575-134258575 on Build GRCh38
rs767992967 Chr.11: 134258652-134258652 on Build GRCh38
rs545303222 Chr.11: 134258770-134258770 on Build GRCh38
rs780776705 Chr.11: 134258658-134258658 on Build GRCh38
rs573818576 Chr.11: 134258659-134258659 on Build GRCh38
rs186756646 Chr.11: 134258615-134258615 on Build GRCh38
rs997131128 Chr.11: 134258540-134258540 on Build GRCh38
rs970379928 Chr.11: 134258680-134258680 on Build GRCh38
rs779437942 Chr.11: 134258536-134258536 on Build GRCh38
rs374584216 Chr.11: 134258577-134258577 on Build GRCh38
rs535644930 Chr.11: 134258499-134258499 on Build GRCh38

View Full Product Details