Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 4644923-4645344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKAP3
Entrez Gene ID
10566
Gene Name
A-kinase anchoring protein 3
Gene Aliases
AKAP 110, AKAP110, CT82, FSP95, HEL159, PRKA3, SOB1
UniGene
Hs.98397
Gene Chromosome Location
Chr.12: 4615508-4649047 on Build GRCh38
Transcripts
NM_001278309.1 NM_006422.3 XM_005253664.2 XM_011520908.1 XM_011520909.1 XM_011520910.2

SNP Detail

SNP ID Chromosome Location
rs761572070 Chr.12: 4645294-4645294 on Build GRCh38
rs559395875 Chr.12: 4644950-4644950 on Build GRCh38
rs1016321102 Chr.12: 4645238-4645238 on Build GRCh38
rs989189938 Chr.12: 4645092-4645092 on Build GRCh38
rs954419202 Chr.12: 4645066-4645066 on Build GRCh38
rs1046324040 Chr.12: 4645181-4645181 on Build GRCh38
rs1016289752 Chr.12: 4645236-4645236 on Build GRCh38
rs372137626 Chr.12: 4645218-4645218 on Build GRCh38
rs536668926 Chr.12: 4645084-4645084 on Build GRCh38
rs1022643589 Chr.12: 4645137-4645137 on Build GRCh38
rs7971891 Chr.12: 4645255-4645255 on Build GRCh38
rs78292849 Chr.12: 4644947-4644947 on Build GRCh38
rs932718859 Chr.12: 4644952-4644952 on Build GRCh38
rs994047406 Chr.12: 4645251-4645251 on Build GRCh38
rs954118333 Chr.12: 4645297-4645297 on Build GRCh38
rs1056894434 Chr.12: 4645104-4645104 on Build GRCh38
rs962162455 Chr.12: 4644996-4644996 on Build GRCh38
rs898007208 Chr.12: 4645249-4645249 on Build GRCh38
rs1006312567 Chr.12: 4645237-4645237 on Build GRCh38
rs537431466 Chr.12: 4644948-4644948 on Build GRCh38
rs566932982 Chr.12: 4645278-4645278 on Build GRCh38
rs986226170 Chr.12: 4645081-4645081 on Build GRCh38
rs952459758 Chr.12: 4645200-4645200 on Build GRCh38
rs754046591 Chr.12: 4645198-4645198 on Build GRCh38
rs368284258 Chr.12: 4645023-4645023 on Build GRCh38
rs550814500 Chr.12: 4645112-4645112 on Build GRCh38
rs910207823 Chr.12: 4645082-4645082 on Build GRCh38
rs985917975 Chr.12: 4645316-4645316 on Build GRCh38
rs976958264 Chr.12: 4645269-4645269 on Build GRCh38
rs149641452 Chr.12: 4645073-4645073 on Build GRCh38
rs1012587725 Chr.12: 4645124-4645124 on Build GRCh38
rs78712892 Chr.12: 4645031-4645031 on Build GRCh38
rs947336525 Chr.12: 4645043-4645043 on Build GRCh38
rs138250320 Chr.12: 4644975-4644975 on Build GRCh38
rs571064074 Chr.12: 4645116-4645116 on Build GRCh38
rs1043339477 Chr.12: 4645052-4645052 on Build GRCh38
rs906421142 Chr.12: 4645193-4645193 on Build GRCh38
rs1049759256 Chr.12: 4644960-4644960 on Build GRCh38
rs1030969633 Chr.12: 4645038-4645038 on Build GRCh38
rs768562851 Chr.12: 4645171-4645171 on Build GRCh38

View Full Product Details