Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 5563438-5563936 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO2
Entrez Gene ID
57101
Gene Name
anoctamin 2
Gene Aliases
C12orf3, TMEM16B
UniGene
Hs.148970
Gene Chromosome Location
Chr.12: 5560241-5948790 on Build GRCh38
Transcripts
NM_001278596.1 NM_001278597.1 XM_006718988.3 XM_011520975.2 XM_011520978.2 XM_017019672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4953071 c.2731-1G>T p.? Chr.12: 5563569-5563569 on Build GRCh38
7355782 c.2859G>T p.K953N Chr.12: 5563440-5563440 on Build GRCh38
6073060 c.2817C>A p.S939R Chr.12: 5563479-5563479 on Build GRCh38
468668 c.2749G>A p.V917I Chr.12: 5563547-5563547 on Build GRCh38
6601024 c.2752G>A p.V918I Chr.12: 5563547-5563547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201870276 Chr.12: 5563553-5563553 on Build GRCh38
rs997615340 Chr.12: 5563627-5563627 on Build GRCh38
rs773059306 Chr.12: 5563502-5563502 on Build GRCh38
rs1005829679 Chr.12: 5563824-5563824 on Build GRCh38
rs1055471825 Chr.12: 5563827-5563827 on Build GRCh38
rs373466590 Chr.12: 5563545-5563545 on Build GRCh38
rs750959277 Chr.12: 5563471-5563471 on Build GRCh38
rs552348717 Chr.12: 5563778-5563778 on Build GRCh38
rs372300716 Chr.12: 5563508-5563508 on Build GRCh38
rs112221309 Chr.12: 5563794-5563794 on Build GRCh38
rs147424620 Chr.12: 5563738-5563738 on Build GRCh38
rs777481159 Chr.12: 5563562-5563562 on Build GRCh38
rs868260763 Chr.12: 5563551-5563551 on Build GRCh38
rs749065299 Chr.12: 5563563-5563563 on Build GRCh38
rs779577005 Chr.12: 5563463-5563463 on Build GRCh38
rs747947447 Chr.12: 5563487-5563487 on Build GRCh38
rs976837712 Chr.12: 5563889-5563889 on Build GRCh38
rs759047811 Chr.12: 5563512-5563512 on Build GRCh38
rs545229849 Chr.12: 5563696-5563696 on Build GRCh38
rs765444481 Chr.12: 5563842-5563842 on Build GRCh38
rs762841872 Chr.12: 5563503-5563503 on Build GRCh38
rs967341584 Chr.12: 5563739-5563739 on Build GRCh38
rs373910465 Chr.12: 5563608-5563608 on Build GRCh38
rs566836662 Chr.12: 5563796-5563796 on Build GRCh38
rs529327394 Chr.12: 5563806-5563806 on Build GRCh38
rs775699209 Chr.12: 5563902-5563902 on Build GRCh38
rs1012459294 Chr.12: 5563590-5563590 on Build GRCh38
rs952142420 Chr.12: 5563474-5563474 on Build GRCh38
rs578224921 Chr.12: 5563473-5563473 on Build GRCh38
rs372919273 Chr.12: 5563523-5563523 on Build GRCh38
rs760452306 Chr.12: 5563594-5563594 on Build GRCh38
rs763308283 Chr.12: 5563610-5563610 on Build GRCh38
rs538503111 Chr.12: 5563616-5563616 on Build GRCh38
rs371310851 Chr.12: 5563604-5563604 on Build GRCh38
rs200912800 Chr.12: 5563577-5563577 on Build GRCh38
rs375548202 Chr.12: 5563513-5563513 on Build GRCh38
rs781231821 Chr.12: 5563547-5563547 on Build GRCh38
rs569001028 Chr.12: 5563760-5563760 on Build GRCh38
rs895310143 Chr.12: 5563585-5563585 on Build GRCh38
rs866112601 Chr.12: 5563525-5563525 on Build GRCh38
rs990482740 Chr.12: 5563539-5563539 on Build GRCh38
rs182361207 Chr.12: 5563837-5563837 on Build GRCh38
rs776959556 Chr.12: 5563805-5563805 on Build GRCh38
rs775108825 Chr.12: 5563586-5563586 on Build GRCh38
rs549184021 Chr.12: 5563835-5563835 on Build GRCh38
rs781322541 Chr.12: 5563483-5563483 on Build GRCh38
rs953338331 Chr.12: 5563673-5563673 on Build GRCh38
rs138818944 Chr.12: 5563532-5563532 on Build GRCh38
rs961856188 Chr.12: 5563861-5563861 on Build GRCh38
rs773758339 Chr.12: 5563509-5563509 on Build GRCh38
rs1052883114 Chr.12: 5563679-5563679 on Build GRCh38
rs185209166 Chr.12: 5563542-5563542 on Build GRCh38
rs376930143 Chr.12: 5563541-5563541 on Build GRCh38
rs990041605 Chr.12: 5563710-5563710 on Build GRCh38
rs1049258376 Chr.12: 5563893-5563893 on Build GRCh38
rs948212443 Chr.12: 5563846-5563846 on Build GRCh38
rs768895523 Chr.12: 5563479-5563479 on Build GRCh38
rs267603589 Chr.12: 5563461-5563461 on Build GRCh38
rs369360823 Chr.12: 5563521-5563521 on Build GRCh38
rs751593582 Chr.12: 5563544-5563544 on Build GRCh38
rs766806181 Chr.12: 5563611-5563611 on Build GRCh38
rs769630475 Chr.12: 5563495-5563495 on Build GRCh38
rs377720747 Chr.12: 5563548-5563548 on Build GRCh38
rs1028798499 Chr.12: 5563647-5563647 on Build GRCh38
rs142941746 Chr.12: 5563583-5563583 on Build GRCh38
rs891648955 Chr.12: 5563718-5563718 on Build GRCh38
rs879876423 Chr.12: 5563847-5563847 on Build GRCh38
rs747060528 Chr.12: 5563476-5563476 on Build GRCh38
rs190525282 Chr.12: 5563707-5563707 on Build GRCh38

View Full Product Details