Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 5612823-5613317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO2
Entrez Gene ID
57101
Gene Name
anoctamin 2
Gene Aliases
C12orf3, TMEM16B
UniGene
Hs.148970
Gene Chromosome Location
Chr.12: 5560241-5948790 on Build GRCh38
Transcripts
NM_001278596.1 NM_001278597.1 XM_006718988.3 XM_011520975.2 XM_011520978.2 XM_017019672.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4864557 c.1984G>T p.E662* Chr.12: 5612906-5612906 on Build GRCh38
694422 c.1935G>T p.V645V Chr.12: 5612952-5612952 on Build GRCh38
6462779 c.1929G>T p.R643S Chr.12: 5612958-5612958 on Build GRCh38
4533439 c.1981G>A p.E661K Chr.12: 5612906-5612906 on Build GRCh38
6462775 c.1930T>G p.F644V Chr.12: 5612957-5612957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950268473 Chr.12: 5613154-5613154 on Build GRCh38
rs759312015 Chr.12: 5612978-5612978 on Build GRCh38
rs575875855 Chr.12: 5613094-5613094 on Build GRCh38
rs928721177 Chr.12: 5613229-5613230 on Build GRCh38
rs755546576 Chr.12: 5612933-5612933 on Build GRCh38
rs982097000 Chr.12: 5613160-5613160 on Build GRCh38
rs183097689 Chr.12: 5612997-5612997 on Build GRCh38
rs919984707 Chr.12: 5613189-5613189 on Build GRCh38
rs915918668 Chr.12: 5613266-5613266 on Build GRCh38
rs941894332 Chr.12: 5613292-5613292 on Build GRCh38
rs3741838 Chr.12: 5613015-5613015 on Build GRCh38
rs1007172389 Chr.12: 5613123-5613123 on Build GRCh38
rs557701376 Chr.12: 5613205-5613205 on Build GRCh38
rs754933071 Chr.12: 5612921-5612921 on Build GRCh38
rs1047207510 Chr.12: 5613225-5613225 on Build GRCh38
rs771139190 Chr.12: 5612954-5612954 on Build GRCh38
rs913387429 Chr.12: 5613099-5613099 on Build GRCh38
rs781741599 Chr.12: 5612934-5612934 on Build GRCh38
rs373586770 Chr.12: 5612912-5612912 on Build GRCh38
rs758152600 Chr.12: 5613072-5613072 on Build GRCh38
rs61910084 Chr.12: 5612953-5612953 on Build GRCh38
rs770411402 Chr.12: 5613173-5613173 on Build GRCh38
rs187515117 Chr.12: 5613249-5613249 on Build GRCh38
rs112771756 Chr.12: 5613177-5613177 on Build GRCh38
rs535646339 Chr.12: 5613059-5613059 on Build GRCh38
rs956460564 Chr.12: 5613081-5613081 on Build GRCh38
rs930058693 Chr.12: 5613224-5613224 on Build GRCh38
rs899758422 Chr.12: 5613263-5613263 on Build GRCh38
rs376053738 Chr.12: 5612993-5612993 on Build GRCh38
rs138831946 Chr.12: 5613018-5613018 on Build GRCh38
rs967588816 Chr.12: 5613214-5613215 on Build GRCh38
rs553158058 Chr.12: 5613032-5613032 on Build GRCh38
rs185116864 Chr.12: 5612854-5612854 on Build GRCh38
rs748647903 Chr.12: 5612937-5612937 on Build GRCh38
rs770062677 Chr.12: 5612939-5612939 on Build GRCh38
rs1038381850 Chr.12: 5613185-5613185 on Build GRCh38
rs745914759 Chr.12: 5612906-5612906 on Build GRCh38
rs753503093 Chr.12: 5612911-5612911 on Build GRCh38
rs190902838 Chr.12: 5612941-5612941 on Build GRCh38
rs774176718 Chr.12: 5612853-5612853 on Build GRCh38
rs990274913 Chr.12: 5612907-5612907 on Build GRCh38
rs772011104 Chr.12: 5612987-5612987 on Build GRCh38
rs141938606 Chr.12: 5613230-5613231 on Build GRCh38
rs960147298 Chr.12: 5613259-5613259 on Build GRCh38
rs964048369 Chr.12: 5613033-5613033 on Build GRCh38
rs760602676 Chr.12: 5612995-5612995 on Build GRCh38
rs760120358 Chr.12: 5612887-5612887 on Build GRCh38
rs772396654 Chr.12: 5612975-5612976 on Build GRCh38
rs367647989 Chr.12: 5612922-5612922 on Build GRCh38
rs752212447 Chr.12: 5612866-5612866 on Build GRCh38
rs941967904 Chr.12: 5613022-5613022 on Build GRCh38
rs796702972 Chr.12: 5613228-5613229 on Build GRCh38
rs763582568 Chr.12: 5612896-5612896 on Build GRCh38
rs993961810 Chr.12: 5612884-5612884 on Build GRCh38
rs774048620 Chr.12: 5612974-5612974 on Build GRCh38
rs1038455142 Chr.12: 5612856-5612856 on Build GRCh38
rs890226481 Chr.12: 5613110-5613110 on Build GRCh38
rs191124858 Chr.12: 5613054-5613054 on Build GRCh38
rs183831523 Chr.12: 5613078-5613078 on Build GRCh38
rs1000451628 Chr.12: 5613287-5613287 on Build GRCh38
rs370814340 Chr.12: 5612857-5612857 on Build GRCh38
rs746637117 Chr.12: 5613170-5613170 on Build GRCh38
rs890936621 Chr.12: 5613232-5613232 on Build GRCh38
rs745311514 Chr.12: 5613288-5613288 on Build GRCh38
rs186694643 Chr.12: 5613012-5613012 on Build GRCh38
rs1040020435 Chr.12: 5613257-5613257 on Build GRCh38

View Full Product Details