Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7650875-7651317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC1
Entrez Gene ID
339
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 1
Gene Aliases
APOBEC-1, BEDP, CDAR1, HEPR
UniGene
Hs.560
Gene Chromosome Location
Chr.12: 7649400-7670599 on Build GRCh38
Transcripts
NM_001304566.1 NM_001644.4 NM_005889.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4551224 c.522G>A p.W174* Chr.12: 7651062-7651062 on Build GRCh38
695014 c.511C>A p.P171T Chr.12: 7651073-7651073 on Build GRCh38
5490610 c.496C>T p.H166Y Chr.12: 7651088-7651088 on Build GRCh38
147408 c.534C>T p.Y178Y Chr.12: 7651050-7651050 on Build GRCh38
695015 c.531G>A p.L177L Chr.12: 7651053-7651053 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771301145 Chr.12: 7651007-7651007 on Build GRCh38
rs779708510 Chr.12: 7650975-7650975 on Build GRCh38
rs747133733 Chr.12: 7651167-7651167 on Build GRCh38
rs373316326 Chr.12: 7651021-7651021 on Build GRCh38
rs973896833 Chr.12: 7651047-7651047 on Build GRCh38
rs149124082 Chr.12: 7651049-7651049 on Build GRCh38
rs779268089 Chr.12: 7651115-7651115 on Build GRCh38
rs746096886 Chr.12: 7651022-7651022 on Build GRCh38
rs754353926 Chr.12: 7651109-7651109 on Build GRCh38
rs757895062 Chr.12: 7650962-7650962 on Build GRCh38
rs999732016 Chr.12: 7650926-7650926 on Build GRCh38
rs746090364 Chr.12: 7651132-7651132 on Build GRCh38
rs12228353 Chr.12: 7650949-7650949 on Build GRCh38
rs201126517 Chr.12: 7651142-7651142 on Build GRCh38
rs764744663 Chr.12: 7651091-7651091 on Build GRCh38
rs1039779409 Chr.12: 7651117-7651117 on Build GRCh38
rs773006098 Chr.12: 7651186-7651186 on Build GRCh38
rs754502311 Chr.12: 7651071-7651071 on Build GRCh38
rs544725997 Chr.12: 7651218-7651218 on Build GRCh38
rs772048636 Chr.12: 7651035-7651035 on Build GRCh38
rs902635824 Chr.12: 7651232-7651232 on Build GRCh38
rs778310856 Chr.12: 7651003-7651003 on Build GRCh38
rs145595004 Chr.12: 7651256-7651256 on Build GRCh38
rs370728062 Chr.12: 7651069-7651069 on Build GRCh38
rs565348963 Chr.12: 7651036-7651036 on Build GRCh38
rs756636866 Chr.12: 7651001-7651001 on Build GRCh38
rs10431308 Chr.12: 7650966-7650966 on Build GRCh38
rs769485309 Chr.12: 7651181-7651181 on Build GRCh38
rs200489962 Chr.12: 7651159-7651159 on Build GRCh38
rs774763032 Chr.12: 7651231-7651231 on Build GRCh38
rs748076636 Chr.12: 7651000-7651000 on Build GRCh38
rs992241455 Chr.12: 7650911-7650911 on Build GRCh38
rs748114827 Chr.12: 7651180-7651180 on Build GRCh38
rs191285090 Chr.12: 7651112-7651112 on Build GRCh38
rs900486189 Chr.12: 7651204-7651204 on Build GRCh38
rs768701520 Chr.12: 7651173-7651173 on Build GRCh38
rs768541800 Chr.12: 7651039-7651039 on Build GRCh38
rs759640991 Chr.12: 7651229-7651229 on Build GRCh38
rs890884076 Chr.12: 7651093-7651093 on Build GRCh38
rs776493196 Chr.12: 7651178-7651178 on Build GRCh38
rs772809787 Chr.12: 7651057-7651057 on Build GRCh38
rs561124244 Chr.12: 7651255-7651255 on Build GRCh38
rs10431309 Chr.12: 7651050-7651050 on Build GRCh38
rs780138830 Chr.12: 7651161-7651161 on Build GRCh38
rs1013608998 Chr.12: 7651266-7651266 on Build GRCh38
rs183871671 Chr.12: 7651219-7651219 on Build GRCh38
rs967266953 Chr.12: 7650909-7650909 on Build GRCh38
rs942357724 Chr.12: 7651097-7651097 on Build GRCh38
rs746706745 Chr.12: 7650931-7650931 on Build GRCh38
rs148255219 Chr.12: 7651070-7651070 on Build GRCh38
rs376504316 Chr.12: 7651061-7651061 on Build GRCh38
rs991965940 Chr.12: 7651108-7651108 on Build GRCh38

View Full Product Details