Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 7652757-7653269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOBEC1
Entrez Gene ID
339
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 1
Gene Aliases
APOBEC-1, BEDP, CDAR1, HEPR
UniGene
Hs.560
Gene Chromosome Location
Chr.12: 7649400-7670599 on Build GRCh38
Transcripts
NM_001304566.1 NM_001644.4 NM_005889.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4637696 c.119A>G p.Y40C Chr.12: 7652761-7652761 on Build GRCh38
6138022 c.120C>T p.Y40Y Chr.12: 7652760-7652760 on Build GRCh38
3399118 c.73G>A p.V25I Chr.12: 7652807-7652807 on Build GRCh38
6014828 c.108C>T p.A36A Chr.12: 7652772-7652772 on Build GRCh38
5913329 c.103G>A p.E35K Chr.12: 7652777-7652777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904203034 Chr.12: 7653140-7653140 on Build GRCh38
rs78510245 Chr.12: 7652916-7652916 on Build GRCh38
rs780974536 Chr.12: 7653010-7653010 on Build GRCh38
rs267603686 Chr.12: 7652825-7652825 on Build GRCh38
rs969352260 Chr.12: 7653060-7653060 on Build GRCh38
rs373437987 Chr.12: 7653133-7653133 on Build GRCh38
rs757381551 Chr.12: 7652837-7652837 on Build GRCh38
rs908716692 Chr.12: 7652842-7652842 on Build GRCh38
rs983254342 Chr.12: 7653242-7653242 on Build GRCh38
rs1027330192 Chr.12: 7653176-7653176 on Build GRCh38
rs768219565 Chr.12: 7652784-7652784 on Build GRCh38
rs199783827 Chr.12: 7652817-7652817 on Build GRCh38
rs751050076 Chr.12: 7652939-7652939 on Build GRCh38
rs952760718 Chr.12: 7653238-7653238 on Build GRCh38
rs374067690 Chr.12: 7652920-7652920 on Build GRCh38
rs780046917 Chr.12: 7652916-7652919 on Build GRCh38
rs919942724 Chr.12: 7653079-7653079 on Build GRCh38
rs189158951 Chr.12: 7653042-7653042 on Build GRCh38
rs5796283 Chr.12: 7653096-7653097 on Build GRCh38
rs761206062 Chr.12: 7653144-7653144 on Build GRCh38
rs12422951 Chr.12: 7653124-7653124 on Build GRCh38
rs1019519260 Chr.12: 7653196-7653196 on Build GRCh38
rs934384239 Chr.12: 7653142-7653142 on Build GRCh38
rs547285344 Chr.12: 7653033-7653033 on Build GRCh38
rs762320087 Chr.12: 7652856-7652856 on Build GRCh38
rs768807626 Chr.12: 7653096-7653096 on Build GRCh38
rs867200012 Chr.12: 7652783-7652783 on Build GRCh38
rs1014109155 Chr.12: 7653035-7653035 on Build GRCh38
rs765832841 Chr.12: 7652913-7652921 on Build GRCh38
rs913276039 Chr.12: 7653221-7653221 on Build GRCh38
rs750373058 Chr.12: 7652861-7652861 on Build GRCh38
rs774350785 Chr.12: 7652966-7652966 on Build GRCh38
rs760748421 Chr.12: 7652821-7652821 on Build GRCh38
rs1042209937 Chr.12: 7652893-7652893 on Build GRCh38
rs754848038 Chr.12: 7653032-7653032 on Build GRCh38
rs1027884570 Chr.12: 7652992-7652992 on Build GRCh38
rs369728515 Chr.12: 7653123-7653123 on Build GRCh38
rs1003334941 Chr.12: 7652922-7652922 on Build GRCh38
rs112046539 Chr.12: 7652913-7652916 on Build GRCh38
rs906268998 Chr.12: 7652901-7652901 on Build GRCh38
rs990102605 Chr.12: 7653216-7653216 on Build GRCh38
rs868470644 Chr.12: 7652977-7652977 on Build GRCh38
rs747039859 Chr.12: 7652820-7652820 on Build GRCh38
rs1005176071 Chr.12: 7653181-7653181 on Build GRCh38
rs191789228 Chr.12: 7653237-7653237 on Build GRCh38
rs772991611 Chr.12: 7653172-7653172 on Build GRCh38
rs950935366 Chr.12: 7653180-7653180 on Build GRCh38
rs921547272 Chr.12: 7653130-7653130 on Build GRCh38
rs1041337710 Chr.12: 7652892-7652892 on Build GRCh38
rs922003358 Chr.12: 7653210-7653210 on Build GRCh38
rs766494598 Chr.12: 7652957-7652957 on Build GRCh38
rs945330295 Chr.12: 7653132-7653132 on Build GRCh38
rs34275479 Chr.12: 7652818-7652818 on Build GRCh38
rs776064348 Chr.12: 7652790-7652790 on Build GRCh38
rs893745121 Chr.12: 7652826-7652826 on Build GRCh38
rs767765861 Chr.12: 7652819-7652819 on Build GRCh38
rs376768287 Chr.12: 7652939-7652940 on Build GRCh38
rs895627994 Chr.12: 7653153-7653153 on Build GRCh38
rs994720259 Chr.12: 7653220-7653220 on Build GRCh38
rs779649867 Chr.12: 7652878-7652878 on Build GRCh38
rs767231515 Chr.12: 7652916-7652920 on Build GRCh38
rs957460354 Chr.12: 7653212-7653212 on Build GRCh38
rs746331216 Chr.12: 7653138-7653138 on Build GRCh38
rs906389697 Chr.12: 7653137-7653137 on Build GRCh38
rs974885476 Chr.12: 7653078-7653078 on Build GRCh38
rs1022866498 Chr.12: 7653044-7653044 on Build GRCh38
rs753928830 Chr.12: 7652827-7652827 on Build GRCh38
rs33949127 Chr.12: 7652916-7652917 on Build GRCh38
rs998624788 Chr.12: 7652976-7652976 on Build GRCh38
rs966521633 Chr.12: 7653041-7653041 on Build GRCh38
rs894907149 Chr.12: 7653162-7653162 on Build GRCh38
rs376504384 Chr.12: 7653175-7653175 on Build GRCh38
rs966210644 Chr.12: 7653208-7653208 on Build GRCh38
rs200390394 Chr.12: 7652807-7652807 on Build GRCh38
rs138302046 Chr.12: 7652935-7652935 on Build GRCh38
rs768751278 Chr.12: 7652915-7652919 on Build GRCh38
rs746301833 Chr.12: 7652898-7652898 on Build GRCh38
rs542787441 Chr.12: 7652812-7652812 on Build GRCh38
rs935995586 Chr.12: 7653112-7653112 on Build GRCh38
rs994518240 Chr.12: 7652970-7652970 on Build GRCh38
rs201808135 Chr.12: 7652915-7652915 on Build GRCh38
rs5796282 Chr.12: 7652915-7652916 on Build GRCh38
rs758510531 Chr.12: 7652808-7652808 on Build GRCh38
rs945278068 Chr.12: 7653222-7653222 on Build GRCh38
rs950944669 Chr.12: 7653006-7653006 on Build GRCh38
rs905749790 Chr.12: 7653218-7653218 on Build GRCh38
rs2080127 Chr.12: 7652871-7652871 on Build GRCh38
rs187059919 Chr.12: 7653209-7653209 on Build GRCh38
rs183088817 Chr.12: 7653151-7653151 on Build GRCh38

View Full Product Details