Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12974773-12975280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM234B
Entrez Gene ID
57613
Gene Name
family with sequence similarity 234 member B
Gene Aliases
KIAA1467
UniGene
Hs.132660
Gene Chromosome Location
Chr.12: 12927680-13083449 on Build GRCh38
Transcripts
XM_017019706.1 NM_015987.4

Gene Symbol
HEBP1
Entrez Gene ID
50865
Gene Name
heme binding protein 1
Gene Aliases
HBP, HEBP
UniGene
Hs.642618
Gene Chromosome Location
Chr.12: 12974864-13000309 on Build GRCh38
Transcripts
XM_017019706.1 NM_015987.4

SNP Detail

SNP ID Chromosome Location
rs958143540 Chr.12: 12974808-12974808 on Build GRCh38
rs188958425 Chr.12: 12975005-12975005 on Build GRCh38
rs966721230 Chr.12: 12975157-12975157 on Build GRCh38
rs772656568 Chr.12: 12975128-12975128 on Build GRCh38
rs7968708 Chr.12: 12975142-12975142 on Build GRCh38
rs142044107 Chr.12: 12974852-12974852 on Build GRCh38
rs906873830 Chr.12: 12975057-12975057 on Build GRCh38
rs940602008 Chr.12: 12975030-12975030 on Build GRCh38
rs1017457671 Chr.12: 12975095-12975095 on Build GRCh38
rs990990504 Chr.12: 12974925-12974925 on Build GRCh38
rs898825164 Chr.12: 12975069-12975069 on Build GRCh38
rs552339689 Chr.12: 12974861-12974861 on Build GRCh38
rs1030994190 Chr.12: 12975239-12975239 on Build GRCh38
rs916625545 Chr.12: 12974917-12974917 on Build GRCh38
rs13479 Chr.12: 12975124-12975124 on Build GRCh38
rs146842330 Chr.12: 12975064-12975064 on Build GRCh38
rs570127716 Chr.12: 12975083-12975083 on Build GRCh38
rs777259078 Chr.12: 12975081-12975081 on Build GRCh38
rs958028816 Chr.12: 12975253-12975253 on Build GRCh38
rs368584778 Chr.12: 12975031-12975031 on Build GRCh38
rs2232244 Chr.12: 12975256-12975256 on Build GRCh38
rs916705422 Chr.12: 12974951-12974951 on Build GRCh38
rs1046201248 Chr.12: 12974992-12974992 on Build GRCh38
rs1045850831 Chr.12: 12974978-12974978 on Build GRCh38
rs79429827 Chr.12: 12975254-12975254 on Build GRCh38
rs900295675 Chr.12: 12975122-12975122 on Build GRCh38
rs966037904 Chr.12: 12975215-12975215 on Build GRCh38
rs769884768 Chr.12: 12975170-12975170 on Build GRCh38
rs947890239 Chr.12: 12974961-12974961 on Build GRCh38
rs999719526 Chr.12: 12975223-12975223 on Build GRCh38
rs539452125 Chr.12: 12975156-12975156 on Build GRCh38
rs73068796 Chr.12: 12975088-12975088 on Build GRCh38
rs562298053 Chr.12: 12975077-12975077 on Build GRCh38
rs927890075 Chr.12: 12974996-12974996 on Build GRCh38
rs765952019 Chr.12: 12975146-12975146 on Build GRCh38

View Full Product Details