Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 14942328-14942833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGDIB
Entrez Gene ID
397
Gene Name
Rho GDP dissociation inhibitor beta
Gene Aliases
D4, GDIA2, GDID4, LYGDI, Ly-GDI, RAP1GN1, RhoGDI2
UniGene
Hs.504877
Gene Chromosome Location
Chr.12: 14942015-14961770 on Build GRCh38
Transcripts
NM_001175.6 NM_001321420.1 NM_001321421.1 NM_001321422.1 NM_001321423.1 NR_135637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2003153 c.506G>A p.R169Q Chr.12: 14942622-14942622 on Build GRCh38
5012499 c.423T>G p.F141L Chr.12: 14942705-14942705 on Build GRCh38
937583 c.581C>T p.S194L Chr.12: 14942547-14942547 on Build GRCh38
4995109 c.505C>T p.R169* Chr.12: 14942623-14942623 on Build GRCh38
2003147 c.564C>T p.S188S Chr.12: 14942564-14942564 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046129429 Chr.12: 14942707-14942707 on Build GRCh38
rs903256176 Chr.12: 14942375-14942375 on Build GRCh38
rs566545782 Chr.12: 14942513-14942513 on Build GRCh38
rs1054555099 Chr.12: 14942561-14942561 on Build GRCh38
rs767992929 Chr.12: 14942758-14942758 on Build GRCh38
rs772149449 Chr.12: 14942555-14942555 on Build GRCh38
rs958619131 Chr.12: 14942468-14942468 on Build GRCh38
rs760868737 Chr.12: 14942500-14942500 on Build GRCh38
rs1012961622 Chr.12: 14942652-14942652 on Build GRCh38
rs776957092 Chr.12: 14942588-14942588 on Build GRCh38
rs1000245450 Chr.12: 14942745-14942746 on Build GRCh38
rs1024253870 Chr.12: 14942800-14942800 on Build GRCh38
rs771982112 Chr.12: 14942554-14942554 on Build GRCh38
rs748771676 Chr.12: 14942546-14942546 on Build GRCh38
rs550309820 Chr.12: 14942441-14942441 on Build GRCh38
rs551253086 Chr.12: 14942488-14942488 on Build GRCh38
rs748044797 Chr.12: 14942657-14942657 on Build GRCh38
rs745906229 Chr.12: 14942532-14942532 on Build GRCh38
rs369111462 Chr.12: 14942625-14942625 on Build GRCh38
rs201666982 Chr.12: 14942591-14942591 on Build GRCh38
rs765241849 Chr.12: 14942725-14942725 on Build GRCh38
rs773158474 Chr.12: 14942556-14942556 on Build GRCh38
rs532853866 Chr.12: 14942448-14942448 on Build GRCh38
rs764609149 Chr.12: 14942739-14942739 on Build GRCh38
rs745791654 Chr.12: 14942746-14942746 on Build GRCh38
rs1003476823 Chr.12: 14942736-14942736 on Build GRCh38
rs776710306 Chr.12: 14942709-14942709 on Build GRCh38
rs901147282 Chr.12: 14942363-14942363 on Build GRCh38
rs2430710 Chr.12: 14942806-14942806 on Build GRCh38
rs542973414 Chr.12: 14942801-14942801 on Build GRCh38
rs767464138 Chr.12: 14942491-14942491 on Build GRCh38
rs755743098 Chr.12: 14942622-14942622 on Build GRCh38
rs961848897 Chr.12: 14942356-14942356 on Build GRCh38
rs34914037 Chr.12: 14942508-14942508 on Build GRCh38
rs995069753 Chr.12: 14942794-14942794 on Build GRCh38
rs749131670 Chr.12: 14942611-14942618 on Build GRCh38
rs778988055 Chr.12: 14942531-14942531 on Build GRCh38
rs770609750 Chr.12: 14942627-14942649 on Build GRCh38
rs746754433 Chr.12: 14942674-14942674 on Build GRCh38
rs759500495 Chr.12: 14942590-14942590 on Build GRCh38
rs1032804452 Chr.12: 14942446-14942446 on Build GRCh38
rs973177864 Chr.12: 14942369-14942369 on Build GRCh38
rs772143582 Chr.12: 14942666-14942666 on Build GRCh38
rs777252647 Chr.12: 14942505-14942506 on Build GRCh38
rs377289696 Chr.12: 14942607-14942607 on Build GRCh38
rs10772822 Chr.12: 14942773-14942773 on Build GRCh38
rs1027445940 Chr.12: 14942802-14942802 on Build GRCh38
rs200511147 Chr.12: 14942682-14942682 on Build GRCh38
rs752158593 Chr.12: 14942745-14942745 on Build GRCh38
rs11056249 Chr.12: 14942394-14942394 on Build GRCh38
rs764374478 Chr.12: 14942615-14942615 on Build GRCh38
rs750671560 Chr.12: 14942495-14942495 on Build GRCh38
rs912169682 Chr.12: 14942471-14942471 on Build GRCh38
rs1012811178 Chr.12: 14942470-14942470 on Build GRCh38
rs761775273 Chr.12: 14942534-14942534 on Build GRCh38
rs568417535 Chr.12: 14942724-14942724 on Build GRCh38
rs758920170 Chr.12: 14942612-14942612 on Build GRCh38
rs7973855 Chr.12: 14942776-14942776 on Build GRCh38
rs775551539 Chr.12: 14942726-14942726 on Build GRCh38
rs1042106739 Chr.12: 14942683-14942683 on Build GRCh38
rs757533858 Chr.12: 14942750-14942750 on Build GRCh38
rs993217823 Chr.12: 14942807-14942807 on Build GRCh38
rs953105967 Chr.12: 14942427-14942427 on Build GRCh38
rs202169635 Chr.12: 14942530-14942530 on Build GRCh38
rs1020392554 Chr.12: 14942370-14942370 on Build GRCh38
rs751901004 Chr.12: 14942616-14942616 on Build GRCh38
rs773088892 Chr.12: 14942669-14942669 on Build GRCh38
rs1803929 Chr.12: 14942689-14942689 on Build GRCh38
rs753891686 Chr.12: 14942503-14942503 on Build GRCh38
rs769335849 Chr.12: 14942529-14942529 on Build GRCh38
rs139160089 Chr.12: 14942648-14942648 on Build GRCh38
rs774413441 Chr.12: 14942673-14942686 on Build GRCh38
rs944947131 Chr.12: 14942483-14942483 on Build GRCh38
rs760888228 Chr.12: 14942573-14942573 on Build GRCh38
rs4703 Chr.12: 14942624-14942624 on Build GRCh38
rs753423402 Chr.12: 14942772-14942772 on Build GRCh38
rs35772331 Chr.12: 14942509-14942509 on Build GRCh38
rs774401441 Chr.12: 14942547-14942547 on Build GRCh38
rs907171476 Chr.12: 14942721-14942721 on Build GRCh38
rs766528316 Chr.12: 14942586-14942586 on Build GRCh38
rs41276684 Chr.12: 14942511-14942511 on Build GRCh38
rs745465573 Chr.12: 14942519-14942519 on Build GRCh38
rs780587141 Chr.12: 14942631-14942631 on Build GRCh38
rs1054303003 Chr.12: 14942747-14942747 on Build GRCh38
rs763409543 Chr.12: 14942737-14942737 on Build GRCh38
rs751956207 Chr.12: 14942400-14942400 on Build GRCh38
rs769127083 Chr.12: 14942790-14942790 on Build GRCh38

View Full Product Details