Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 21800825-21801321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC9
Entrez Gene ID
10060
Gene Name
ATP binding cassette subfamily C member 9
Gene Aliases
ABC37, ATFB12, CANTU, CMD1O, SUR2
UniGene
Hs.732701
Gene Chromosome Location
Chr.12: 21797389-21941869 on Build GRCh38
Transcripts
NM_020297.3 XM_005253284.3 XM_005253286.3 XM_005253287.4 XM_005253288.3 XM_005253289.3 XM_005253290.3 XM_006719025.3 XM_011520545.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3398605 c.4535C>T p.T1512M Chr.12: 21801159-21801159 on Build GRCh38
1283184 c.4545G>C p.L1515L Chr.12: 21801149-21801149 on Build GRCh38
73534 c.4637G>A p.R1546H Chr.12: 21801057-21801057 on Build GRCh38
1746892 c.4573T>G p.L1525V Chr.12: 21801121-21801121 on Build GRCh38
1283185 c.4584C>T p.D1528D Chr.12: 21801110-21801110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755718495 Chr.12: 21801123-21801123 on Build GRCh38
rs756748926 Chr.12: 21801222-21801222 on Build GRCh38
rs529676218 Chr.12: 21800897-21800897 on Build GRCh38
rs1000050511 Chr.12: 21800940-21800941 on Build GRCh38
rs1011019352 Chr.12: 21801002-21801002 on Build GRCh38
rs890069742 Chr.12: 21801081-21801081 on Build GRCh38
rs762798124 Chr.12: 21801018-21801018 on Build GRCh38
rs947970608 Chr.12: 21800871-21800871 on Build GRCh38
rs773377070 Chr.12: 21801057-21801057 on Build GRCh38
rs376736105 Chr.12: 21801022-21801022 on Build GRCh38
rs370423028 Chr.12: 21800996-21800996 on Build GRCh38
rs918326565 Chr.12: 21801172-21801172 on Build GRCh38
rs121909304 Chr.12: 21801157-21801157 on Build GRCh38
rs771800460 Chr.12: 21801195-21801195 on Build GRCh38
rs560738082 Chr.12: 21800892-21800892 on Build GRCh38
rs766613117 Chr.12: 21801296-21801296 on Build GRCh38
rs753079722 Chr.12: 21801078-21801078 on Build GRCh38
rs180770035 Chr.12: 21801037-21801037 on Build GRCh38
rs868198875 Chr.12: 21801111-21801111 on Build GRCh38
rs878916894 Chr.12: 21801076-21801076 on Build GRCh38
rs147521845 Chr.12: 21801070-21801070 on Build GRCh38
rs765745602 Chr.12: 21801074-21801074 on Build GRCh38
rs772916231 Chr.12: 21800881-21800881 on Build GRCh38
rs762319597 Chr.12: 21800861-21800861 on Build GRCh38
rs143310355 Chr.12: 21801056-21801056 on Build GRCh38
rs773424178 Chr.12: 21801201-21801201 on Build GRCh38
rs774236898 Chr.12: 21801167-21801167 on Build GRCh38
rs566226557 Chr.12: 21800955-21800955 on Build GRCh38
rs747962000 Chr.12: 21801175-21801175 on Build GRCh38
rs568984710 Chr.12: 21801108-21801108 on Build GRCh38
rs775503041 Chr.12: 21801016-21801016 on Build GRCh38
rs876657736 Chr.12: 21801039-21801039 on Build GRCh38
rs913054111 Chr.12: 21800971-21800971 on Build GRCh38
rs906864509 Chr.12: 21801299-21801299 on Build GRCh38
rs372899855 Chr.12: 21801229-21801229 on Build GRCh38
rs755422776 Chr.12: 21801061-21801061 on Build GRCh38
rs766534765 Chr.12: 21801216-21801216 on Build GRCh38
rs373396917 Chr.12: 21801099-21801099 on Build GRCh38
rs796370044 Chr.12: 21800864-21800864 on Build GRCh38
rs772458579 Chr.12: 21800987-21800987 on Build GRCh38
rs114906131 Chr.12: 21801025-21801025 on Build GRCh38
rs927163179 Chr.12: 21800886-21800886 on Build GRCh38
rs753102079 Chr.12: 21801022-21801026 on Build GRCh38
rs768172354 Chr.12: 21800984-21800984 on Build GRCh38
rs766300688 Chr.12: 21801058-21801058 on Build GRCh38
rs186493048 Chr.12: 21801158-21801158 on Build GRCh38
rs781012885 Chr.12: 21801100-21801100 on Build GRCh38
rs957267834 Chr.12: 21800895-21800895 on Build GRCh38
rs922613300 Chr.12: 21801028-21801028 on Build GRCh38
rs971057093 Chr.12: 21800878-21800878 on Build GRCh38
rs567584120 Chr.12: 21801268-21801268 on Build GRCh38
rs554811993 Chr.12: 21801159-21801159 on Build GRCh38
rs906499382 Chr.12: 21800893-21800894 on Build GRCh38
rs767798962 Chr.12: 21801021-21801021 on Build GRCh38
rs760801245 Chr.12: 21801209-21801209 on Build GRCh38
rs727504538 Chr.12: 21801125-21801125 on Build GRCh38
rs188290333 Chr.12: 21800941-21800941 on Build GRCh38
rs916376725 Chr.12: 21801230-21801230 on Build GRCh38
rs947971665 Chr.12: 21801248-21801248 on Build GRCh38
rs78275359 Chr.12: 21801114-21801114 on Build GRCh38
rs1046302647 Chr.12: 21801297-21801297 on Build GRCh38
rs866552706 Chr.12: 21801278-21801278 on Build GRCh38
rs149722127 Chr.12: 21801085-21801085 on Build GRCh38
rs769135819 Chr.12: 21801000-21801000 on Build GRCh38

View Full Product Details