Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 4769737-4770249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AJAP1
Entrez Gene ID
55966
Gene Name
adherens junctions associated protein 1
Gene Aliases
MOT8, SHREW-1, SHREW1
UniGene
Hs.12473
Gene Chromosome Location
Chr.1: 4654775-4792518 on Build GRCh38
Transcripts
NM_001042478.1 NM_018836.3 XM_011541786.2 XM_011541787.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7354982 c.880A>G p.I294V Chr.1: 4769903-4769903 on Build GRCh38
6127026 c.849C>A p.I283I Chr.1: 4769872-4769872 on Build GRCh38
910430 c.869T>A p.L290H Chr.1: 4769892-4769892 on Build GRCh38
278728 c.857T>C p.I286T Chr.1: 4769880-4769880 on Build GRCh38
5089624 c.902T>C p.L301P Chr.1: 4769925-4769925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767919803 Chr.1: 4769974-4769976 on Build GRCh38
rs531970793 Chr.1: 4770122-4770122 on Build GRCh38
rs890664507 Chr.1: 4770220-4770220 on Build GRCh38
rs1054558134 Chr.1: 4770043-4770043 on Build GRCh38
rs115280211 Chr.1: 4770051-4770051 on Build GRCh38
rs549943362 Chr.1: 4770110-4770110 on Build GRCh38
rs745633032 Chr.1: 4769962-4769962 on Build GRCh38
rs1045343452 Chr.1: 4770009-4770009 on Build GRCh38
rs949298581 Chr.1: 4769995-4769995 on Build GRCh38
rs765475216 Chr.1: 4770114-4770114 on Build GRCh38
rs926501404 Chr.1: 4770179-4770179 on Build GRCh38
rs982374682 Chr.1: 4769978-4769978 on Build GRCh38
rs945157048 Chr.1: 4770190-4770190 on Build GRCh38
rs575870855 Chr.1: 4769928-4769928 on Build GRCh38
rs144818013 Chr.1: 4769903-4769903 on Build GRCh38
rs750590324 Chr.1: 4769885-4769885 on Build GRCh38
rs774959047 Chr.1: 4769904-4769904 on Build GRCh38
rs867488858 Chr.1: 4769858-4769858 on Build GRCh38
rs377403165 Chr.1: 4769849-4769849 on Build GRCh38
rs1026829619 Chr.1: 4770076-4770076 on Build GRCh38
rs770490485 Chr.1: 4769800-4769800 on Build GRCh38
rs201919883 Chr.1: 4769768-4769768 on Build GRCh38
rs868337392 Chr.1: 4770013-4770013 on Build GRCh38
rs760547919 Chr.1: 4769984-4769984 on Build GRCh38
rs370279253 Chr.1: 4770186-4770186 on Build GRCh38
rs779275107 Chr.1: 4769909-4769909 on Build GRCh38
rs936533675 Chr.1: 4770196-4770196 on Build GRCh38
rs775228816 Chr.1: 4769980-4769980 on Build GRCh38
rs377411350 Chr.1: 4770177-4770177 on Build GRCh38
rs926528178 Chr.1: 4770170-4770170 on Build GRCh38
rs527559491 Chr.1: 4770035-4770035 on Build GRCh38
rs1008475914 Chr.1: 4770093-4770093 on Build GRCh38
rs866397254 Chr.1: 4769833-4769833 on Build GRCh38
rs1018435717 Chr.1: 4769870-4769870 on Build GRCh38
rs2072902 Chr.1: 4769821-4769821 on Build GRCh38
rs761790946 Chr.1: 4769871-4769871 on Build GRCh38
rs746618145 Chr.1: 4769955-4769955 on Build GRCh38
rs534197768 Chr.1: 4770197-4770197 on Build GRCh38
rs1024840137 Chr.1: 4770109-4770109 on Build GRCh38
rs369150387 Chr.1: 4769799-4769799 on Build GRCh38
rs575716568 Chr.1: 4770020-4770020 on Build GRCh38
rs776535148 Chr.1: 4769960-4769960 on Build GRCh38
rs919308453 Chr.1: 4769770-4769770 on Build GRCh38
rs541618931 Chr.1: 4769948-4769948 on Build GRCh38
rs189453469 Chr.1: 4770178-4770178 on Build GRCh38
rs563881344 Chr.1: 4770058-4770058 on Build GRCh38
rs769833205 Chr.1: 4769972-4769972 on Build GRCh38
rs899163471 Chr.1: 4770039-4770039 on Build GRCh38
rs370080285 Chr.1: 4769884-4769884 on Build GRCh38
rs377571944 Chr.1: 4769985-4769985 on Build GRCh38
rs780358493 Chr.1: 4769898-4769898 on Build GRCh38
rs996055107 Chr.1: 4770106-4770111 on Build GRCh38
rs777590121 Chr.1: 4769951-4769951 on Build GRCh38
rs573349841 Chr.1: 4770208-4770208 on Build GRCh38
rs771908537 Chr.1: 4769818-4769818 on Build GRCh38
rs756495343 Chr.1: 4769895-4769895 on Build GRCh38
rs770313254 Chr.1: 4770094-4770094 on Build GRCh38
rs754123384 Chr.1: 4769900-4769900 on Build GRCh38
rs762712108 Chr.1: 4770031-4770031 on Build GRCh38
rs554664356 Chr.1: 4770086-4770086 on Build GRCh38
rs184473115 Chr.1: 4769774-4769774 on Build GRCh38
rs373712306 Chr.1: 4769771-4769771 on Build GRCh38
rs949302738 Chr.1: 4769789-4769789 on Build GRCh38
rs770802565 Chr.1: 4769827-4769827 on Build GRCh38
rs201471947 Chr.1: 4769820-4769820 on Build GRCh38
rs553844963 Chr.1: 4769795-4769795 on Build GRCh38
rs2072901 Chr.1: 4769775-4769775 on Build GRCh38
rs568643977 Chr.1: 4769783-4769783 on Build GRCh38
rs980722325 Chr.1: 4770161-4770161 on Build GRCh38
rs570714770 Chr.1: 4770215-4770215 on Build GRCh38
rs536613641 Chr.1: 4770142-4770142 on Build GRCh38
rs1001625967 Chr.1: 4769779-4769779 on Build GRCh38
rs1010241634 Chr.1: 4770059-4770059 on Build GRCh38
rs758990703 Chr.1: 4770171-4770171 on Build GRCh38
rs945569303 Chr.1: 4769923-4769923 on Build GRCh38
rs1046979525 Chr.1: 4769765-4769765 on Build GRCh38
rs533070992 Chr.1: 4770087-4770087 on Build GRCh38
rs569427006 Chr.1: 4769772-4769772 on Build GRCh38
rs383231 Chr.1: 4770204-4770204 on Build GRCh38
rs200921379 Chr.1: 4769957-4769957 on Build GRCh38
rs555946725 Chr.1: 4769828-4769828 on Build GRCh38
rs992891401 Chr.1: 4770108-4770108 on Build GRCh38
rs569765759 Chr.1: 4770113-4770113 on Build GRCh38
rs890611334 Chr.1: 4770001-4770001 on Build GRCh38
rs534232162 Chr.1: 4769790-4769790 on Build GRCh38
rs79867902 Chr.1: 4770078-4770078 on Build GRCh38

View Full Product Details