Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49956412-49956904 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP2
Entrez Gene ID
359
Gene Name
aquaporin 2
Gene Aliases
AQP-CD, WCH-CD
UniGene
Hs.130730
Gene Chromosome Location
Chr.12: 49950741-49958881 on Build GRCh38
Transcripts
NM_000486.5 NR_110590.1 NR_110591.1

Gene Symbol
LOC101927318
Entrez Gene ID
101927318
Gene Name
uncharacterized LOC101927318
Gene Aliases
-
UniGene
Hs.668233
Gene Chromosome Location
Chr.12: 49951512-49962924 on Build GRCh38
Transcripts
NM_000486.5 NR_110590.1 NR_110591.1

SNP Detail

SNP ID Chromosome Location
rs761696713 Chr.12: 49956629-49956629 on Build GRCh38
rs146499105 Chr.12: 49956654-49956654 on Build GRCh38
rs886049548 Chr.12: 49956551-49956551 on Build GRCh38
rs867574338 Chr.12: 49956569-49956569 on Build GRCh38
rs771673301 Chr.12: 49956854-49956854 on Build GRCh38
rs946083576 Chr.12: 49956673-49956673 on Build GRCh38
rs893204856 Chr.12: 49956635-49956635 on Build GRCh38
rs886049549 Chr.12: 49956689-49956689 on Build GRCh38
rs922187935 Chr.12: 49956551-49956551 on Build GRCh38
rs926539763 Chr.12: 49956883-49956883 on Build GRCh38
rs980680714 Chr.12: 49956568-49956568 on Build GRCh38
rs143529505 Chr.12: 49956631-49956631 on Build GRCh38
rs895550551 Chr.12: 49956704-49956704 on Build GRCh38
rs886049550 Chr.12: 49956815-49956815 on Build GRCh38
rs1041841696 Chr.12: 49956675-49956675 on Build GRCh38
rs542433413 Chr.12: 49956702-49956702 on Build GRCh38
rs149776268 Chr.12: 49956516-49956516 on Build GRCh38
rs968339953 Chr.12: 49956829-49956829 on Build GRCh38
rs953637875 Chr.12: 49956845-49956845 on Build GRCh38
rs372756747 Chr.12: 49956584-49956584 on Build GRCh38
rs60887132 Chr.12: 49956515-49956515 on Build GRCh38
rs886049547 Chr.12: 49956496-49956496 on Build GRCh38
rs1001317597 Chr.12: 49956689-49956689 on Build GRCh38
rs577861581 Chr.12: 49956571-49956571 on Build GRCh38
rs981054186 Chr.12: 49956873-49956873 on Build GRCh38
rs139933330 Chr.12: 49956487-49956487 on Build GRCh38
rs764385692 Chr.12: 49956454-49956454 on Build GRCh38
rs1003880111 Chr.12: 49956827-49956827 on Build GRCh38
rs186839162 Chr.12: 49956695-49956695 on Build GRCh38
rs982509005 Chr.12: 49956517-49956517 on Build GRCh38
rs770864493 Chr.12: 49956658-49956658 on Build GRCh38
rs557230385 Chr.12: 49956682-49956682 on Build GRCh38
rs1010380212 Chr.12: 49956739-49956739 on Build GRCh38
rs145603433 Chr.12: 49956519-49956519 on Build GRCh38
rs987637162 Chr.12: 49956871-49956871 on Build GRCh38
rs960095940 Chr.12: 49956472-49956472 on Build GRCh38
rs886049546 Chr.12: 49956450-49956450 on Build GRCh38
rs374844138 Chr.12: 49956491-49956491 on Build GRCh38
rs974178535 Chr.12: 49956485-49956485 on Build GRCh38
rs145489632 Chr.12: 49956495-49956495 on Build GRCh38
rs930651311 Chr.12: 49956630-49956630 on Build GRCh38
rs1027038404 Chr.12: 49956812-49956812 on Build GRCh38
rs1020396845 Chr.12: 49956503-49956503 on Build GRCh38
rs1029239319 Chr.12: 49956839-49956839 on Build GRCh38
rs1020045741 Chr.12: 49956786-49956786 on Build GRCh38
rs995169981 Chr.12: 49956780-49956780 on Build GRCh38
rs150878207 Chr.12: 49956481-49956481 on Build GRCh38
rs560850476 Chr.12: 49956526-49956526 on Build GRCh38
rs886049551 Chr.12: 49956851-49956851 on Build GRCh38
rs900107693 Chr.12: 49956770-49956770 on Build GRCh38
rs1018519398 Chr.12: 49956717-49956717 on Build GRCh38
rs1048258162 Chr.12: 49956672-49956672 on Build GRCh38
rs776361548 Chr.12: 49956696-49956696 on Build GRCh38
rs148904180 Chr.12: 49956609-49956609 on Build GRCh38
rs749447331 Chr.12: 49956451-49956451 on Build GRCh38
rs1035962828 Chr.12: 49956699-49956699 on Build GRCh38
rs376984675 Chr.12: 49956826-49956826 on Build GRCh38
rs541028359 Chr.12: 49956846-49956846 on Build GRCh38
rs147666604 Chr.12: 49956509-49956509 on Build GRCh38
rs77904078 Chr.12: 49956836-49956836 on Build GRCh38

View Full Product Details