Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49956794-49957291 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP2
Entrez Gene ID
359
Gene Name
aquaporin 2
Gene Aliases
AQP-CD, WCH-CD
UniGene
Hs.130730
Gene Chromosome Location
Chr.12: 49950741-49958881 on Build GRCh38
Transcripts
NM_000486.5 NR_110590.1 NR_110591.1

Gene Symbol
LOC101927318
Entrez Gene ID
101927318
Gene Name
uncharacterized LOC101927318
Gene Aliases
-
UniGene
Hs.668233
Gene Chromosome Location
Chr.12: 49951512-49962924 on Build GRCh38
Transcripts
NM_000486.5 NR_110590.1 NR_110591.1

SNP Detail

SNP ID Chromosome Location
rs914551036 Chr.12: 49957001-49957001 on Build GRCh38
rs900010855 Chr.12: 49957083-49957083 on Build GRCh38
rs142248293 Chr.12: 49957259-49957259 on Build GRCh38
rs771673301 Chr.12: 49956854-49956854 on Build GRCh38
rs1040305364 Chr.12: 49957046-49957046 on Build GRCh38
rs926539763 Chr.12: 49956883-49956883 on Build GRCh38
rs989423908 Chr.12: 49956990-49956990 on Build GRCh38
rs138508276 Chr.12: 49957150-49957150 on Build GRCh38
rs144781042 Chr.12: 49957074-49957074 on Build GRCh38
rs968339953 Chr.12: 49956829-49956829 on Build GRCh38
rs138918672 Chr.12: 49957016-49957016 on Build GRCh38
rs953637875 Chr.12: 49956845-49956845 on Build GRCh38
rs772839440 Chr.12: 49957142-49957142 on Build GRCh38
rs150827515 Chr.12: 49957267-49957267 on Build GRCh38
rs920681801 Chr.12: 49956962-49956962 on Build GRCh38
rs970473412 Chr.12: 49957203-49957203 on Build GRCh38
rs912022096 Chr.12: 49956885-49956885 on Build GRCh38
rs996139104 Chr.12: 49957130-49957130 on Build GRCh38
rs750350039 Chr.12: 49956964-49956964 on Build GRCh38
rs144704751 Chr.12: 49956987-49956987 on Build GRCh38
rs981054186 Chr.12: 49956873-49956873 on Build GRCh38
rs150302183 Chr.12: 49957015-49957015 on Build GRCh38
rs1003880111 Chr.12: 49956827-49956827 on Build GRCh38
rs944217295 Chr.12: 49957035-49957035 on Build GRCh38
rs1010081818 Chr.12: 49957168-49957168 on Build GRCh38
rs987637162 Chr.12: 49956871-49956871 on Build GRCh38
rs140054228 Chr.12: 49957003-49957003 on Build GRCh38
rs140840803 Chr.12: 49957246-49957246 on Build GRCh38
rs143902080 Chr.12: 49957154-49957154 on Build GRCh38
rs34780794 Chr.12: 49957233-49957234 on Build GRCh38
rs1029239319 Chr.12: 49956839-49956839 on Build GRCh38
rs905775984 Chr.12: 49957036-49957036 on Build GRCh38
rs149432877 Chr.12: 49957032-49957032 on Build GRCh38
rs937279347 Chr.12: 49957058-49957058 on Build GRCh38
rs999848112 Chr.12: 49957208-49957208 on Build GRCh38
rs296766 Chr.12: 49957170-49957170 on Build GRCh38
rs138521926 Chr.12: 49956993-49956993 on Build GRCh38
rs1041787870 Chr.12: 49957009-49957009 on Build GRCh38
rs1003870285 Chr.12: 49957169-49957169 on Build GRCh38
rs886049551 Chr.12: 49956851-49956851 on Build GRCh38
rs972523833 Chr.12: 49956940-49956940 on Build GRCh38
rs879585566 Chr.12: 49957212-49957212 on Build GRCh38
rs886049552 Chr.12: 49957207-49957207 on Build GRCh38
rs1033900150 Chr.12: 49957230-49957230 on Build GRCh38
rs904314398 Chr.12: 49957207-49957207 on Build GRCh38
rs376984675 Chr.12: 49956826-49956826 on Build GRCh38
rs958221212 Chr.12: 49956923-49956923 on Build GRCh38
rs541028359 Chr.12: 49956846-49956846 on Build GRCh38
rs77904078 Chr.12: 49956836-49956836 on Build GRCh38

View Full Product Details