Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 49958152-49958646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AQP2
Entrez Gene ID
359
Gene Name
aquaporin 2
Gene Aliases
AQP-CD, WCH-CD
UniGene
Hs.130730
Gene Chromosome Location
Chr.12: 49950741-49958881 on Build GRCh38
Transcripts
NM_000486.5 NR_110590.1 NR_110591.1

Gene Symbol
LOC101927318
Entrez Gene ID
101927318
Gene Name
uncharacterized LOC101927318
Gene Aliases
-
UniGene
Hs.668233
Gene Chromosome Location
Chr.12: 49951512-49962924 on Build GRCh38
Transcripts
NM_000486.5 NR_110590.1 NR_110591.1

SNP Detail

SNP ID Chromosome Location
rs2336709 Chr.12: 49958567-49958567 on Build GRCh38
rs146241781 Chr.12: 49958591-49958591 on Build GRCh38
rs952858239 Chr.12: 49958324-49958324 on Build GRCh38
rs552569294 Chr.12: 49958352-49958352 on Build GRCh38
rs2878771 Chr.12: 49958610-49958610 on Build GRCh38
rs138229213 Chr.12: 49958449-49958449 on Build GRCh38
rs529322978 Chr.12: 49958314-49958314 on Build GRCh38
rs928974116 Chr.12: 49958542-49958542 on Build GRCh38
rs376235969 Chr.12: 49958221-49958221 on Build GRCh38
rs34704996 Chr.12: 49958580-49958581 on Build GRCh38
rs928271258 Chr.12: 49958463-49958463 on Build GRCh38
rs147733377 Chr.12: 49958239-49958239 on Build GRCh38
rs893533041 Chr.12: 49958558-49958558 on Build GRCh38
rs1054823124 Chr.12: 49958538-49958538 on Build GRCh38
rs530539331 Chr.12: 49958477-49958477 on Build GRCh38
rs985943047 Chr.12: 49958503-49958503 on Build GRCh38
rs148876709 Chr.12: 49958223-49958223 on Build GRCh38
rs968657633 Chr.12: 49958608-49958608 on Build GRCh38
rs142738661 Chr.12: 49958480-49958480 on Build GRCh38
rs910363723 Chr.12: 49958517-49958517 on Build GRCh38
rs1077521 Chr.12: 49958403-49958403 on Build GRCh38
rs886049553 Chr.12: 49958412-49958412 on Build GRCh38
rs965753390 Chr.12: 49958290-49958290 on Build GRCh38
rs1077520 Chr.12: 49958333-49958333 on Build GRCh38
rs570342310 Chr.12: 49958256-49958256 on Build GRCh38
rs145783831 Chr.12: 49958230-49958230 on Build GRCh38
rs886049554 Chr.12: 49958546-49958546 on Build GRCh38
rs139949172 Chr.12: 49958488-49958488 on Build GRCh38
rs1049265944 Chr.12: 49958247-49958247 on Build GRCh38
rs113903023 Chr.12: 49958446-49958446 on Build GRCh38
rs896616338 Chr.12: 49958618-49958618 on Build GRCh38
rs187623325 Chr.12: 49958534-49958534 on Build GRCh38
rs563819167 Chr.12: 49958224-49958224 on Build GRCh38
rs957301964 Chr.12: 49958464-49958464 on Build GRCh38
rs1021463287 Chr.12: 49958412-49958412 on Build GRCh38
rs146060042 Chr.12: 49958487-49958487 on Build GRCh38
rs141894535 Chr.12: 49958524-49958524 on Build GRCh38
rs147698325 Chr.12: 49958572-49958572 on Build GRCh38
rs975480604 Chr.12: 49958296-49958296 on Build GRCh38
rs1032920617 Chr.12: 49958450-49958450 on Build GRCh38
rs1020771314 Chr.12: 49958198-49958198 on Build GRCh38
rs928982256 Chr.12: 49958235-49958235 on Build GRCh38
rs944552267 Chr.12: 49958519-49958519 on Build GRCh38
rs141457763 Chr.12: 49958444-49958444 on Build GRCh38
rs142648983 Chr.12: 49958323-49958323 on Build GRCh38
rs919005317 Chr.12: 49958539-49958539 on Build GRCh38
rs1048751748 Chr.12: 49958545-49958545 on Build GRCh38

View Full Product Details