Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51953302-51953812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR1B
Entrez Gene ID
91
Gene Name
activin A receptor type 1B
Gene Aliases
ACTRIB, ACVRLK4, ALK4, SKR2
UniGene
Hs.438918
Gene Chromosome Location
Chr.12: 51951667-51997079 on Build GRCh38
Transcripts
NM_004302.4 NM_020327.3 NM_020328.3 XM_011538966.2 XM_011538967.2 XM_017020199.1 XM_017020200.1 XM_017020201.1

SNP Detail

SNP ID Chromosome Location
rs11404836 Chr.12: 51953594-51953595 on Build GRCh38
rs758802561 Chr.12: 51953610-51953610 on Build GRCh38
rs918186235 Chr.12: 51953729-51953729 on Build GRCh38
rs943718829 Chr.12: 51953401-51953401 on Build GRCh38
rs1006720345 Chr.12: 51953553-51953553 on Build GRCh38
rs9634266 Chr.12: 51953673-51953673 on Build GRCh38
rs533249956 Chr.12: 51953742-51953742 on Build GRCh38
rs751466818 Chr.12: 51953617-51953617 on Build GRCh38
rs1029605585 Chr.12: 51953521-51953521 on Build GRCh38
rs113610098 Chr.12: 51953524-51953524 on Build GRCh38
rs896713291 Chr.12: 51953392-51953392 on Build GRCh38
rs1018270152 Chr.12: 51953535-51953535 on Build GRCh38
rs1032882788 Chr.12: 51953475-51953475 on Build GRCh38
rs545925219 Chr.12: 51953437-51953437 on Build GRCh38
rs1042401560 Chr.12: 51953510-51953510 on Build GRCh38
rs779181691 Chr.12: 51953527-51953527 on Build GRCh38
rs896575737 Chr.12: 51953497-51953497 on Build GRCh38
rs1019414175 Chr.12: 51953575-51953575 on Build GRCh38
rs912299208 Chr.12: 51953390-51953390 on Build GRCh38
rs576318004 Chr.12: 51953492-51953492 on Build GRCh38
rs953886952 Chr.12: 51953533-51953533 on Build GRCh38
rs948316923 Chr.12: 51953774-51953774 on Build GRCh38
rs909409923 Chr.12: 51953473-51953473 on Build GRCh38
rs1014354751 Chr.12: 51953417-51953417 on Build GRCh38
rs185729781 Chr.12: 51953338-51953338 on Build GRCh38
rs999938719 Chr.12: 51953446-51953446 on Build GRCh38
rs535443412 Chr.12: 51953339-51953339 on Build GRCh38
rs147277864 Chr.12: 51953439-51953439 on Build GRCh38
rs1010037190 Chr.12: 51953506-51953506 on Build GRCh38
rs544570541 Chr.12: 51953595-51953596 on Build GRCh38
rs902481241 Chr.12: 51953520-51953520 on Build GRCh38
rs972860465 Chr.12: 51953671-51953671 on Build GRCh38
rs985896541 Chr.12: 51953532-51953532 on Build GRCh38
rs963160110 Chr.12: 51953557-51953557 on Build GRCh38
rs188997574 Chr.12: 51953644-51953644 on Build GRCh38
rs768968606 Chr.12: 51953453-51953453 on Build GRCh38
rs938169410 Chr.12: 51953496-51953496 on Build GRCh38
rs1056652167 Chr.12: 51953372-51953372 on Build GRCh38
rs149933291 Chr.12: 51953676-51953676 on Build GRCh38
rs565817940 Chr.12: 51953328-51953328 on Build GRCh38
rs562134421 Chr.12: 51953639-51953639 on Build GRCh38
rs796941507 Chr.12: 51953713-51953713 on Build GRCh38
rs398055575 Chr.12: 51953608-51953609 on Build GRCh38
rs766692511 Chr.12: 51953419-51953419 on Build GRCh38

View Full Product Details