Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51982603-51983109 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR1B
Entrez Gene ID
91
Gene Name
activin A receptor type 1B
Gene Aliases
ACTRIB, ACVRLK4, ALK4, SKR2
UniGene
Hs.438918
Gene Chromosome Location
Chr.12: 51951667-51997079 on Build GRCh38
Transcripts
NM_004302.4 NM_020327.3 NM_020328.3 XM_011538966.2 XM_011538967.2 XM_017020199.1 XM_017020200.1 XM_017020201.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5636675 c.840G>A p.W280* Chr.12: 51982714-51982714 on Build GRCh38
4819415 c.917G>T p.R306I Chr.12: 51982791-51982791 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs149714823 Chr.12: 51983008-51983008 on Build GRCh38
rs921256096 Chr.12: 51982913-51982913 on Build GRCh38
rs987647105 Chr.12: 51982705-51982705 on Build GRCh38
rs191020997 Chr.12: 51982739-51982739 on Build GRCh38
rs1004307336 Chr.12: 51982964-51982964 on Build GRCh38
rs906742414 Chr.12: 51982972-51982972 on Build GRCh38
rs779924865 Chr.12: 51982734-51982734 on Build GRCh38
rs1037612606 Chr.12: 51982967-51982967 on Build GRCh38
rs932683234 Chr.12: 51982937-51982937 on Build GRCh38
rs962112771 Chr.12: 51982753-51982753 on Build GRCh38
rs1041174024 Chr.12: 51982910-51982910 on Build GRCh38
rs757217593 Chr.12: 51982866-51982866 on Build GRCh38
rs927055835 Chr.12: 51982930-51982930 on Build GRCh38
rs891654380 Chr.12: 51983012-51983012 on Build GRCh38
rs117766220 Chr.12: 51982883-51982883 on Build GRCh38
rs958434031 Chr.12: 51982836-51982836 on Build GRCh38
rs780803213 Chr.12: 51982636-51982636 on Build GRCh38
rs2172603 Chr.12: 51983028-51983028 on Build GRCh38
rs750557999 Chr.12: 51982992-51982992 on Build GRCh38
rs973642710 Chr.12: 51982853-51982853 on Build GRCh38
rs1021523989 Chr.12: 51983043-51983043 on Build GRCh38
rs140512254 Chr.12: 51982830-51982830 on Build GRCh38
rs758879315 Chr.12: 51982923-51982924 on Build GRCh38
rs547955784 Chr.12: 51983050-51983050 on Build GRCh38
rs1014651549 Chr.12: 51982745-51982745 on Build GRCh38
rs772140419 Chr.12: 51982772-51982772 on Build GRCh38
rs557422924 Chr.12: 51982710-51982710 on Build GRCh38
rs551375052 Chr.12: 51982733-51982733 on Build GRCh38
rs888601587 Chr.12: 51982727-51982727 on Build GRCh38
rs1029082949 Chr.12: 51982696-51982696 on Build GRCh38
rs1027226983 Chr.12: 51982725-51982725 on Build GRCh38
rs997051385 Chr.12: 51982700-51982700 on Build GRCh38
rs562905372 Chr.12: 51982848-51982848 on Build GRCh38
rs1003028931 Chr.12: 51982978-51982978 on Build GRCh38
rs977266977 Chr.12: 51982882-51982882 on Build GRCh38
rs1048606126 Chr.12: 51982940-51982940 on Build GRCh38
rs888714282 Chr.12: 51982946-51982946 on Build GRCh38
rs1002507628 Chr.12: 51982770-51982770 on Build GRCh38
rs540762560 Chr.12: 51982682-51982682 on Build GRCh38
rs919564044 Chr.12: 51982856-51982856 on Build GRCh38
rs944121892 Chr.12: 51982902-51982902 on Build GRCh38
rs773517703 Chr.12: 51982806-51982806 on Build GRCh38
rs962248820 Chr.12: 51982834-51982834 on Build GRCh38
rs767578218 Chr.12: 51982941-51982941 on Build GRCh38
rs542517915 Chr.12: 51983006-51983006 on Build GRCh38
rs533142333 Chr.12: 51982702-51982702 on Build GRCh38
rs938459834 Chr.12: 51982938-51982938 on Build GRCh38
rs968515897 Chr.12: 51982875-51982875 on Build GRCh38

View Full Product Details