Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52320862-52321367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT83
Entrez Gene ID
3889
Gene Name
keratin 83
Gene Aliases
HB3, Hb-3, KRTHB3
UniGene
Hs.720768
Gene Chromosome Location
Chr.12: 52314301-52321398 on Build GRCh38
Transcripts
NM_002282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
110402 c.204C>T p.F68F Chr.12: 52321132-52321132 on Build GRCh38
3462221 c.170G>A p.G57E Chr.12: 52321166-52321166 on Build GRCh38
3462219 c.328G>A p.E110K Chr.12: 52321008-52321008 on Build GRCh38
1705630 c.218G>A p.G73E Chr.12: 52321118-52321118 on Build GRCh38
7114337 c.281C>T p.T94M Chr.12: 52321055-52321055 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549134397 Chr.12: 52321036-52321036 on Build GRCh38
rs754356225 Chr.12: 52321175-52321175 on Build GRCh38
rs764292091 Chr.12: 52321136-52321136 on Build GRCh38
rs752994861 Chr.12: 52321081-52321081 on Build GRCh38
rs758097210 Chr.12: 52321191-52321191 on Build GRCh38
rs772461137 Chr.12: 52321218-52321218 on Build GRCh38
rs750995033 Chr.12: 52321147-52321147 on Build GRCh38
rs771358344 Chr.12: 52320994-52320994 on Build GRCh38
rs894390168 Chr.12: 52321023-52321023 on Build GRCh38
rs200833070 Chr.12: 52321257-52321257 on Build GRCh38
rs1051771662 Chr.12: 52320991-52320991 on Build GRCh38
rs773139403 Chr.12: 52321115-52321115 on Build GRCh38
rs990215562 Chr.12: 52321130-52321130 on Build GRCh38
rs749161579 Chr.12: 52321259-52321259 on Build GRCh38
rs760039172 Chr.12: 52321235-52321235 on Build GRCh38
rs756638517 Chr.12: 52321151-52321151 on Build GRCh38
rs777374672 Chr.12: 52321192-52321192 on Build GRCh38
rs573946841 Chr.12: 52321172-52321172 on Build GRCh38
rs201594412 Chr.12: 52321240-52321240 on Build GRCh38
rs751158922 Chr.12: 52321248-52321248 on Build GRCh38
rs111332863 Chr.12: 52320897-52320897 on Build GRCh38
rs762816475 Chr.12: 52321217-52321217 on Build GRCh38
rs558016037 Chr.12: 52321094-52321094 on Build GRCh38
rs542814857 Chr.12: 52320887-52320887 on Build GRCh38
rs747873972 Chr.12: 52321313-52321313 on Build GRCh38
rs1054467808 Chr.12: 52320942-52320942 on Build GRCh38
rs748989577 Chr.12: 52321099-52321099 on Build GRCh38
rs751797447 Chr.12: 52321140-52321140 on Build GRCh38
rs752308926 Chr.12: 52320926-52320926 on Build GRCh38
rs765642444 Chr.12: 52321236-52321236 on Build GRCh38
rs774373912 Chr.12: 52320902-52320902 on Build GRCh38
rs754334165 Chr.12: 52321231-52321231 on Build GRCh38
rs758760872 Chr.12: 52321241-52321241 on Build GRCh38
rs367891695 Chr.12: 52321069-52321069 on Build GRCh38
rs754825251 Chr.12: 52321310-52321310 on Build GRCh38
rs779172264 Chr.12: 52321026-52321026 on Build GRCh38
rs773581872 Chr.12: 52321266-52321266 on Build GRCh38
rs777675184 Chr.12: 52321030-52321030 on Build GRCh38
rs755572344 Chr.12: 52321300-52321300 on Build GRCh38
rs751265769 Chr.12: 52321260-52321269 on Build GRCh38
rs775302468 Chr.12: 52321212-52321212 on Build GRCh38
rs930417357 Chr.12: 52321178-52321178 on Build GRCh38
rs376391383 Chr.12: 52320955-52320955 on Build GRCh38
rs773559995 Chr.12: 52320915-52320915 on Build GRCh38
rs749177729 Chr.12: 52321308-52321308 on Build GRCh38
rs760565887 Chr.12: 52321119-52321119 on Build GRCh38
rs141964696 Chr.12: 52321158-52321158 on Build GRCh38
rs149947658 Chr.12: 52321298-52321298 on Build GRCh38
rs775460575 Chr.12: 52321029-52321030 on Build GRCh38
rs200214916 Chr.12: 52321343-52321343 on Build GRCh38
rs550281827 Chr.12: 52321312-52321312 on Build GRCh38
rs751435724 Chr.12: 52320943-52320943 on Build GRCh38
rs201909879 Chr.12: 52321025-52321025 on Build GRCh38
rs778809722 Chr.12: 52321311-52321311 on Build GRCh38
rs146287955 Chr.12: 52321263-52321263 on Build GRCh38
rs764380315 Chr.12: 52321286-52321286 on Build GRCh38
rs900271515 Chr.12: 52321031-52321031 on Build GRCh38
rs747408486 Chr.12: 52320986-52320986 on Build GRCh38
rs748675592 Chr.12: 52320909-52320909 on Build GRCh38
rs776735812 Chr.12: 52321303-52321303 on Build GRCh38
rs775280849 Chr.12: 52321009-52321009 on Build GRCh38
rs754254775 Chr.12: 52321224-52321224 on Build GRCh38
rs776341669 Chr.12: 52321131-52321131 on Build GRCh38
rs761189505 Chr.12: 52321138-52321139 on Build GRCh38
rs61485872 Chr.12: 52321269-52321269 on Build GRCh38
rs746315687 Chr.12: 52321095-52321095 on Build GRCh38
rs765840475 Chr.12: 52321282-52321282 on Build GRCh38
rs745755826 Chr.12: 52321204-52321204 on Build GRCh38
rs895315297 Chr.12: 52321221-52321221 on Build GRCh38
rs971315230 Chr.12: 52321087-52321087 on Build GRCh38
rs977098433 Chr.12: 52321253-52321253 on Build GRCh38
rs551528470 Chr.12: 52320928-52320928 on Build GRCh38
rs746021183 Chr.12: 52321331-52321331 on Build GRCh38
rs770273202 Chr.12: 52321043-52321043 on Build GRCh38
rs747396835 Chr.12: 52320914-52320914 on Build GRCh38
rs747770852 Chr.12: 52321156-52321156 on Build GRCh38
rs747575012 Chr.12: 52321033-52321033 on Build GRCh38
rs777462202 Chr.12: 52321245-52321245 on Build GRCh38
rs757428966 Chr.12: 52321143-52321143 on Build GRCh38
rs759508548 Chr.12: 52320989-52320992 on Build GRCh38
rs769503856 Chr.12: 52321211-52321211 on Build GRCh38
rs758512053 Chr.12: 52321028-52321028 on Build GRCh38
rs886159537 Chr.12: 52321195-52321195 on Build GRCh38
rs563212614 Chr.12: 52321003-52321005 on Build GRCh38
rs757758399 Chr.12: 52321227-52321228 on Build GRCh38
rs769578632 Chr.12: 52321167-52321167 on Build GRCh38
rs936031259 Chr.12: 52320918-52320918 on Build GRCh38
rs766108026 Chr.12: 52321257-52321258 on Build GRCh38
rs767320922 Chr.12: 52321022-52321022 on Build GRCh38
rs781009358 Chr.12: 52321197-52321197 on Build GRCh38
rs763324093 Chr.12: 52321283-52321283 on Build GRCh38
rs143776387 Chr.12: 52321017-52321017 on Build GRCh38
rs201373391 Chr.12: 52321008-52321008 on Build GRCh38
rs957481705 Chr.12: 52321125-52321125 on Build GRCh38
rs757410511 Chr.12: 52321089-52321089 on Build GRCh38
rs777091313 Chr.12: 52320999-52320999 on Build GRCh38
rs776629118 Chr.12: 52321163-52321163 on Build GRCh38
rs771602377 Chr.12: 52321157-52321157 on Build GRCh38
rs886049622 Chr.12: 52321253-52321253 on Build GRCh38
rs766424277 Chr.12: 52321227-52321227 on Build GRCh38
rs780705567 Chr.12: 52321275-52321276 on Build GRCh38
rs759961703 Chr.12: 52321001-52321001 on Build GRCh38
rs571144595 Chr.12: 52320973-52320973 on Build GRCh38
rs762443407 Chr.12: 52321046-52321046 on Build GRCh38
rs935645175 Chr.12: 52321201-52321218 on Build GRCh38
rs766097425 Chr.12: 52321080-52321080 on Build GRCh38
rs200595102 Chr.12: 52320967-52320967 on Build GRCh38
rs978593553 Chr.12: 52321253-52321254 on Build GRCh38
rs199997738 Chr.12: 52321144-52321144 on Build GRCh38
rs774437079 Chr.12: 52321168-52321168 on Build GRCh38
rs538643123 Chr.12: 52321076-52321076 on Build GRCh38
rs565969399 Chr.12: 52321041-52321041 on Build GRCh38
rs766185773 Chr.12: 52321124-52321124 on Build GRCh38
rs559602201 Chr.12: 52320916-52320916 on Build GRCh38
rs374309148 Chr.12: 52321068-52321068 on Build GRCh38
rs769816860 Chr.12: 52321256-52321256 on Build GRCh38
rs780780227 Chr.12: 52320957-52320957 on Build GRCh38
rs537011326 Chr.12: 52321113-52321113 on Build GRCh38
rs768240052 Chr.12: 52321058-52321058 on Build GRCh38
rs138833898 Chr.12: 52321114-52321114 on Build GRCh38
rs750577277 Chr.12: 52321174-52321174 on Build GRCh38
rs751634646 Chr.12: 52321086-52321086 on Build GRCh38
rs755310254 Chr.12: 52321186-52321186 on Build GRCh38
rs778487552 Chr.12: 52321155-52321155 on Build GRCh38
rs570205730 Chr.12: 52321018-52321018 on Build GRCh38
rs148290789 Chr.12: 52321132-52321132 on Build GRCh38
rs749653752 Chr.12: 52321152-52321152 on Build GRCh38
rs1025233131 Chr.12: 52321232-52321232 on Build GRCh38
rs542978203 Chr.12: 52321189-52321189 on Build GRCh38
rs756727430 Chr.12: 52321250-52321250 on Build GRCh38
rs779083731 Chr.12: 52320970-52320970 on Build GRCh38
rs771394395 Chr.12: 52321035-52321035 on Build GRCh38
rs368221217 Chr.12: 52321098-52321098 on Build GRCh38
rs756881584 Chr.12: 52321291-52321291 on Build GRCh38
rs749987201 Chr.12: 52321299-52321299 on Build GRCh38
rs780709406 Chr.12: 52321254-52321254 on Build GRCh38
rs150640038 Chr.12: 52321045-52321045 on Build GRCh38
rs764162535 Chr.12: 52320929-52320929 on Build GRCh38
rs759312131 Chr.12: 52321166-52321166 on Build GRCh38
rs751248490 Chr.12: 52321288-52321288 on Build GRCh38
rs776714515 Chr.12: 52321329-52321329 on Build GRCh38
rs778359333 Chr.12: 52320979-52320979 on Build GRCh38
rs764865449 Chr.12: 52320923-52320923 on Build GRCh38
rs558452885 Chr.12: 52321222-52321222 on Build GRCh38
rs745810779 Chr.12: 52321255-52321255 on Build GRCh38
rs763451943 Chr.12: 52321013-52321013 on Build GRCh38
rs752369680 Chr.12: 52321190-52321190 on Build GRCh38
rs201780536 Chr.12: 52320917-52320917 on Build GRCh38
rs550558420 Chr.12: 52321012-52321012 on Build GRCh38
rs780196908 Chr.12: 52321307-52321307 on Build GRCh38
rs776821705 Chr.12: 52321273-52321273 on Build GRCh38
rs150875897 Chr.12: 52321097-52321097 on Build GRCh38
rs747188137 Chr.12: 52321267-52321267 on Build GRCh38
rs768240634 Chr.12: 52321100-52321100 on Build GRCh38
rs746592309 Chr.12: 52321194-52321194 on Build GRCh38
rs774021555 Chr.12: 52321107-52321107 on Build GRCh38
rs764705836 Chr.12: 52321189-52321189 on Build GRCh38
rs767725820 Chr.12: 52321173-52321173 on Build GRCh38
rs746502340 Chr.12: 52321159-52321159 on Build GRCh38
rs774610454 Chr.12: 52320926-52320927 on Build GRCh38
rs111267865 Chr.12: 52321024-52321024 on Build GRCh38
rs754620118 Chr.12: 52321260-52321260 on Build GRCh38
rs771227120 Chr.12: 52321328-52321328 on Build GRCh38
rs755364196 Chr.12: 52321153-52321153 on Build GRCh38
rs752860418 Chr.12: 52321027-52321027 on Build GRCh38
rs770899040 Chr.12: 52321162-52321162 on Build GRCh38

View Full Product Details