Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53314161-53314682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AAAS
Entrez Gene ID
8086
Gene Name
aladin WD repeat nucleoporin
Gene Aliases
AAA, AAASb, ADRACALA, ADRACALIN, ALADIN, GL003
UniGene
Hs.369144
Gene Chromosome Location
Chr.12: 53307456-53321628 on Build GRCh38
Transcripts
NM_001173466.1 NM_015665.5 XM_011538778.1 XM_011538780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
940975 c.614G>A p.S205N Chr.12: 53314373-53314373 on Build GRCh38
3384485 c.648C>G p.C216W Chr.12: 53314339-53314339 on Build GRCh38
4567075 c.555_556CC>TT p.P186S Chr.12: 53314431-53314432 on Build GRCh38
1181130 c.688C>T p.R230* Chr.12: 53314299-53314299 on Build GRCh38
940976 c.581G>A p.R194Q Chr.12: 53314406-53314406 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs80027466 Chr.12: 53314308-53314308 on Build GRCh38
rs1028490510 Chr.12: 53314641-53314641 on Build GRCh38
rs200513971 Chr.12: 53314456-53314456 on Build GRCh38
rs772913826 Chr.12: 53314377-53314377 on Build GRCh38
rs752360800 Chr.12: 53314298-53314298 on Build GRCh38
rs548631510 Chr.12: 53314294-53314294 on Build GRCh38
rs941172886 Chr.12: 53314632-53314632 on Build GRCh38
rs964759127 Chr.12: 53314530-53314530 on Build GRCh38
rs998702356 Chr.12: 53314638-53314638 on Build GRCh38
rs763760167 Chr.12: 53314406-53314406 on Build GRCh38
rs768784941 Chr.12: 53314352-53314352 on Build GRCh38
rs767168679 Chr.12: 53314186-53314186 on Build GRCh38
rs866079099 Chr.12: 53314392-53314392 on Build GRCh38
rs371783768 Chr.12: 53314449-53314449 on Build GRCh38
rs145235560 Chr.12: 53314416-53314416 on Build GRCh38
rs984412481 Chr.12: 53314347-53314347 on Build GRCh38
rs78667074 Chr.12: 53314653-53314653 on Build GRCh38
rs139160221 Chr.12: 53314467-53314467 on Build GRCh38
rs948367683 Chr.12: 53314433-53314433 on Build GRCh38
rs957436344 Chr.12: 53314266-53314266 on Build GRCh38
rs562064426 Chr.12: 53314605-53314605 on Build GRCh38
rs754431109 Chr.12: 53314262-53314262 on Build GRCh38
rs908400676 Chr.12: 53314616-53314616 on Build GRCh38
rs150015340 Chr.12: 53314397-53314397 on Build GRCh38
rs147512475 Chr.12: 53314355-53314355 on Build GRCh38
rs771220747 Chr.12: 53314566-53314566 on Build GRCh38
rs570290321 Chr.12: 53314585-53314585 on Build GRCh38
rs772310506 Chr.12: 53314373-53314373 on Build GRCh38
rs778906427 Chr.12: 53314429-53314429 on Build GRCh38
rs748222885 Chr.12: 53314431-53314431 on Build GRCh38
rs756720428 Chr.12: 53314312-53314312 on Build GRCh38
rs772208074 Chr.12: 53314445-53314445 on Build GRCh38
rs140046968 Chr.12: 53314380-53314380 on Build GRCh38
rs1054947013 Chr.12: 53314500-53314500 on Build GRCh38
rs1031770747 Chr.12: 53314229-53314229 on Build GRCh38
rs901247093 Chr.12: 53314626-53314626 on Build GRCh38
rs78212742 Chr.12: 53314396-53314396 on Build GRCh38
rs999029003 Chr.12: 53314507-53314507 on Build GRCh38
rs1011688479 Chr.12: 53314523-53314524 on Build GRCh38
rs7953255 Chr.12: 53314227-53314227 on Build GRCh38
rs939634100 Chr.12: 53314369-53314369 on Build GRCh38
rs1031714041 Chr.12: 53314234-53314234 on Build GRCh38
rs1021577263 Chr.12: 53314205-53314205 on Build GRCh38
rs77331189 Chr.12: 53314395-53314395 on Build GRCh38
rs758057774 Chr.12: 53314299-53314299 on Build GRCh38
rs770780563 Chr.12: 53314459-53314459 on Build GRCh38
rs201891426 Chr.12: 53314310-53314310 on Build GRCh38
rs1029541319 Chr.12: 53314553-53314553 on Build GRCh38
rs752230551 Chr.12: 53314399-53314399 on Build GRCh38
rs776399575 Chr.12: 53314488-53314488 on Build GRCh38
rs144873582 Chr.12: 53314349-53314349 on Build GRCh38
rs756489312 Chr.12: 53314415-53314415 on Build GRCh38
rs893285758 Chr.12: 53314516-53314516 on Build GRCh38
rs1010800386 Chr.12: 53314198-53314198 on Build GRCh38
rs781457941 Chr.12: 53314300-53314300 on Build GRCh38
rs886049650 Chr.12: 53314324-53314324 on Build GRCh38
rs141013947 Chr.12: 53314320-53314320 on Build GRCh38
rs111686817 Chr.12: 53314515-53314515 on Build GRCh38
rs1018085208 Chr.12: 53314528-53314530 on Build GRCh38
rs894165422 Chr.12: 53314429-53314429 on Build GRCh38
rs796232206 Chr.12: 53314334-53314334 on Build GRCh38
rs904564669 Chr.12: 53314209-53314209 on Build GRCh38
rs796419389 Chr.12: 53314607-53314607 on Build GRCh38
rs533184867 Chr.12: 53314372-53314372 on Build GRCh38
rs1043587785 Chr.12: 53314552-53314552 on Build GRCh38
rs747189660 Chr.12: 53314565-53314565 on Build GRCh38
rs758026430 Chr.12: 53314401-53314401 on Build GRCh38
rs750775106 Chr.12: 53314407-53314407 on Build GRCh38
rs946627146 Chr.12: 53314539-53314539 on Build GRCh38
rs755526295 Chr.12: 53314423-53314423 on Build GRCh38

View Full Product Details