Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53425365-53425861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMHR2
Entrez Gene ID
269
Gene Name
anti-Mullerian hormone receptor type 2
Gene Aliases
AMHR, MISR2, MISRII, MRII
UniGene
Hs.659889
Gene Chromosome Location
Chr.12: 53423855-53431672 on Build GRCh38
Transcripts
NM_001164690.1 NM_001164691.1 NM_020547.2 XM_011538173.1 XM_011538174.1 XM_011538176.1 XM_011538178.1 XM_011538179.1 XM_011538180.1 XM_011538181.1 XM_011538183.2 XM_011538184.2 XM_011538185.1 XM_011538186.2 XM_011538187.2 XM_017019179.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
431394 c.658G>C p.G220R Chr.12: 53425725-53425725 on Build GRCh38
5375474 c.748C>T p.P250S Chr.12: 53425815-53425815 on Build GRCh38
5069583 c.793_794insG p.P268fs*30 Chr.12: 53425860-53425861 on Build GRCh38
7224099 c.622-6C>T p.? Chr.12: 53425683-53425683 on Build GRCh38
5375476 c.794G>A p.R265Q Chr.12: 53425861-53425861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771506798 Chr.12: 53425608-53425608 on Build GRCh38
rs919906447 Chr.12: 53425518-53425518 on Build GRCh38
rs150262652 Chr.12: 53425485-53425485 on Build GRCh38
rs2071558 Chr.12: 53425683-53425683 on Build GRCh38
rs748079944 Chr.12: 53425600-53425600 on Build GRCh38
rs536389361 Chr.12: 53425812-53425812 on Build GRCh38
rs200782636 Chr.12: 53425642-53425642 on Build GRCh38
rs751661136 Chr.12: 53425454-53425454 on Build GRCh38
rs368855951 Chr.12: 53425588-53425588 on Build GRCh38
rs777917652 Chr.12: 53425444-53425444 on Build GRCh38
rs35355410 Chr.12: 53425459-53425459 on Build GRCh38
rs761279948 Chr.12: 53425480-53425480 on Build GRCh38
rs267603539 Chr.12: 53425772-53425772 on Build GRCh38
rs764475549 Chr.12: 53425671-53425671 on Build GRCh38
rs200276375 Chr.12: 53425431-53425431 on Build GRCh38
rs111460751 Chr.12: 53425427-53425427 on Build GRCh38
rs767707614 Chr.12: 53425679-53425679 on Build GRCh38
rs766527208 Chr.12: 53425804-53425804 on Build GRCh38
rs1019005711 Chr.12: 53425405-53425405 on Build GRCh38
rs764947789 Chr.12: 53425817-53425817 on Build GRCh38
rs528390504 Chr.12: 53425462-53425462 on Build GRCh38
rs781306620 Chr.12: 53425456-53425456 on Build GRCh38
rs763262910 Chr.12: 53425668-53425668 on Build GRCh38
rs1009450197 Chr.12: 53425415-53425415 on Build GRCh38
rs920098365 Chr.12: 53425656-53425656 on Build GRCh38
rs776292420 Chr.12: 53425765-53425765 on Build GRCh38
rs35774817 Chr.12: 53425616-53425617 on Build GRCh38
rs760273526 Chr.12: 53425496-53425496 on Build GRCh38
rs966443999 Chr.12: 53425407-53425407 on Build GRCh38
rs772653981 Chr.12: 53425610-53425610 on Build GRCh38
rs752995431 Chr.12: 53425521-53425521 on Build GRCh38
rs964077853 Chr.12: 53425584-53425584 on Build GRCh38
rs746584163 Chr.12: 53425760-53425760 on Build GRCh38
rs949006402 Chr.12: 53425784-53425784 on Build GRCh38
rs758445401 Chr.12: 53425391-53425391 on Build GRCh38
rs372745663 Chr.12: 53425547-53425547 on Build GRCh38
rs56226427 Chr.12: 53425769-53425769 on Build GRCh38
rs1041369932 Chr.12: 53425662-53425662 on Build GRCh38
rs146554515 Chr.12: 53425809-53425809 on Build GRCh38
rs757310194 Chr.12: 53425550-53425550 on Build GRCh38
rs535466975 Chr.12: 53425520-53425520 on Build GRCh38
rs778757150 Chr.12: 53425591-53425591 on Build GRCh38
rs145761355 Chr.12: 53425455-53425455 on Build GRCh38
rs115935733 Chr.12: 53425534-53425534 on Build GRCh38
rs116317145 Chr.12: 53425768-53425768 on Build GRCh38
rs150321213 Chr.12: 53425659-53425659 on Build GRCh38
rs746905091 Chr.12: 53425842-53425842 on Build GRCh38
rs764571819 Chr.12: 53425524-53425524 on Build GRCh38
rs187803948 Chr.12: 53425466-53425466 on Build GRCh38
rs778721310 Chr.12: 53425708-53425708 on Build GRCh38
rs754761311 Chr.12: 53425698-53425698 on Build GRCh38
rs753228275 Chr.12: 53425429-53425429 on Build GRCh38
rs762077507 Chr.12: 53425774-53425774 on Build GRCh38
rs114182001 Chr.12: 53425484-53425484 on Build GRCh38
rs754059893 Chr.12: 53425695-53425695 on Build GRCh38
rs868480524 Chr.12: 53425696-53425696 on Build GRCh38
rs35820869 Chr.12: 53425581-53425582 on Build GRCh38
rs193298272 Chr.12: 53425477-53425477 on Build GRCh38
rs115267458 Chr.12: 53425829-53425829 on Build GRCh38
rs770641486 Chr.12: 53425649-53425649 on Build GRCh38
rs758179091 Chr.12: 53425832-53425832 on Build GRCh38
rs376064725 Chr.12: 53425680-53425680 on Build GRCh38
rs374421820 Chr.12: 53425414-53425414 on Build GRCh38
rs768542840 Chr.12: 53425474-53425474 on Build GRCh38
rs760877575 Chr.12: 53425803-53425803 on Build GRCh38
rs749153412 Chr.12: 53425571-53425571 on Build GRCh38
rs200459389 Chr.12: 53425569-53425569 on Build GRCh38
rs142124054 Chr.12: 53425725-53425725 on Build GRCh38
rs926386950 Chr.12: 53425815-53425815 on Build GRCh38
rs138904291 Chr.12: 53425724-53425724 on Build GRCh38
rs774740207 Chr.12: 53425675-53425675 on Build GRCh38
rs781368665 Chr.12: 53425561-53425561 on Build GRCh38
rs763375589 Chr.12: 53425523-53425523 on Build GRCh38
rs766756135 Chr.12: 53425689-53425689 on Build GRCh38
rs146343768 Chr.12: 53425770-53425770 on Build GRCh38
rs770410518 Chr.12: 53425762-53425762 on Build GRCh38
rs1055470723 Chr.12: 53425604-53425604 on Build GRCh38
rs768196317 Chr.12: 53425780-53425780 on Build GRCh38
rs762321378 Chr.12: 53425677-53425677 on Build GRCh38
rs774776142 Chr.12: 53425836-53425836 on Build GRCh38
rs141101903 Chr.12: 53425482-53425482 on Build GRCh38
rs917000947 Chr.12: 53425628-53425628 on Build GRCh38
rs773833238 Chr.12: 53425802-53425802 on Build GRCh38
rs750685505 Chr.12: 53425562-53425562 on Build GRCh38
rs375425684 Chr.12: 53425464-53425464 on Build GRCh38
rs55982347 Chr.12: 53425808-53425808 on Build GRCh38

View Full Product Details