Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6785257-6785769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMTA1
Entrez Gene ID
23261
Gene Name
calmodulin binding transcription activator 1
Gene Aliases
CANPMR
UniGene
Hs.397705
Gene Chromosome Location
Chr.1: 6785324-7769706 on Build GRCh38
Transcripts
NM_001195563.1 NM_001242701.1 NM_015215.3 NR_038934.1 XM_011541083.2 XM_011541084.2 XM_011541085.2 XM_011541086.2 XM_011541087.2 XM_011541088.2 XM_011541089.2 XM_011541090.2 XM_011541091.2 XM_011541092.2 XM_017000774.1 XM_017000775.1 XM_017000776.1 XM_017000777.1 XM_017000778.1 XM_017000779.1 XM_017000780.1 XM_017000781.1

SNP Detail

SNP ID Chromosome Location
rs906690672 Chr.1: 6785729-6785729 on Build GRCh38
rs925692317 Chr.1: 6785683-6785699 on Build GRCh38
rs1049631249 Chr.1: 6785693-6785693 on Build GRCh38
rs748334484 Chr.1: 6785621-6785621 on Build GRCh38
rs977209338 Chr.1: 6785352-6785352 on Build GRCh38
rs1021927803 Chr.1: 6785592-6785592 on Build GRCh38
rs1033666479 Chr.1: 6785516-6785516 on Build GRCh38
rs1007524420 Chr.1: 6785710-6785710 on Build GRCh38
rs970705367 Chr.1: 6785293-6785293 on Build GRCh38
rs982054734 Chr.1: 6785298-6785298 on Build GRCh38
rs1049111385 Chr.1: 6785466-6785466 on Build GRCh38
rs1039356730 Chr.1: 6785726-6785741 on Build GRCh38
rs977636214 Chr.1: 6785614-6785614 on Build GRCh38
rs914450977 Chr.1: 6785336-6785336 on Build GRCh38
rs140309561 Chr.1: 6785349-6785350 on Build GRCh38
rs192783463 Chr.1: 6785504-6785504 on Build GRCh38
rs1012870310 Chr.1: 6785590-6785590 on Build GRCh38
rs945856050 Chr.1: 6785673-6785687 on Build GRCh38
rs774568430 Chr.1: 6785598-6785598 on Build GRCh38
rs943705745 Chr.1: 6785448-6785449 on Build GRCh38
rs781157444 Chr.1: 6785519-6785519 on Build GRCh38
rs994955937 Chr.1: 6785689-6785689 on Build GRCh38
rs902144260 Chr.1: 6785730-6785730 on Build GRCh38
rs955415196 Chr.1: 6785302-6785302 on Build GRCh38
rs543906692 Chr.1: 6785398-6785398 on Build GRCh38
rs777288848 Chr.1: 6785491-6785491 on Build GRCh38
rs755609750 Chr.1: 6785518-6785520 on Build GRCh38
rs920912464 Chr.1: 6785454-6785454 on Build GRCh38
rs1055128232 Chr.1: 6785731-6785731 on Build GRCh38
rs1004750068 Chr.1: 6785486-6785486 on Build GRCh38
rs779326250 Chr.1: 6785517-6785518 on Build GRCh38
rs757843745 Chr.1: 6785485-6785485 on Build GRCh38
rs375462112 Chr.1: 6785299-6785299 on Build GRCh38
rs911170562 Chr.1: 6785334-6785334 on Build GRCh38
rs985379191 Chr.1: 6785325-6785325 on Build GRCh38
rs1003622983 Chr.1: 6785730-6785731 on Build GRCh38
rs992651549 Chr.1: 6785662-6785688 on Build GRCh38
rs932955592 Chr.1: 6785460-6785460 on Build GRCh38
rs889179032 Chr.1: 6785697-6785697 on Build GRCh38
rs755656812 Chr.1: 6785646-6785646 on Build GRCh38
rs941360016 Chr.1: 6785350-6785350 on Build GRCh38
rs532386291 Chr.1: 6785644-6785644 on Build GRCh38
rs745666229 Chr.1: 6785547-6785547 on Build GRCh38
rs920964374 Chr.1: 6785658-6785658 on Build GRCh38
rs959364667 Chr.1: 6785588-6785588 on Build GRCh38
rs1011288352 Chr.1: 6785745-6785745 on Build GRCh38
rs1034249138 Chr.1: 6785452-6785453 on Build GRCh38
rs924320349 Chr.1: 6785430-6785430 on Build GRCh38
rs968586749 Chr.1: 6785597-6785597 on Build GRCh38
rs1018862409 Chr.1: 6785715-6785715 on Build GRCh38
rs186614298 Chr.1: 6785312-6785312 on Build GRCh38
rs976494405 Chr.1: 6785637-6785637 on Build GRCh38
rs1037136642 Chr.1: 6785487-6785487 on Build GRCh38
rs1048641492 Chr.1: 6785665-6785665 on Build GRCh38
rs769546370 Chr.1: 6785564-6785564 on Build GRCh38
rs34262581 Chr.1: 6785422-6785422 on Build GRCh38
rs561022348 Chr.1: 6785291-6785291 on Build GRCh38
rs965154299 Chr.1: 6785632-6785632 on Build GRCh38
rs991797363 Chr.1: 6785333-6785333 on Build GRCh38
rs1003569073 Chr.1: 6785509-6785509 on Build GRCh38
rs746911118 Chr.1: 6785495-6785495 on Build GRCh38
rs924372573 Chr.1: 6785616-6785616 on Build GRCh38
rs547797731 Chr.1: 6785703-6785703 on Build GRCh38

View Full Product Details