Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100205317-100205853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR6
Entrez Gene ID
64431
Gene Name
ARP6 actin-related protein 6 homolog
Gene Aliases
ARP6, CDA12, HSPC281, MSTP136, hARP6, hARPX
UniGene
Hs.115088
Gene Chromosome Location
Chr.12: 100200087-100224425 on Build GRCh38
Transcripts
NM_022496.4 NR_048568.1 NR_048569.1 XM_017019821.1 NR_135073.1

Gene Symbol
DEPDC4
Entrez Gene ID
120863
Gene Name
DEP domain containing 4
Gene Aliases
DEP.4
UniGene
Hs.653118
Gene Chromosome Location
Chr.12: 100203669-100268094 on Build GRCh38
Transcripts
NM_022496.4 NR_048568.1 NR_048569.1 XM_017019821.1 NR_135073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5991928 c.187-8delT p.? Chr.12: 100205668-100205668 on Build GRCh38
5361753 c.217C>T p.Q73* Chr.12: 100205706-100205706 on Build GRCh38
6248977 c.203G>T p.W68L Chr.12: 100205692-100205692 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1055289985 Chr.12: 100205522-100205522 on Build GRCh38
rs985315264 Chr.12: 100205484-100205484 on Build GRCh38
rs550605100 Chr.12: 100205789-100205789 on Build GRCh38
rs761842900 Chr.12: 100205646-100205646 on Build GRCh38
rs754428694 Chr.12: 100205742-100205743 on Build GRCh38
rs758238959 Chr.12: 100205661-100205661 on Build GRCh38
rs747598274 Chr.12: 100205685-100205685 on Build GRCh38
rs904598929 Chr.12: 100205441-100205441 on Build GRCh38
rs191567788 Chr.12: 100205674-100205674 on Build GRCh38
rs910930111 Chr.12: 100205488-100205488 on Build GRCh38
rs925546462 Chr.12: 100205518-100205518 on Build GRCh38
rs772265819 Chr.12: 100205780-100205780 on Build GRCh38
rs937237842 Chr.12: 100205381-100205381 on Build GRCh38
rs200351125 Chr.12: 100205687-100205687 on Build GRCh38
rs182355337 Chr.12: 100205500-100205500 on Build GRCh38
rs1024970176 Chr.12: 100205348-100205348 on Build GRCh38
rs17030036 Chr.12: 100205720-100205720 on Build GRCh38
rs967013105 Chr.12: 100205708-100205708 on Build GRCh38
rs1042635576 Chr.12: 100205508-100205508 on Build GRCh38
rs972206284 Chr.12: 100205513-100205514 on Build GRCh38
rs936786523 Chr.12: 100205520-100205520 on Build GRCh38
rs1000193578 Chr.12: 100205465-100205465 on Build GRCh38
rs965148977 Chr.12: 100205489-100205489 on Build GRCh38
rs764738691 Chr.12: 100205665-100205666 on Build GRCh38
rs1018104504 Chr.12: 100205648-100205648 on Build GRCh38
rs542383628 Chr.12: 100205654-100205654 on Build GRCh38
rs765221024 Chr.12: 100205649-100205649 on Build GRCh38
rs760801748 Chr.12: 100205796-100205796 on Build GRCh38
rs952430307 Chr.12: 100205545-100205546 on Build GRCh38
rs779806999 Chr.12: 100205667-100205667 on Build GRCh38
rs778211114 Chr.12: 100205689-100205689 on Build GRCh38
rs201301665 Chr.12: 100205781-100205781 on Build GRCh38
rs976947834 Chr.12: 100205497-100205497 on Build GRCh38
rs983793894 Chr.12: 100205570-100205570 on Build GRCh38
rs780573802 Chr.12: 100205681-100205681 on Build GRCh38
rs1057369011 Chr.12: 100205402-100205402 on Build GRCh38
rs150399997 Chr.12: 100205412-100205412 on Build GRCh38
rs1041814312 Chr.12: 100205439-100205439 on Build GRCh38
rs767479530 Chr.12: 100205639-100205639 on Build GRCh38
rs897133376 Chr.12: 100205589-100205589 on Build GRCh38
rs994146173 Chr.12: 100205610-100205610 on Build GRCh38
rs774689043 Chr.12: 100205751-100205751 on Build GRCh38
rs574052357 Chr.12: 100205426-100205426 on Build GRCh38
rs1013650990 Chr.12: 100205544-100205544 on Build GRCh38
rs563199129 Chr.12: 100205625-100205625 on Build GRCh38
rs1028943373 Chr.12: 100205467-100205467 on Build GRCh38
rs193262573 Chr.12: 100205499-100205499 on Build GRCh38
rs544893822 Chr.12: 100205550-100205550 on Build GRCh38
rs184084745 Chr.12: 100205767-100205767 on Build GRCh38
rs895341054 Chr.12: 100205535-100205535 on Build GRCh38
rs776993103 Chr.12: 100205371-100205371 on Build GRCh38
rs756242290 Chr.12: 100205635-100205635 on Build GRCh38
rs1019543768 Chr.12: 100205500-100205500 on Build GRCh38
rs144717521 Chr.12: 100205698-100205698 on Build GRCh38
rs759708097 Chr.12: 100205634-100205634 on Build GRCh38
rs952524084 Chr.12: 100205471-100205471 on Build GRCh38
rs138937703 Chr.12: 100205640-100205640 on Build GRCh38
rs187948951 Chr.12: 100205545-100205545 on Build GRCh38

View Full Product Details