Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100211951-100212463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR6
Entrez Gene ID
64431
Gene Name
ARP6 actin-related protein 6 homolog
Gene Aliases
ARP6, CDA12, HSPC281, MSTP136, hARP6, hARPX
UniGene
Hs.115088
Gene Chromosome Location
Chr.12: 100200087-100224425 on Build GRCh38
Transcripts
NM_022496.4 NR_048568.1 NR_048569.1 XM_017019821.1 NR_135073.1

Gene Symbol
DEPDC4
Entrez Gene ID
120863
Gene Name
DEP domain containing 4
Gene Aliases
DEP.4
UniGene
Hs.653118
Gene Chromosome Location
Chr.12: 100203669-100268094 on Build GRCh38
Transcripts
NM_022496.4 NR_048568.1 NR_048569.1 XM_017019821.1 NR_135073.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4004606 c.671+10A>G p.? Chr.12: 100212364-100212364 on Build GRCh38
6360178 c.657C>A p.D219E Chr.12: 100212340-100212340 on Build GRCh38
5613478 c.656A>T p.D219V Chr.12: 100212339-100212339 on Build GRCh38
6257078 c.583G>T p.V195F Chr.12: 100212266-100212266 on Build GRCh38
6382498 c.596C>A p.T199K Chr.12: 100212279-100212279 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895051359 Chr.12: 100212420-100212420 on Build GRCh38
rs190308955 Chr.12: 100212092-100212092 on Build GRCh38
rs896959296 Chr.12: 100212067-100212067 on Build GRCh38
rs985038393 Chr.12: 100212101-100212101 on Build GRCh38
rs566384999 Chr.12: 100212288-100212288 on Build GRCh38
rs1048510018 Chr.12: 100212095-100212095 on Build GRCh38
rs760940737 Chr.12: 100212282-100212283 on Build GRCh38
rs770549446 Chr.12: 100212240-100212240 on Build GRCh38
rs775942188 Chr.12: 100212251-100212251 on Build GRCh38
rs754049145 Chr.12: 100212297-100212297 on Build GRCh38
rs7313835 Chr.12: 100212242-100212242 on Build GRCh38
rs536142752 Chr.12: 100212239-100212239 on Build GRCh38
rs780952137 Chr.12: 100212370-100212370 on Build GRCh38
rs912493087 Chr.12: 100211996-100211996 on Build GRCh38
rs777538480 Chr.12: 100212410-100212410 on Build GRCh38
rs905932785 Chr.12: 100212096-100212096 on Build GRCh38
rs558407012 Chr.12: 100212340-100212340 on Build GRCh38
rs757397599 Chr.12: 100212336-100212336 on Build GRCh38
rs753910672 Chr.12: 100212406-100212409 on Build GRCh38
rs959174378 Chr.12: 100211976-100211976 on Build GRCh38
rs764290841 Chr.12: 100212366-100212367 on Build GRCh38
rs369544302 Chr.12: 100212244-100212244 on Build GRCh38
rs775065690 Chr.12: 100212241-100212241 on Build GRCh38
rs961111878 Chr.12: 100212079-100212079 on Build GRCh38
rs369148044 Chr.12: 100212432-100212432 on Build GRCh38
rs985117655 Chr.12: 100211977-100211977 on Build GRCh38
rs567178706 Chr.12: 100212196-100212196 on Build GRCh38
rs754965993 Chr.12: 100212360-100212360 on Build GRCh38
rs896089380 Chr.12: 100212130-100212130 on Build GRCh38
rs371881017 Chr.12: 100212357-100212357 on Build GRCh38
rs755841608 Chr.12: 100212408-100212408 on Build GRCh38
rs867446158 Chr.12: 100212191-100212191 on Build GRCh38
rs17030047 Chr.12: 100212099-100212099 on Build GRCh38
rs73147811 Chr.12: 100212116-100212116 on Build GRCh38
rs1040546428 Chr.12: 100212189-100212189 on Build GRCh38
rs78866321 Chr.12: 100212025-100212025 on Build GRCh38
rs953742921 Chr.12: 100212281-100212281 on Build GRCh38
rs140734277 Chr.12: 100212347-100212347 on Build GRCh38
rs772709909 Chr.12: 100212232-100212232 on Build GRCh38
rs991964495 Chr.12: 100212018-100212018 on Build GRCh38
rs185593406 Chr.12: 100212262-100212262 on Build GRCh38
rs1036886904 Chr.12: 100212055-100212055 on Build GRCh38
rs180800280 Chr.12: 100212396-100212396 on Build GRCh38
rs1001290926 Chr.12: 100212368-100212368 on Build GRCh38
rs912209670 Chr.12: 100212372-100212372 on Build GRCh38
rs189500291 Chr.12: 100212295-100212295 on Build GRCh38
rs943630719 Chr.12: 100212162-100212162 on Build GRCh38
rs972930549 Chr.12: 100212081-100212081 on Build GRCh38
rs530533736 Chr.12: 100212100-100212100 on Build GRCh38
rs368544342 Chr.12: 100212206-100212206 on Build GRCh38

View Full Product Details