Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 105176742-105177247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APPL2
Entrez Gene ID
55198
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Gene Aliases
DIP13B
UniGene
Hs.506603
Gene Chromosome Location
Chr.12: 105173297-105236230 on Build GRCh38
Transcripts
NM_001251904.1 NM_001251905.1 NM_018171.3 XM_006719472.1 XM_011538530.2 XM_011538531.2 XM_011538532.2 XM_017019551.1 XM_017019552.1 XM_017019553.1 XM_017019554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5422135 c.1672-6C>A p.? Chr.12: 105177022-105177022 on Build GRCh38
2064875 c.1765G>A p.E589K Chr.12: 105176923-105176923 on Build GRCh38
934304 c.1741C>T p.R581C Chr.12: 105176947-105176947 on Build GRCh38
1183286 c.1682C>A p.T561N Chr.12: 105177006-105177006 on Build GRCh38
934303 c.1781A>G p.Y594C Chr.12: 105176907-105176907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892602312 Chr.12: 105177080-105177080 on Build GRCh38
rs369065711 Chr.12: 105177115-105177115 on Build GRCh38
rs751734458 Chr.12: 105177167-105177167 on Build GRCh38
rs761544638 Chr.12: 105176771-105176771 on Build GRCh38
rs755325811 Chr.12: 105177028-105177028 on Build GRCh38
rs538776087 Chr.12: 105176792-105176792 on Build GRCh38
rs776979017 Chr.12: 105176916-105176917 on Build GRCh38
rs555725549 Chr.12: 105176785-105176785 on Build GRCh38
rs755223246 Chr.12: 105177169-105177169 on Build GRCh38
rs949743544 Chr.12: 105176889-105176889 on Build GRCh38
rs767297070 Chr.12: 105176929-105176929 on Build GRCh38
rs1009209895 Chr.12: 105177108-105177116 on Build GRCh38
rs781552911 Chr.12: 105176996-105176996 on Build GRCh38
rs370254990 Chr.12: 105176963-105176963 on Build GRCh38
rs756654740 Chr.12: 105177195-105177195 on Build GRCh38
rs767224181 Chr.12: 105177049-105177049 on Build GRCh38
rs73179990 Chr.12: 105177203-105177203 on Build GRCh38
rs759283532 Chr.12: 105176913-105176913 on Build GRCh38
rs745577754 Chr.12: 105177217-105177217 on Build GRCh38
rs906780960 Chr.12: 105177058-105177058 on Build GRCh38
rs748523680 Chr.12: 105177179-105177179 on Build GRCh38
rs750468282 Chr.12: 105176979-105176981 on Build GRCh38
rs1007268101 Chr.12: 105177143-105177143 on Build GRCh38
rs1047495707 Chr.12: 105176870-105176870 on Build GRCh38
rs780610422 Chr.12: 105176843-105176843 on Build GRCh38
rs764085045 Chr.12: 105177212-105177214 on Build GRCh38
rs756070695 Chr.12: 105176905-105176905 on Build GRCh38
rs940772285 Chr.12: 105176951-105176951 on Build GRCh38
rs769298089 Chr.12: 105176887-105176887 on Build GRCh38
rs771729906 Chr.12: 105177225-105177225 on Build GRCh38
rs777023806 Chr.12: 105176881-105176881 on Build GRCh38
rs955515776 Chr.12: 105177120-105177120 on Build GRCh38
rs1053037026 Chr.12: 105177080-105177081 on Build GRCh38
rs139517407 Chr.12: 105177083-105177083 on Build GRCh38
rs769011584 Chr.12: 105176904-105176907 on Build GRCh38
rs780899503 Chr.12: 105176819-105176819 on Build GRCh38
rs775875847 Chr.12: 105176970-105176971 on Build GRCh38
rs377356032 Chr.12: 105176825-105176825 on Build GRCh38
rs763402698 Chr.12: 105177006-105177007 on Build GRCh38
rs751938287 Chr.12: 105177025-105177026 on Build GRCh38
rs765995069 Chr.12: 105176899-105176899 on Build GRCh38
rs373779141 Chr.12: 105177061-105177061 on Build GRCh38
rs766506811 Chr.12: 105177164-105177164 on Build GRCh38
rs891948992 Chr.12: 105177085-105177085 on Build GRCh38
rs770390221 Chr.12: 105177023-105177023 on Build GRCh38
rs765293784 Chr.12: 105176923-105176923 on Build GRCh38
rs752593594 Chr.12: 105176944-105176944 on Build GRCh38
rs373485718 Chr.12: 105176846-105176846 on Build GRCh38
rs758730830 Chr.12: 105176973-105176973 on Build GRCh38
rs2289773 Chr.12: 105177112-105177112 on Build GRCh38
rs773848624 Chr.12: 105177216-105177216 on Build GRCh38
rs199559987 Chr.12: 105176891-105176891 on Build GRCh38
rs748767178 Chr.12: 105177021-105177021 on Build GRCh38
rs779187749 Chr.12: 105176947-105176947 on Build GRCh38
rs1018278264 Chr.12: 105177180-105177180 on Build GRCh38
rs753784396 Chr.12: 105177140-105177140 on Build GRCh38
rs747203652 Chr.12: 105176992-105176992 on Build GRCh38
rs756017295 Chr.12: 105176946-105176946 on Build GRCh38
rs748669786 Chr.12: 105177002-105177002 on Build GRCh38
rs781468127 Chr.12: 105177174-105177174 on Build GRCh38
rs761995028 Chr.12: 105176915-105176916 on Build GRCh38
rs868559097 Chr.12: 105177018-105177019 on Build GRCh38
rs780452705 Chr.12: 105176984-105176984 on Build GRCh38
rs184843985 Chr.12: 105176821-105176821 on Build GRCh38
rs772758389 Chr.12: 105177018-105177018 on Build GRCh38
rs773899510 Chr.12: 105177042-105177042 on Build GRCh38
rs770443632 Chr.12: 105177213-105177213 on Build GRCh38
rs770271918 Chr.12: 105177014-105177014 on Build GRCh38
rs189720343 Chr.12: 105177212-105177212 on Build GRCh38
rs574673409 Chr.12: 105177154-105177154 on Build GRCh38
rs111815589 Chr.12: 105177048-105177048 on Build GRCh38
rs867054082 Chr.12: 105177005-105177006 on Build GRCh38
rs766809485 Chr.12: 105177018-105177019 on Build GRCh38
rs768914856 Chr.12: 105176994-105176994 on Build GRCh38
rs963681143 Chr.12: 105176833-105176833 on Build GRCh38
rs920604115 Chr.12: 105176983-105176983 on Build GRCh38
rs767273812 Chr.12: 105176801-105176801 on Build GRCh38
rs769022744 Chr.12: 105176871-105176871 on Build GRCh38
rs762652229 Chr.12: 105176896-105176896 on Build GRCh38
rs760740351 Chr.12: 105177054-105177054 on Build GRCh38
rs759172553 Chr.12: 105177046-105177046 on Build GRCh38

View Full Product Details