Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111709970-111710481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD10
Entrez Gene ID
80724
Gene Name
acyl-CoA dehydrogenase family member 10
Gene Aliases
-
UniGene
Hs.331141
Gene Chromosome Location
Chr.12: 111686053-111757107 on Build GRCh38
Transcripts
NM_001136538.1 NM_025247.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6397229 c.716A>G p.K239R Chr.12: 111710298-111710298 on Build GRCh38
3396055 c.759C>G p.Y253* Chr.12: 111710341-111710341 on Build GRCh38
3396056 c.772G>C p.V258L Chr.12: 111710354-111710354 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs539739506 Chr.12: 111710416-111710416 on Build GRCh38
rs919097364 Chr.12: 111710400-111710400 on Build GRCh38
rs762530458 Chr.12: 111710091-111710091 on Build GRCh38
rs894104651 Chr.12: 111710184-111710184 on Build GRCh38
rs547422439 Chr.12: 111710325-111710325 on Build GRCh38
rs984541389 Chr.12: 111710005-111710005 on Build GRCh38
rs375941386 Chr.12: 111710375-111710375 on Build GRCh38
rs930400207 Chr.12: 111710152-111710152 on Build GRCh38
rs543591426 Chr.12: 111710391-111710391 on Build GRCh38
rs186488391 Chr.12: 111710307-111710307 on Build GRCh38
rs984525213 Chr.12: 111710419-111710419 on Build GRCh38
rs150771041 Chr.12: 111710172-111710172 on Build GRCh38
rs560526859 Chr.12: 111710079-111710079 on Build GRCh38
rs527885685 Chr.12: 111710154-111710154 on Build GRCh38
rs892529254 Chr.12: 111710090-111710090 on Build GRCh38
rs117374315 Chr.12: 111710308-111710308 on Build GRCh38
rs74909944 Chr.12: 111710089-111710089 on Build GRCh38
rs763670312 Chr.12: 111710189-111710189 on Build GRCh38
rs922819299 Chr.12: 111710113-111710113 on Build GRCh38
rs919111138 Chr.12: 111710048-111710048 on Build GRCh38
rs934236032 Chr.12: 111710124-111710124 on Build GRCh38
rs542994043 Chr.12: 111710160-111710160 on Build GRCh38
rs944497656 Chr.12: 111709992-111709992 on Build GRCh38
rs946795805 Chr.12: 111710181-111710181 on Build GRCh38
rs962104406 Chr.12: 111710449-111710449 on Build GRCh38
rs1008248212 Chr.12: 111710333-111710333 on Build GRCh38
rs1006313553 Chr.12: 111710431-111710431 on Build GRCh38
rs117545816 Chr.12: 111710450-111710450 on Build GRCh38
rs999087186 Chr.12: 111710253-111710253 on Build GRCh38
rs751727677 Chr.12: 111710227-111710227 on Build GRCh38
rs750987649 Chr.12: 111710339-111710340 on Build GRCh38
rs915659942 Chr.12: 111710441-111710441 on Build GRCh38
rs112110213 Chr.12: 111710033-111710033 on Build GRCh38
rs554600613 Chr.12: 111710418-111710418 on Build GRCh38
rs907484313 Chr.12: 111710247-111710247 on Build GRCh38
rs972006567 Chr.12: 111710397-111710397 on Build GRCh38
rs181174401 Chr.12: 111710296-111710296 on Build GRCh38
rs752084259 Chr.12: 111710017-111710017 on Build GRCh38
rs754976139 Chr.12: 111710398-111710399 on Build GRCh38
rs1045934754 Chr.12: 111710244-111710244 on Build GRCh38
rs907071637 Chr.12: 111710326-111710326 on Build GRCh38
rs753217344 Chr.12: 111710292-111710292 on Build GRCh38
rs972884359 Chr.12: 111710105-111710105 on Build GRCh38
rs1010032205 Chr.12: 111710321-111710321 on Build GRCh38
rs774584444 Chr.12: 111710399-111710399 on Build GRCh38
rs531360509 Chr.12: 111710246-111710246 on Build GRCh38
rs773902648 Chr.12: 111710442-111710443 on Build GRCh38
rs1031787909 Chr.12: 111710288-111710288 on Build GRCh38
rs1034293951 Chr.12: 111710407-111710415 on Build GRCh38

View Full Product Details