Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12191682-12192193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR4632
Entrez Gene ID
100616438
Gene Name
microRNA 4632
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.1: 12191713-12191773 on Build GRCh38
Transcripts
NR_039775.2 NM_001066.2 XM_011542060.2 XM_011542063.2 XM_017002211.1 XM_017002212.1 XM_017002213.1 XM_017002214.1 XM_017002215.1

Gene Symbol
TNFRSF1B
Entrez Gene ID
7133
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
UniGene
Hs.256278
Gene Chromosome Location
Chr.1: 12166948-12209222 on Build GRCh38
Transcripts
NR_039775.2 NM_001066.2 XM_011542060.2 XM_011542063.2 XM_017002211.1 XM_017002212.1 XM_017002213.1 XM_017002214.1 XM_017002215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4021355 c.416C>T p.P139L Chr.1: 12191882-12191882 on Build GRCh38
6430976 c.329G>A p.C110Y Chr.1: 12191795-12191795 on Build GRCh38
6805056 c.379G>A p.A127T Chr.1: 12191845-12191845 on Build GRCh38
6057763 c.425A>T p.K142M Chr.1: 12191891-12191891 on Build GRCh38
3788530 c.436G>T p.G146C Chr.1: 12191902-12191902 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372809836 Chr.1: 12191755-12191755 on Build GRCh38
rs538653610 Chr.1: 12191859-12191859 on Build GRCh38
rs776137552 Chr.1: 12191725-12191726 on Build GRCh38
rs774708133 Chr.1: 12191958-12191958 on Build GRCh38
rs778072312 Chr.1: 12191787-12191787 on Build GRCh38
rs954263552 Chr.1: 12191996-12191996 on Build GRCh38
rs749531606 Chr.1: 12191942-12191942 on Build GRCh38
rs952383926 Chr.1: 12192125-12192125 on Build GRCh38
rs752962082 Chr.1: 12191728-12191728 on Build GRCh38
rs771305913 Chr.1: 12191930-12191930 on Build GRCh38
rs747434014 Chr.1: 12191901-12191901 on Build GRCh38
rs375867757 Chr.1: 12191927-12191927 on Build GRCh38
rs763190463 Chr.1: 12191841-12191841 on Build GRCh38
rs746367262 Chr.1: 12191923-12191923 on Build GRCh38
rs372784869 Chr.1: 12191731-12191731 on Build GRCh38
rs778125323 Chr.1: 12191897-12191897 on Build GRCh38
rs765439526 Chr.1: 12191723-12191723 on Build GRCh38
rs757616668 Chr.1: 12191904-12191904 on Build GRCh38
rs983748594 Chr.1: 12192146-12192146 on Build GRCh38
rs765864632 Chr.1: 12191882-12191882 on Build GRCh38
rs5746024 Chr.1: 12192004-12192004 on Build GRCh38
rs1021760143 Chr.1: 12191994-12191994 on Build GRCh38
rs761655488 Chr.1: 12191887-12191888 on Build GRCh38
rs761685259 Chr.1: 12191847-12191847 on Build GRCh38
rs985050720 Chr.1: 12191735-12191735 on Build GRCh38
rs985340039 Chr.1: 12191997-12191997 on Build GRCh38
rs767627829 Chr.1: 12191858-12191858 on Build GRCh38
rs759820006 Chr.1: 12191720-12191720 on Build GRCh38
rs145097690 Chr.1: 12191883-12191883 on Build GRCh38
rs375550027 Chr.1: 12191941-12191941 on Build GRCh38
rs773044555 Chr.1: 12191975-12191975 on Build GRCh38
rs746004548 Chr.1: 12191813-12191813 on Build GRCh38
rs761738546 Chr.1: 12191962-12191962 on Build GRCh38
rs150483691 Chr.1: 12191802-12191802 on Build GRCh38
rs763391486 Chr.1: 12191929-12191929 on Build GRCh38
rs747520649 Chr.1: 12191797-12191797 on Build GRCh38
rs754560287 Chr.1: 12191888-12191888 on Build GRCh38
rs498906 Chr.1: 12191708-12191708 on Build GRCh38
rs918468125 Chr.1: 12192105-12192105 on Build GRCh38
rs757190166 Chr.1: 12191743-12191743 on Build GRCh38
rs775628074 Chr.1: 12191836-12191836 on Build GRCh38
rs758783633 Chr.1: 12191758-12191758 on Build GRCh38
rs916364499 Chr.1: 12192160-12192160 on Build GRCh38
rs1044948428 Chr.1: 12191865-12191865 on Build GRCh38
rs1011546567 Chr.1: 12191989-12191989 on Build GRCh38
rs763467359 Chr.1: 12191934-12191940 on Build GRCh38
rs907479646 Chr.1: 12191900-12191900 on Build GRCh38
rs201949042 Chr.1: 12191892-12191892 on Build GRCh38
rs751575604 Chr.1: 12191742-12191742 on Build GRCh38
rs771371338 Chr.1: 12191801-12191801 on Build GRCh38
rs965219186 Chr.1: 12192074-12192074 on Build GRCh38
rs769890498 Chr.1: 12191833-12191833 on Build GRCh38
rs762700397 Chr.1: 12191730-12191730 on Build GRCh38
rs375442414 Chr.1: 12191846-12191846 on Build GRCh38
rs780859909 Chr.1: 12191749-12191749 on Build GRCh38
rs653667 Chr.1: 12191751-12191751 on Build GRCh38
rs891811791 Chr.1: 12191954-12191954 on Build GRCh38
rs764377878 Chr.1: 12191971-12191972 on Build GRCh38
rs138462821 Chr.1: 12191845-12191845 on Build GRCh38
rs775568440 Chr.1: 12191940-12191940 on Build GRCh38
rs901570828 Chr.1: 12192087-12192087 on Build GRCh38
rs750342650 Chr.1: 12191869-12191869 on Build GRCh38
rs781592930 Chr.1: 12191920-12191920 on Build GRCh38
rs774736074 Chr.1: 12191995-12191995 on Build GRCh38
rs13306721 Chr.1: 12192159-12192159 on Build GRCh38
rs5746023 Chr.1: 12191986-12191986 on Build GRCh38
rs972636109 Chr.1: 12192082-12192082 on Build GRCh38
rs1040467878 Chr.1: 12192083-12192083 on Build GRCh38
rs993666900 Chr.1: 12192115-12192115 on Build GRCh38
rs1026450185 Chr.1: 12192118-12192118 on Build GRCh38
rs1019848979 Chr.1: 12192044-12192044 on Build GRCh38
rs755627203 Chr.1: 12191878-12191878 on Build GRCh38
rs776627969 Chr.1: 12191713-12191713 on Build GRCh38
rs17880512 Chr.1: 12191953-12191953 on Build GRCh38
rs554390462 Chr.1: 12191966-12191966 on Build GRCh38

View Full Product Details