Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12207031-12207521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF1B
Entrez Gene ID
7133
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
UniGene
Hs.256278
Gene Chromosome Location
Chr.1: 12166948-12209222 on Build GRCh38
Transcripts
NM_001066.2 XM_011542060.2 XM_011542063.2 XM_017002214.1 XM_017002215.1

SNP Detail

SNP ID Chromosome Location
rs936570944 Chr.1: 12207354-12207354 on Build GRCh38
rs919987446 Chr.1: 12207075-12207075 on Build GRCh38
rs10157774 Chr.1: 12207445-12207445 on Build GRCh38
rs557951117 Chr.1: 12207166-12207166 on Build GRCh38
rs534561339 Chr.1: 12207157-12207157 on Build GRCh38
rs577935304 Chr.1: 12207170-12207170 on Build GRCh38
rs3397 Chr.1: 12207235-12207235 on Build GRCh38
rs1012551059 Chr.1: 12207387-12207387 on Build GRCh38
rs1053867261 Chr.1: 12207363-12207363 on Build GRCh38
rs900125067 Chr.1: 12207367-12207367 on Build GRCh38
rs767669838 Chr.1: 12207054-12207054 on Build GRCh38
rs368738070 Chr.1: 12207330-12207330 on Build GRCh38
rs537912812 Chr.1: 12207490-12207490 on Build GRCh38
rs1027403913 Chr.1: 12207493-12207493 on Build GRCh38
rs540109288 Chr.1: 12207196-12207196 on Build GRCh38
rs949216081 Chr.1: 12207315-12207315 on Build GRCh38
rs5746066 Chr.1: 12207473-12207473 on Build GRCh38
rs1042284268 Chr.1: 12207127-12207127 on Build GRCh38
rs374945013 Chr.1: 12207062-12207062 on Build GRCh38
rs5746064 Chr.1: 12207390-12207391 on Build GRCh38
rs368441565 Chr.1: 12207207-12207207 on Build GRCh38
rs1031981858 Chr.1: 12207154-12207154 on Build GRCh38
rs896289977 Chr.1: 12207107-12207107 on Build GRCh38
rs1042315853 Chr.1: 12207316-12207316 on Build GRCh38
rs200873417 Chr.1: 12207158-12207158 on Build GRCh38
rs5746065 Chr.1: 12207407-12207407 on Build GRCh38
rs186736505 Chr.1: 12207140-12207140 on Build GRCh38
rs902223162 Chr.1: 12207141-12207141 on Build GRCh38
rs751085905 Chr.1: 12207360-12207367 on Build GRCh38
rs1061624 Chr.1: 12207208-12207208 on Build GRCh38
rs374272314 Chr.1: 12207195-12207195 on Build GRCh38
rs1008380652 Chr.1: 12207433-12207433 on Build GRCh38
rs551473542 Chr.1: 12207242-12207242 on Build GRCh38
rs917790470 Chr.1: 12207306-12207306 on Build GRCh38
rs755156523 Chr.1: 12207346-12207346 on Build GRCh38
rs760912981 Chr.1: 12207067-12207067 on Build GRCh38
rs987083843 Chr.1: 12207234-12207234 on Build GRCh38
rs747764180 Chr.1: 12207084-12207085 on Build GRCh38
rs5746067 Chr.1: 12207492-12207492 on Build GRCh38
rs1015748162 Chr.1: 12207450-12207450 on Build GRCh38
rs1000919433 Chr.1: 12207142-12207142 on Build GRCh38
rs996367156 Chr.1: 12207370-12207370 on Build GRCh38
rs753644221 Chr.1: 12207108-12207108 on Build GRCh38
rs977155835 Chr.1: 12207461-12207461 on Build GRCh38
rs114236595 Chr.1: 12207268-12207268 on Build GRCh38
rs990826717 Chr.1: 12207302-12207302 on Build GRCh38
rs527887561 Chr.1: 12207276-12207276 on Build GRCh38
rs577235106 Chr.1: 12207434-12207434 on Build GRCh38
rs555160047 Chr.1: 12207069-12207069 on Build GRCh38
rs5030792 Chr.1: 12207213-12207213 on Build GRCh38
rs1051501920 Chr.1: 12207401-12207401 on Build GRCh38
rs568886932 Chr.1: 12207480-12207480 on Build GRCh38
rs773310858 Chr.1: 12207060-12207060 on Build GRCh38
rs902455632 Chr.1: 12207339-12207339 on Build GRCh38
rs5746068 Chr.1: 12207498-12207498 on Build GRCh38
rs916968123 Chr.1: 12207138-12207138 on Build GRCh38
rs957501544 Chr.1: 12207476-12207476 on Build GRCh38
rs547731360 Chr.1: 12207359-12207359 on Build GRCh38
rs764108902 Chr.1: 12207081-12207081 on Build GRCh38
rs746246949 Chr.1: 12207364-12207367 on Build GRCh38
rs201385323 Chr.1: 12207059-12207059 on Build GRCh38
rs370508215 Chr.1: 12207082-12207082 on Build GRCh38
rs575457978 Chr.1: 12207087-12207087 on Build GRCh38
rs938086908 Chr.1: 12207086-12207086 on Build GRCh38
rs531722591 Chr.1: 12207236-12207236 on Build GRCh38
rs974500442 Chr.1: 12207244-12207244 on Build GRCh38
rs868677549 Chr.1: 12207379-12207379 on Build GRCh38
rs1055380816 Chr.1: 12207106-12207106 on Build GRCh38

View Full Product Details