Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27925661-27926162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDX1
Entrez Gene ID
3651
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1
UniGene
Hs.32938
Gene Chromosome Location
Chr.13: 27919982-27926314 on Build GRCh38
Transcripts
NM_000209.3

SNP Detail

SNP ID Chromosome Location
rs74905790 Chr.13: 27925734-27925734 on Build GRCh38
rs988071587 Chr.13: 27925890-27925890 on Build GRCh38
rs965503360 Chr.13: 27925820-27925820 on Build GRCh38
rs954250190 Chr.13: 27926016-27926016 on Build GRCh38
rs555696245 Chr.13: 27926099-27926099 on Build GRCh38
rs1004438085 Chr.13: 27925757-27925757 on Build GRCh38
rs112637982 Chr.13: 27925716-27925716 on Build GRCh38
rs1046702086 Chr.13: 27925916-27925916 on Build GRCh38
rs9319402 Chr.13: 27926043-27926043 on Build GRCh38
rs1046179539 Chr.13: 27925941-27925941 on Build GRCh38
rs901471926 Chr.13: 27926006-27926006 on Build GRCh38
rs75697516 Chr.13: 27925741-27925741 on Build GRCh38
rs541144902 Chr.13: 27925706-27925706 on Build GRCh38
rs957680474 Chr.13: 27926125-27926125 on Build GRCh38
rs963114404 Chr.13: 27925728-27925728 on Build GRCh38
rs925421874 Chr.13: 27926058-27926058 on Build GRCh38
rs7981781 Chr.13: 27925825-27925825 on Build GRCh38
rs986943849 Chr.13: 27925764-27925764 on Build GRCh38
rs985780045 Chr.13: 27925833-27925833 on Build GRCh38
rs1015684824 Chr.13: 27925786-27925786 on Build GRCh38
rs113776422 Chr.13: 27926056-27926056 on Build GRCh38
rs558894682 Chr.13: 27925769-27925769 on Build GRCh38
rs888458039 Chr.13: 27925951-27925951 on Build GRCh38
rs987059380 Chr.13: 27926042-27926042 on Build GRCh38
rs963359999 Chr.13: 27925956-27925956 on Build GRCh38
rs1004748205 Chr.13: 27926138-27926138 on Build GRCh38
rs936549507 Chr.13: 27925850-27925850 on Build GRCh38
rs916587775 Chr.13: 27926109-27926109 on Build GRCh38
rs1055017332 Chr.13: 27926092-27926092 on Build GRCh38
rs886275720 Chr.13: 27925944-27925944 on Build GRCh38
rs976911916 Chr.13: 27925822-27925822 on Build GRCh38
rs998300221 Chr.13: 27926021-27926021 on Build GRCh38
rs112431822 Chr.13: 27925921-27925921 on Build GRCh38
rs905301326 Chr.13: 27925725-27925725 on Build GRCh38
rs894976456 Chr.13: 27925691-27925691 on Build GRCh38
rs888673510 Chr.13: 27925747-27925747 on Build GRCh38
rs545512348 Chr.13: 27926003-27926003 on Build GRCh38
rs953117306 Chr.13: 27926054-27926054 on Build GRCh38
rs776354464 Chr.13: 27926017-27926017 on Build GRCh38
rs113931775 Chr.13: 27925939-27925939 on Build GRCh38
rs574699287 Chr.13: 27925832-27925832 on Build GRCh38
rs913850313 Chr.13: 27925896-27925896 on Build GRCh38
rs892322969 Chr.13: 27925701-27925701 on Build GRCh38
rs764829593 Chr.13: 27926131-27926131 on Build GRCh38
rs949184150 Chr.13: 27925907-27925907 on Build GRCh38
rs954176479 Chr.13: 27925753-27925753 on Build GRCh38
rs112525679 Chr.13: 27925723-27925723 on Build GRCh38
rs1054945680 Chr.13: 27925895-27925895 on Build GRCh38
rs953283792 Chr.13: 27925831-27925831 on Build GRCh38
rs530321473 Chr.13: 27925946-27925946 on Build GRCh38
rs548276958 Chr.13: 27926124-27926124 on Build GRCh38
rs945420930 Chr.13: 27925788-27925788 on Build GRCh38
rs1037225548 Chr.13: 27926001-27926001 on Build GRCh38
rs1004197141 Chr.13: 27925991-27925991 on Build GRCh38
rs183052606 Chr.13: 27926136-27926136 on Build GRCh38
rs1031978470 Chr.13: 27926067-27926067 on Build GRCh38
rs1013394384 Chr.13: 27925700-27925700 on Build GRCh38
rs936767535 Chr.13: 27926059-27926059 on Build GRCh38
rs776277133 Chr.13: 27925836-27925836 on Build GRCh38
rs974722717 Chr.13: 27925733-27925733 on Build GRCh38
rs554902597 Chr.13: 27925826-27925826 on Build GRCh38
rs572475449 Chr.13: 27925791-27925791 on Build GRCh38
rs924944124 Chr.13: 27925837-27925837 on Build GRCh38

View Full Product Details