Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95072862-95073367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC4
Entrez Gene ID
10257
Gene Name
ATP binding cassette subfamily C member 4
Gene Aliases
MOAT-B, MOATB, MRP4
UniGene
Hs.508423
Gene Chromosome Location
Chr.13: 95019829-95301550 on Build GRCh38
Transcripts
NM_001301829.1 NM_005845.4 XM_005254025.2 XM_017020319.1 XM_017020321.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
948950 c.2992C>T p.R998* Chr.13: 95073230-95073230 on Build GRCh38
6315693 c.2932C>A p.Q978K Chr.13: 95073290-95073290 on Build GRCh38
1181345 c.2963C>T p.T988M Chr.13: 95073259-95073259 on Build GRCh38
5437219 c.3018+2T>C p.? Chr.13: 95073202-95073202 on Build GRCh38
5920577 c.2997A>T p.Q999H Chr.13: 95073225-95073225 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973894438 Chr.13: 95072941-95072941 on Build GRCh38
rs1044398044 Chr.13: 95073131-95073131 on Build GRCh38
rs764107411 Chr.13: 95073310-95073310 on Build GRCh38
rs370539767 Chr.13: 95073341-95073341 on Build GRCh38
rs377216030 Chr.13: 95073242-95073242 on Build GRCh38
rs1018157692 Chr.13: 95072919-95072919 on Build GRCh38
rs911951872 Chr.13: 95073058-95073058 on Build GRCh38
rs915583752 Chr.13: 95072888-95072888 on Build GRCh38
rs764028696 Chr.13: 95073189-95073189 on Build GRCh38
rs778302118 Chr.13: 95073266-95073266 on Build GRCh38
rs536521419 Chr.13: 95072945-95072945 on Build GRCh38
rs201886144 Chr.13: 95073309-95073309 on Build GRCh38
rs377095942 Chr.13: 95073167-95073167 on Build GRCh38
rs750227626 Chr.13: 95073332-95073332 on Build GRCh38
rs906126408 Chr.13: 95073068-95073068 on Build GRCh38
rs369993880 Chr.13: 95073294-95073294 on Build GRCh38
rs755985570 Chr.13: 95073334-95073334 on Build GRCh38
rs147550379 Chr.13: 95073133-95073133 on Build GRCh38
rs754450666 Chr.13: 95073265-95073265 on Build GRCh38
rs775696518 Chr.13: 95073308-95073308 on Build GRCh38
rs2274401 Chr.13: 95073105-95073105 on Build GRCh38
rs970160658 Chr.13: 95073293-95073293 on Build GRCh38
rs1029697679 Chr.13: 95072899-95072899 on Build GRCh38
rs62637636 Chr.13: 95073267-95073267 on Build GRCh38
rs920430382 Chr.13: 95072972-95072972 on Build GRCh38
rs769948414 Chr.13: 95073302-95073302 on Build GRCh38
rs771509326 Chr.13: 95073268-95073268 on Build GRCh38
rs771562574 Chr.13: 95073198-95073198 on Build GRCh38
rs765512296 Chr.13: 95073171-95073171 on Build GRCh38
rs776977943 Chr.13: 95073291-95073291 on Build GRCh38
rs148428145 Chr.13: 95072982-95072982 on Build GRCh38
rs767627685 Chr.13: 95073319-95073319 on Build GRCh38
rs773604803 Chr.13: 95073154-95073154 on Build GRCh38
rs112970489 Chr.13: 95072937-95072937 on Build GRCh38
rs749827594 Chr.13: 95073161-95073161 on Build GRCh38
rs753050401 Chr.13: 95072981-95072981 on Build GRCh38
rs755821526 Chr.13: 95073235-95073235 on Build GRCh38
rs947714611 Chr.13: 95073013-95073013 on Build GRCh38
rs148288608 Chr.13: 95073082-95073083 on Build GRCh38
rs944782335 Chr.13: 95073061-95073061 on Build GRCh38
rs761621325 Chr.13: 95073311-95073311 on Build GRCh38
rs992225267 Chr.13: 95072954-95072954 on Build GRCh38
rs149024948 Chr.13: 95073230-95073230 on Build GRCh38
rs771230786 Chr.13: 95073165-95073165 on Build GRCh38
rs955469123 Chr.13: 95072901-95072901 on Build GRCh38
rs1025464331 Chr.13: 95073275-95073275 on Build GRCh38
rs920486617 Chr.13: 95073306-95073306 on Build GRCh38
rs187944090 Chr.13: 95073089-95073089 on Build GRCh38
rs775535241 Chr.13: 95073186-95073186 on Build GRCh38
rs774369738 Chr.13: 95073324-95073324 on Build GRCh38
rs767394657 Chr.13: 95073229-95073229 on Build GRCh38
rs779443384 Chr.13: 95073259-95073259 on Build GRCh38
rs560531339 Chr.13: 95073005-95073006 on Build GRCh38
rs757024501 Chr.13: 95073223-95073223 on Build GRCh38
rs539864465 Chr.13: 95073143-95073143 on Build GRCh38
rs762958479 Chr.13: 95073187-95073187 on Build GRCh38
rs985357884 Chr.13: 95072902-95072902 on Build GRCh38
rs145069335 Chr.13: 95073037-95073037 on Build GRCh38
rs962492314 Chr.13: 95072924-95072924 on Build GRCh38
rs542982093 Chr.13: 95073258-95073258 on Build GRCh38
rs1039171556 Chr.13: 95073342-95073342 on Build GRCh38
rs936136147 Chr.13: 95073087-95073087 on Build GRCh38
rs192079317 Chr.13: 95073170-95073170 on Build GRCh38
rs9972066 Chr.13: 95072949-95072949 on Build GRCh38

View Full Product Details