Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110642229-110642724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARS2
Entrez Gene ID
79587
Gene Name
cysteinyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
COXPD27, cysRS
UniGene
Hs.508725
Gene Chromosome Location
Chr.13: 110641410-110713527 on Build GRCh38
Transcripts
NM_024537.3 XM_006719953.2 XM_011521116.1 XM_017020741.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6398469 c.1601C>T p.T534I Chr.13: 110642337-110642337 on Build GRCh38
6620327 c.1465G>A p.E489K Chr.13: 110642473-110642473 on Build GRCh38
6244640 c.1437C>A p.S479R Chr.13: 110642501-110642501 on Build GRCh38
3744066 c.1530C>T p.D510D Chr.13: 110642408-110642408 on Build GRCh38
6321936 c.1521C>A p.A507A Chr.13: 110642417-110642417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764139625 Chr.13: 110642582-110642582 on Build GRCh38
rs768619007 Chr.13: 110642666-110642666 on Build GRCh38
rs869123815 Chr.13: 110642579-110642580 on Build GRCh38
rs372410471 Chr.13: 110642414-110642414 on Build GRCh38
rs866090910 Chr.13: 110642579-110642580 on Build GRCh38
rs776708099 Chr.13: 110642411-110642411 on Build GRCh38
rs554100218 Chr.13: 110642312-110642312 on Build GRCh38
rs1053982606 Chr.13: 110642314-110642314 on Build GRCh38
rs765236726 Chr.13: 110642685-110642685 on Build GRCh38
rs558254593 Chr.13: 110642376-110642376 on Build GRCh38
rs764789216 Chr.13: 110642532-110642532 on Build GRCh38
rs531837939 Chr.13: 110642294-110642294 on Build GRCh38
rs556039252 Chr.13: 110642615-110642615 on Build GRCh38
rs371876889 Chr.13: 110642346-110642346 on Build GRCh38
rs200491395 Chr.13: 110642403-110642403 on Build GRCh38
rs147216443 Chr.13: 110642407-110642407 on Build GRCh38
rs776614771 Chr.13: 110642669-110642669 on Build GRCh38
rs750963303 Chr.13: 110642364-110642364 on Build GRCh38
rs746765274 Chr.13: 110642664-110642664 on Build GRCh38
rs1018661935 Chr.13: 110642441-110642452 on Build GRCh38
rs201472735 Chr.13: 110642357-110642357 on Build GRCh38
rs568639299 Chr.13: 110642519-110642519 on Build GRCh38
rs758493385 Chr.13: 110642620-110642620 on Build GRCh38
rs200697273 Chr.13: 110642458-110642458 on Build GRCh38
rs761277278 Chr.13: 110642531-110642531 on Build GRCh38
rs762583126 Chr.13: 110642428-110642428 on Build GRCh38
rs200151026 Chr.13: 110642464-110642464 on Build GRCh38
rs539168705 Chr.13: 110642408-110642408 on Build GRCh38
rs955591697 Chr.13: 110642425-110642425 on Build GRCh38
rs748803394 Chr.13: 110642596-110642596 on Build GRCh38
rs367617450 Chr.13: 110642686-110642686 on Build GRCh38
rs59247777 Chr.13: 110642652-110642652 on Build GRCh38
rs1008749357 Chr.13: 110642593-110642593 on Build GRCh38
rs765964848 Chr.13: 110642535-110642535 on Build GRCh38
rs775887829 Chr.13: 110642527-110642527 on Build GRCh38
rs140567973 Chr.13: 110642448-110642448 on Build GRCh38
rs755324076 Chr.13: 110642690-110642690 on Build GRCh38
rs199527718 Chr.13: 110642404-110642404 on Build GRCh38
rs868284111 Chr.13: 110642328-110642328 on Build GRCh38
rs371859424 Chr.13: 110642540-110642540 on Build GRCh38
rs940558340 Chr.13: 110642283-110642283 on Build GRCh38
rs372084282 Chr.13: 110642492-110642492 on Build GRCh38
rs747748040 Chr.13: 110642575-110642576 on Build GRCh38
rs761927444 Chr.13: 110642617-110642617 on Build GRCh38
rs758790375 Chr.13: 110642588-110642588 on Build GRCh38
rs769106048 Chr.13: 110642594-110642594 on Build GRCh38
rs753574371 Chr.13: 110642397-110642397 on Build GRCh38
rs780333994 Chr.13: 110642589-110642589 on Build GRCh38
rs142158842 Chr.13: 110642274-110642274 on Build GRCh38
rs753109147 Chr.13: 110642692-110642692 on Build GRCh38
rs915844703 Chr.13: 110642654-110642654 on Build GRCh38
rs550431344 Chr.13: 110642298-110642298 on Build GRCh38
rs775479641 Chr.13: 110642348-110642348 on Build GRCh38
rs749637648 Chr.13: 110642615-110642615 on Build GRCh38
rs138022576 Chr.13: 110642400-110642400 on Build GRCh38
rs771193928 Chr.13: 110642507-110642507 on Build GRCh38
rs778065573 Chr.13: 110642544-110642544 on Build GRCh38
rs547578192 Chr.13: 110642360-110642360 on Build GRCh38
rs752100982 Chr.13: 110642378-110642378 on Build GRCh38
rs765758054 Chr.13: 110642363-110642363 on Build GRCh38
rs994699107 Chr.13: 110642393-110642393 on Build GRCh38
rs568697181 Chr.13: 110642302-110642302 on Build GRCh38
rs375781277 Chr.13: 110642537-110642537 on Build GRCh38
rs373027990 Chr.13: 110642487-110642487 on Build GRCh38
rs758180871 Chr.13: 110642488-110642488 on Build GRCh38
rs756519020 Chr.13: 110642698-110642698 on Build GRCh38
rs548268562 Chr.13: 110642257-110642257 on Build GRCh38
rs760155305 Chr.13: 110642466-110642466 on Build GRCh38
rs779837938 Chr.13: 110642447-110642447 on Build GRCh38
rs777258774 Chr.13: 110642386-110642386 on Build GRCh38
rs77995789 Chr.13: 110642579-110642579 on Build GRCh38
rs377071463 Chr.13: 110642553-110642553 on Build GRCh38
rs397695391 Chr.13: 110642578-110642579 on Build GRCh38
rs535459837 Chr.13: 110642607-110642607 on Build GRCh38
rs376713706 Chr.13: 110642415-110642415 on Build GRCh38
rs759617223 Chr.13: 110642667-110642669 on Build GRCh38
rs1043038561 Chr.13: 110642279-110642279 on Build GRCh38
rs146355429 Chr.13: 110642449-110642449 on Build GRCh38
rs949779416 Chr.13: 110642505-110642505 on Build GRCh38
rs777503937 Chr.13: 110642329-110642329 on Build GRCh38
rs778546592 Chr.13: 110642445-110642445 on Build GRCh38
rs375216303 Chr.13: 110642691-110642691 on Build GRCh38
rs548644174 Chr.13: 110642253-110642253 on Build GRCh38
rs113709957 Chr.13: 110642538-110642538 on Build GRCh38
rs144664137 Chr.13: 110642473-110642473 on Build GRCh38
rs546257922 Chr.13: 110642688-110642688 on Build GRCh38
rs753863282 Chr.13: 110642616-110642616 on Build GRCh38
rs778799638 Chr.13: 110642345-110642345 on Build GRCh38
rs552719429 Chr.13: 110642265-110642265 on Build GRCh38
rs774705663 Chr.13: 110642465-110642465 on Build GRCh38
rs756833981 Chr.13: 110642436-110642436 on Build GRCh38
rs370731584 Chr.13: 110642662-110642662 on Build GRCh38
rs750129111 Chr.13: 110642486-110642486 on Build GRCh38
rs368624802 Chr.13: 110642422-110642422 on Build GRCh38
rs11414164 Chr.13: 110642573-110642574 on Build GRCh38
rs374641891 Chr.13: 110642474-110642474 on Build GRCh38
rs762393974 Chr.13: 110642555-110642555 on Build GRCh38
rs774040091 Chr.13: 110642605-110642605 on Build GRCh38
rs770397058 Chr.13: 110642604-110642604 on Build GRCh38
rs201457197 Chr.13: 110642539-110642539 on Build GRCh38
rs779023166 Chr.13: 110642546-110642546 on Build GRCh38
rs763717023 Chr.13: 110642433-110642433 on Build GRCh38
rs754888866 Chr.13: 110642499-110642499 on Build GRCh38
rs558215984 Chr.13: 110642581-110642581 on Build GRCh38
rs762666073 Chr.13: 110642290-110642290 on Build GRCh38
rs770310270 Chr.13: 110642576-110642577 on Build GRCh38
rs752148416 Chr.13: 110642430-110642430 on Build GRCh38
rs761330441 Chr.13: 110642484-110642484 on Build GRCh38
rs753916889 Chr.13: 110642583-110642583 on Build GRCh38
rs761592702 Chr.13: 110642677-110642677 on Build GRCh38
rs369472568 Chr.13: 110642383-110642383 on Build GRCh38
rs758217572 Chr.13: 110642446-110642446 on Build GRCh38
rs773915666 Chr.13: 110642573-110642573 on Build GRCh38
rs759354502 Chr.13: 110642574-110642574 on Build GRCh38
rs781648351 Chr.13: 110642647-110642647 on Build GRCh38
rs574591117 Chr.13: 110642648-110642648 on Build GRCh38
rs763434479 Chr.13: 110642423-110642423 on Build GRCh38
rs999658016 Chr.13: 110642624-110642624 on Build GRCh38
rs1004519591 Chr.13: 110642351-110642351 on Build GRCh38
rs748996703 Chr.13: 110642330-110642330 on Build GRCh38
rs566143026 Chr.13: 110642347-110642347 on Build GRCh38
rs148358273 Chr.13: 110642256-110642256 on Build GRCh38
rs753970334 Chr.13: 110642545-110642545 on Build GRCh38
rs747585911 Chr.13: 110642550-110642550 on Build GRCh38
rs766266851 Chr.13: 110642490-110642490 on Build GRCh38
rs568391657 Chr.13: 110642333-110642333 on Build GRCh38
rs376293005 Chr.13: 110642547-110642547 on Build GRCh38
rs752848136 Chr.13: 110642316-110642321 on Build GRCh38
rs761608377 Chr.13: 110642559-110642560 on Build GRCh38
rs200021381 Chr.13: 110642580-110642580 on Build GRCh38
rs982253279 Chr.13: 110642559-110642559 on Build GRCh38
rs531077614 Chr.13: 110642699-110642699 on Build GRCh38
rs769159060 Chr.13: 110642552-110642552 on Build GRCh38
rs756303536 Chr.13: 110642501-110642501 on Build GRCh38
rs760818887 Chr.13: 110642349-110642349 on Build GRCh38
rs953211656 Chr.13: 110642398-110642398 on Build GRCh38
rs774917578 Chr.13: 110642632-110642633 on Build GRCh38
rs1004975550 Chr.13: 110642462-110642462 on Build GRCh38
rs770579480 Chr.13: 110642572-110642572 on Build GRCh38
rs930328975 Chr.13: 110642703-110642703 on Build GRCh38
rs377485989 Chr.13: 110642638-110642638 on Build GRCh38
rs759647154 Chr.13: 110642273-110642274 on Build GRCh38
rs767794681 Chr.13: 110642277-110642278 on Build GRCh38
rs912600400 Chr.13: 110642263-110642263 on Build GRCh38
rs750656672 Chr.13: 110642587-110642587 on Build GRCh38
rs374867586 Chr.13: 110642659-110642659 on Build GRCh38
rs757675624 Chr.13: 110642273-110642273 on Build GRCh38
rs774709224 Chr.13: 110642518-110642518 on Build GRCh38
rs751019720 Chr.13: 110642275-110642275 on Build GRCh38
rs373546015 Chr.13: 110642619-110642619 on Build GRCh38
rs200972973 Chr.13: 110642592-110642592 on Build GRCh38
rs750550451 Chr.13: 110642497-110642497 on Build GRCh38
rs370709222 Chr.13: 110642687-110642687 on Build GRCh38
rs775365088 Chr.13: 110642609-110642609 on Build GRCh38
rs780529860 Chr.13: 110642549-110642549 on Build GRCh38
rs549395705 Chr.13: 110642572-110642573 on Build GRCh38
rs781269830 Chr.13: 110642453-110642453 on Build GRCh38
rs772850443 Chr.13: 110642533-110642533 on Build GRCh38
rs538042133 Chr.13: 110642606-110642606 on Build GRCh38
rs766613503 Chr.13: 110642637-110642637 on Build GRCh38
rs754535107 Chr.13: 110642574-110642575 on Build GRCh38
rs771396401 Chr.13: 110642629-110642630 on Build GRCh38
rs368020365 Chr.13: 110642305-110642305 on Build GRCh38
rs748191524 Chr.13: 110642457-110642457 on Build GRCh38
rs377120724 Chr.13: 110642372-110642372 on Build GRCh38
rs767008467 Chr.13: 110642377-110642377 on Build GRCh38
rs564455691 Chr.13: 110642463-110642463 on Build GRCh38
rs372734645 Chr.13: 110642506-110642506 on Build GRCh38
rs192532265 Chr.13: 110642500-110642500 on Build GRCh38
rs766953234 Chr.13: 110642429-110642429 on Build GRCh38
rs772359189 Chr.13: 110642548-110642548 on Build GRCh38
rs781688053 Chr.13: 110642595-110642595 on Build GRCh38
rs143777330 Chr.13: 110642401-110642401 on Build GRCh38
rs757311069 Chr.13: 110642584-110642584 on Build GRCh38
rs150780076 Chr.13: 110642359-110642359 on Build GRCh38
rs753425826 Chr.13: 110642435-110642435 on Build GRCh38
rs902904499 Chr.13: 110642511-110642511 on Build GRCh38

View Full Product Details