Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 110687338-110687855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARS2
Entrez Gene ID
79587
Gene Name
cysteinyl-tRNA synthetase 2, mitochondrial (putative)
Gene Aliases
COXPD27, cysRS
UniGene
Hs.508725
Gene Chromosome Location
Chr.13: 110641410-110713527 on Build GRCh38
Transcripts
NM_024537.3 XM_006719953.2 XM_011521116.1 XM_011521117.2 XM_011521118.2 XM_017020741.1 XM_017020742.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1161075 c.466G>A p.V156I Chr.13: 110687826-110687826 on Build GRCh38
7302285 c.480G>A p.T160T Chr.13: 110687812-110687812 on Build GRCh38
1493138 c.565G>A p.A189T Chr.13: 110687727-110687727 on Build GRCh38
6285324 c.544C>T p.R182C Chr.13: 110687748-110687748 on Build GRCh38
7412279 c.513G>C p.Q171H Chr.13: 110687779-110687779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751237567 Chr.13: 110687757-110687757 on Build GRCh38
rs141903932 Chr.13: 110687468-110687468 on Build GRCh38
rs764848441 Chr.13: 110687757-110687766 on Build GRCh38
rs749912683 Chr.13: 110687700-110687700 on Build GRCh38
rs771107338 Chr.13: 110687548-110687548 on Build GRCh38
rs931379000 Chr.13: 110687528-110687528 on Build GRCh38
rs981574676 Chr.13: 110687408-110687408 on Build GRCh38
rs560243221 Chr.13: 110687485-110687485 on Build GRCh38
rs756008590 Chr.13: 110687770-110687770 on Build GRCh38
rs902277990 Chr.13: 110687645-110687645 on Build GRCh38
rs879031915 Chr.13: 110687678-110687678 on Build GRCh38
rs1000579273 Chr.13: 110687697-110687697 on Build GRCh38
rs538031990 Chr.13: 110687774-110687774 on Build GRCh38
rs767191718 Chr.13: 110687831-110687831 on Build GRCh38
rs765977477 Chr.13: 110687704-110687704 on Build GRCh38
rs777784801 Chr.13: 110687718-110687718 on Build GRCh38
rs763610904 Chr.13: 110687704-110687706 on Build GRCh38
rs989855610 Chr.13: 110687644-110687644 on Build GRCh38
rs756975496 Chr.13: 110687709-110687711 on Build GRCh38
rs532037153 Chr.13: 110687686-110687686 on Build GRCh38
rs1037424130 Chr.13: 110687497-110687497 on Build GRCh38
rs200187461 Chr.13: 110687692-110687692 on Build GRCh38
rs373960867 Chr.13: 110687710-110687710 on Build GRCh38
rs752514843 Chr.13: 110687766-110687766 on Build GRCh38
rs572181410 Chr.13: 110687549-110687549 on Build GRCh38
rs375907408 Chr.13: 110687705-110687706 on Build GRCh38
rs375287344 Chr.13: 110687679-110687679 on Build GRCh38
rs751347522 Chr.13: 110687706-110687706 on Build GRCh38
rs750847494 Chr.13: 110687775-110687776 on Build GRCh38
rs758019231 Chr.13: 110687701-110687701 on Build GRCh38
rs772630304 Chr.13: 110687737-110687737 on Build GRCh38
rs533611037 Chr.13: 110687740-110687740 on Build GRCh38
rs897190821 Chr.13: 110687519-110687520 on Build GRCh38
rs148098469 Chr.13: 110687826-110687826 on Build GRCh38
rs779188351 Chr.13: 110687803-110687803 on Build GRCh38
rs9645916 Chr.13: 110687796-110687796 on Build GRCh38
rs749363417 Chr.13: 110687719-110687719 on Build GRCh38
rs760115460 Chr.13: 110687703-110687711 on Build GRCh38
rs759143144 Chr.13: 110687829-110687829 on Build GRCh38
rs747408982 Chr.13: 110687819-110687819 on Build GRCh38
rs758758975 Chr.13: 110687833-110687833 on Build GRCh38
rs72661692 Chr.13: 110687754-110687754 on Build GRCh38
rs200663762 Chr.13: 110687694-110687695 on Build GRCh38
rs747159621 Chr.13: 110687365-110687365 on Build GRCh38
rs770282381 Chr.13: 110687827-110687827 on Build GRCh38
rs1024851218 Chr.13: 110687377-110687377 on Build GRCh38
rs758955126 Chr.13: 110687679-110687680 on Build GRCh38
rs895235203 Chr.13: 110687530-110687530 on Build GRCh38
rs142034206 Chr.13: 110687729-110687729 on Build GRCh38
rs552751411 Chr.13: 110687692-110687693 on Build GRCh38
rs1005137257 Chr.13: 110687716-110687716 on Build GRCh38
rs759387072 Chr.13: 110687693-110687694 on Build GRCh38
rs769254654 Chr.13: 110687822-110687822 on Build GRCh38
rs974499032 Chr.13: 110687515-110687515 on Build GRCh38
rs944189904 Chr.13: 110687594-110687594 on Build GRCh38
rs748213415 Chr.13: 110687672-110687672 on Build GRCh38
rs985647548 Chr.13: 110687466-110687466 on Build GRCh38
rs771083627 Chr.13: 110687691-110687691 on Build GRCh38
rs955739276 Chr.13: 110687615-110687615 on Build GRCh38
rs9583527 Chr.13: 110687694-110687694 on Build GRCh38
rs143602497 Chr.13: 110687813-110687813 on Build GRCh38
rs78646508 Chr.13: 110687695-110687695 on Build GRCh38
rs1022810530 Chr.13: 110687553-110687553 on Build GRCh38
rs776195135 Chr.13: 110687696-110687696 on Build GRCh38
rs914733209 Chr.13: 110687426-110687426 on Build GRCh38
rs779941643 Chr.13: 110687535-110687535 on Build GRCh38
rs936449486 Chr.13: 110687823-110687823 on Build GRCh38
rs780525993 Chr.13: 110687679-110687681 on Build GRCh38
rs919888079 Chr.13: 110687534-110687534 on Build GRCh38
rs775998657 Chr.13: 110687739-110687739 on Build GRCh38
rs546395837 Chr.13: 110687620-110687620 on Build GRCh38
rs769307767 Chr.13: 110687747-110687747 on Build GRCh38
rs746202062 Chr.13: 110687678-110687679 on Build GRCh38
rs187077112 Chr.13: 110687369-110687369 on Build GRCh38
rs12385854 Chr.13: 110687422-110687422 on Build GRCh38
rs753936012 Chr.13: 110687793-110687793 on Build GRCh38
rs201010636 Chr.13: 110687727-110687727 on Build GRCh38
rs767243208 Chr.13: 110687765-110687765 on Build GRCh38
rs1031354219 Chr.13: 110687368-110687368 on Build GRCh38
rs932859876 Chr.13: 110687458-110687458 on Build GRCh38
rs780516056 Chr.13: 110687816-110687816 on Build GRCh38
rs370120997 Chr.13: 110687692-110687692 on Build GRCh38
rs550255236 Chr.13: 110687688-110687688 on Build GRCh38
rs989886850 Chr.13: 110687416-110687416 on Build GRCh38
rs775497917 Chr.13: 110687699-110687700 on Build GRCh38
rs377385794 Chr.13: 110687786-110687786 on Build GRCh38
rs764771526 Chr.13: 110687698-110687698 on Build GRCh38
rs773286463 Chr.13: 110687685-110687685 on Build GRCh38
rs769930388 Chr.13: 110687673-110687673 on Build GRCh38
rs756061981 Chr.13: 110687715-110687715 on Build GRCh38
rs190348168 Chr.13: 110687625-110687625 on Build GRCh38
rs560857761 Chr.13: 110687807-110687807 on Build GRCh38
rs907091977 Chr.13: 110687561-110687561 on Build GRCh38
rs369230086 Chr.13: 110687706-110687706 on Build GRCh38
rs574529714 Chr.13: 110687433-110687433 on Build GRCh38
rs76988055 Chr.13: 110687728-110687728 on Build GRCh38
rs759194567 Chr.13: 110687764-110687764 on Build GRCh38
rs1042529790 Chr.13: 110687609-110687609 on Build GRCh38
rs142925085 Chr.13: 110687755-110687755 on Build GRCh38
rs912747218 Chr.13: 110687744-110687744 on Build GRCh38
rs368569577 Chr.13: 110687584-110687584 on Build GRCh38
rs768718396 Chr.13: 110687679-110687692 on Build GRCh38
rs770666639 Chr.13: 110687693-110687693 on Build GRCh38
rs115225996 Chr.13: 110687387-110687387 on Build GRCh38
rs145988159 Chr.13: 110687752-110687752 on Build GRCh38
rs773926534 Chr.13: 110687828-110687828 on Build GRCh38
rs201439493 Chr.13: 110687709-110687709 on Build GRCh38
rs748013270 Chr.13: 110687711-110687711 on Build GRCh38
rs746008234 Chr.13: 110687812-110687812 on Build GRCh38
rs918590287 Chr.13: 110687783-110687783 on Build GRCh38
rs1026981869 Chr.13: 110687731-110687731 on Build GRCh38
rs944313633 Chr.13: 110687493-110687493 on Build GRCh38
rs912752791 Chr.13: 110687573-110687573 on Build GRCh38
rs757335344 Chr.13: 110687794-110687794 on Build GRCh38

View Full Product Details