Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21312009-21312551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPGRIP1
Entrez Gene ID
57096
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
Gene Aliases
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d
UniGene
Hs.126035
Gene Chromosome Location
Chr.14: 21287977-21351316 on Build GRCh38
Transcripts
NM_020366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
357222 c.1140A>G p.K380K Chr.14: 21312495-21312495 on Build GRCh38
954376 c.1139A>C p.K380T Chr.14: 21312494-21312494 on Build GRCh38
4824564 c.1112G>A p.R371Q Chr.14: 21312467-21312467 on Build GRCh38
954374 c.1112G>T p.R371L Chr.14: 21312467-21312467 on Build GRCh38
4824563 c.1112G>A p.R371Q Chr.14: 21312467-21312467 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766608701 Chr.14: 21312454-21312454 on Build GRCh38
rs61751266 Chr.14: 21312458-21312458 on Build GRCh38
rs758038121 Chr.14: 21312490-21312490 on Build GRCh38
rs965899524 Chr.14: 21312330-21312330 on Build GRCh38
rs527599505 Chr.14: 21312222-21312222 on Build GRCh38
rs375859404 Chr.14: 21312466-21312466 on Build GRCh38
rs553370787 Chr.14: 21312051-21312051 on Build GRCh38
rs773592041 Chr.14: 21312432-21312432 on Build GRCh38
rs1000891344 Chr.14: 21312198-21312198 on Build GRCh38
rs370820376 Chr.14: 21312349-21312349 on Build GRCh38
rs779600963 Chr.14: 21312497-21312497 on Build GRCh38
rs1032728230 Chr.14: 21312213-21312213 on Build GRCh38
rs893059474 Chr.14: 21312216-21312216 on Build GRCh38
rs755730792 Chr.14: 21312324-21312324 on Build GRCh38
rs61977482 Chr.14: 21312086-21312086 on Build GRCh38
rs1045125176 Chr.14: 21312243-21312243 on Build GRCh38
rs908832510 Chr.14: 21312381-21312381 on Build GRCh38
rs548990299 Chr.14: 21312429-21312429 on Build GRCh38
rs980350108 Chr.14: 21312467-21312467 on Build GRCh38
rs779504787 Chr.14: 21312388-21312388 on Build GRCh38
rs1024600671 Chr.14: 21312442-21312442 on Build GRCh38
rs774569329 Chr.14: 21312458-21312458 on Build GRCh38
rs78876813 Chr.14: 21312160-21312160 on Build GRCh38
rs750114619 Chr.14: 21312485-21312485 on Build GRCh38
rs926273547 Chr.14: 21312250-21312250 on Build GRCh38
rs747174838 Chr.14: 21312423-21312426 on Build GRCh38
rs756520616 Chr.14: 21312468-21312468 on Build GRCh38
rs545500597 Chr.14: 21312078-21312078 on Build GRCh38
rs8021536 Chr.14: 21312157-21312157 on Build GRCh38
rs975897511 Chr.14: 21312339-21312339 on Build GRCh38
rs546703278 Chr.14: 21312178-21312178 on Build GRCh38
rs371312060 Chr.14: 21312424-21312424 on Build GRCh38
rs890365588 Chr.14: 21312326-21312326 on Build GRCh38
rs576378570 Chr.14: 21312073-21312073 on Build GRCh38
rs954761837 Chr.14: 21312085-21312085 on Build GRCh38
rs993317720 Chr.14: 21312271-21312271 on Build GRCh38
rs768628497 Chr.14: 21312416-21312416 on Build GRCh38
rs968775206 Chr.14: 21312125-21312125 on Build GRCh38
rs867552844 Chr.14: 21312474-21312474 on Build GRCh38
rs768072609 Chr.14: 21312463-21312463 on Build GRCh38
rs1037429143 Chr.14: 21312259-21312259 on Build GRCh38
rs368149716 Chr.14: 21312422-21312422 on Build GRCh38
rs949117333 Chr.14: 21312217-21312217 on Build GRCh38
rs568103038 Chr.14: 21312288-21312288 on Build GRCh38
rs547687343 Chr.14: 21312281-21312281 on Build GRCh38
rs776379559 Chr.14: 21312417-21312417 on Build GRCh38
rs764096693 Chr.14: 21312147-21312147 on Build GRCh38
rs190602707 Chr.14: 21312362-21312362 on Build GRCh38
rs1017726981 Chr.14: 21312119-21312119 on Build GRCh38
rs759474031 Chr.14: 21312460-21312460 on Build GRCh38
rs573028664 Chr.14: 21312147-21312147 on Build GRCh38
rs749902482 Chr.14: 21312233-21312233 on Build GRCh38
rs897631598 Chr.14: 21312056-21312056 on Build GRCh38
rs776880045 Chr.14: 21312468-21312468 on Build GRCh38
rs764587478 Chr.14: 21312276-21312276 on Build GRCh38
rs940906655 Chr.14: 21312121-21312121 on Build GRCh38
rs978781775 Chr.14: 21312145-21312145 on Build GRCh38
rs751096098 Chr.14: 21312507-21312507 on Build GRCh38
rs984482762 Chr.14: 21312379-21312379 on Build GRCh38
rs1020208488 Chr.14: 21312270-21312270 on Build GRCh38
rs1010528973 Chr.14: 21312258-21312258 on Build GRCh38
rs561715841 Chr.14: 21312194-21312194 on Build GRCh38
rs1037545213 Chr.14: 21312055-21312055 on Build GRCh38
rs1013700217 Chr.14: 21312167-21312167 on Build GRCh38
rs764415383 Chr.14: 21312471-21312471 on Build GRCh38
rs763245499 Chr.14: 21312446-21312446 on Build GRCh38
rs762000920 Chr.14: 21312496-21312497 on Build GRCh38
rs768719934 Chr.14: 21312439-21312442 on Build GRCh38
rs185939398 Chr.14: 21312348-21312348 on Build GRCh38

View Full Product Details