Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23561884-23562391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1G2
Entrez Gene ID
8906
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Gene Aliases
G2AD
UniGene
Hs.343244
Gene Chromosome Location
Chr.14: 23559563-23569056 on Build GRCh38
Transcripts
NM_001282474.1 NM_001282475.1 NM_003917.4 XM_005268167.3 XM_005268168.4 XM_005268169.3 XM_005268170.3 XM_005268172.3 XM_005268173.3 XM_005268174.3 XM_005268175.3 XM_005268177.3 XM_005268178.3 XM_005268179.3 XM_005268180.4 XM_005268182.3 XM_006720301.3 XM_011537283.2 XM_011537286.2 XM_017021740.1 XM_017021741.1 XM_017021742.1 XM_017021746.1 XM_017021747.1 XM_017021748.1 NR_110555.1

Gene Symbol
LOC102724814
Entrez Gene ID
102724814
Gene Name
uncharacterized LOC102724814
Gene Aliases
-
UniGene
Hs.569375
Gene Chromosome Location
Chr.14: 23561097-23568076 on Build GRCh38
Transcripts
NM_001282474.1 NM_001282475.1 NM_003917.4 XM_005268167.3 XM_005268168.4 XM_005268169.3 XM_005268170.3 XM_005268172.3 XM_005268173.3 XM_005268174.3 XM_005268175.3 XM_005268177.3 XM_005268178.3 XM_005268179.3 XM_005268180.4 XM_005268182.3 XM_006720301.3 XM_011537283.2 XM_011537286.2 XM_017021740.1 XM_017021741.1 XM_017021742.1 XM_017021746.1 XM_017021747.1 XM_017021748.1 NR_110555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6412926 c.1283A>C p.K428T Chr.14: 23561977-23561977 on Build GRCh38
5421269 c.1156C>G p.L386V Chr.14: 23562325-23562325 on Build GRCh38
6428766 c.1241A>C p.E414A Chr.14: 23562019-23562019 on Build GRCh38
5478661 c.1202G>A p.R401H Chr.14: 23562058-23562058 on Build GRCh38
4050093 c.1175G>A p.R392H Chr.14: 23562306-23562306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763974165 Chr.14: 23562330-23562330 on Build GRCh38
rs142884247 Chr.14: 23562306-23562306 on Build GRCh38
rs762973287 Chr.14: 23562314-23562314 on Build GRCh38
rs148525847 Chr.14: 23562008-23562008 on Build GRCh38
rs148282907 Chr.14: 23562059-23562059 on Build GRCh38
rs748934676 Chr.14: 23562285-23562285 on Build GRCh38
rs373148237 Chr.14: 23561935-23561935 on Build GRCh38
rs766645158 Chr.14: 23562283-23562286 on Build GRCh38
rs569062719 Chr.14: 23562368-23562368 on Build GRCh38
rs753122091 Chr.14: 23561979-23561979 on Build GRCh38
rs769210525 Chr.14: 23562364-23562364 on Build GRCh38
rs772214302 Chr.14: 23562288-23562290 on Build GRCh38
rs779648112 Chr.14: 23562094-23562094 on Build GRCh38
rs747633978 Chr.14: 23562345-23562345 on Build GRCh38
rs760007030 Chr.14: 23561973-23561973 on Build GRCh38
rs533190621 Chr.14: 23562346-23562346 on Build GRCh38
rs199999308 Chr.14: 23562012-23562012 on Build GRCh38
rs778910779 Chr.14: 23561994-23561994 on Build GRCh38
rs202232189 Chr.14: 23562065-23562065 on Build GRCh38
rs367610818 Chr.14: 23561972-23561972 on Build GRCh38
rs948473619 Chr.14: 23562106-23562106 on Build GRCh38
rs756923386 Chr.14: 23562286-23562286 on Build GRCh38
rs777757882 Chr.14: 23562270-23562270 on Build GRCh38
rs755991945 Chr.14: 23562264-23562264 on Build GRCh38
rs142900764 Chr.14: 23562309-23562309 on Build GRCh38
rs771300290 Chr.14: 23561969-23561969 on Build GRCh38
rs765355477 Chr.14: 23562071-23562071 on Build GRCh38
rs765391495 Chr.14: 23562308-23562308 on Build GRCh38
rs761777964 Chr.14: 23561997-23561997 on Build GRCh38
rs748756065 Chr.14: 23562038-23562038 on Build GRCh38
rs767581949 Chr.14: 23561917-23561917 on Build GRCh38
rs754663881 Chr.14: 23562193-23562193 on Build GRCh38
rs773276758 Chr.14: 23562313-23562313 on Build GRCh38
rs777821677 Chr.14: 23561926-23561926 on Build GRCh38
rs760502593 Chr.14: 23562363-23562364 on Build GRCh38
rs762082955 Chr.14: 23562307-23562307 on Build GRCh38
rs756399107 Chr.14: 23561923-23561923 on Build GRCh38
rs1015933658 Chr.14: 23562138-23562138 on Build GRCh38
rs968535646 Chr.14: 23562355-23562355 on Build GRCh38
rs772788148 Chr.14: 23561957-23561957 on Build GRCh38
rs377511055 Chr.14: 23561940-23561940 on Build GRCh38
rs767513116 Chr.14: 23562318-23562318 on Build GRCh38
rs762899254 Chr.14: 23562044-23562045 on Build GRCh38
rs562310960 Chr.14: 23562300-23562300 on Build GRCh38
rs1052189238 Chr.14: 23562302-23562302 on Build GRCh38
rs149292012 Chr.14: 23561961-23561961 on Build GRCh38
rs752499143 Chr.14: 23562243-23562243 on Build GRCh38
rs61977722 Chr.14: 23562179-23562179 on Build GRCh38
rs746377212 Chr.14: 23562051-23562051 on Build GRCh38
rs374503007 Chr.14: 23562031-23562031 on Build GRCh38
rs753559390 Chr.14: 23562139-23562139 on Build GRCh38
rs368747270 Chr.14: 23562039-23562039 on Build GRCh38
rs768979520 Chr.14: 23562305-23562305 on Build GRCh38
rs778637630 Chr.14: 23562239-23562239 on Build GRCh38
rs890597421 Chr.14: 23561999-23561999 on Build GRCh38
rs754938216 Chr.14: 23562003-23562003 on Build GRCh38
rs576634833 Chr.14: 23562067-23562067 on Build GRCh38
rs769627554 Chr.14: 23561956-23561956 on Build GRCh38
rs541567290 Chr.14: 23562325-23562325 on Build GRCh38
rs758206386 Chr.14: 23561937-23561937 on Build GRCh38
rs773328826 Chr.14: 23562370-23562370 on Build GRCh38
rs747797122 Chr.14: 23561949-23561949 on Build GRCh38
rs915979081 Chr.14: 23562215-23562215 on Build GRCh38
rs199694194 Chr.14: 23562327-23562327 on Build GRCh38
rs952501873 Chr.14: 23562075-23562075 on Build GRCh38
rs369245203 Chr.14: 23561947-23561947 on Build GRCh38
rs978635859 Chr.14: 23562033-23562033 on Build GRCh38
rs773343932 Chr.14: 23561973-23561974 on Build GRCh38
rs768476906 Chr.14: 23561942-23561942 on Build GRCh38
rs926767095 Chr.14: 23562202-23562202 on Build GRCh38
rs754886923 Chr.14: 23562113-23562113 on Build GRCh38
rs371365778 Chr.14: 23561981-23561981 on Build GRCh38
rs746734267 Chr.14: 23561941-23561941 on Build GRCh38
rs773770477 Chr.14: 23562044-23562044 on Build GRCh38
rs959853315 Chr.14: 23562200-23562200 on Build GRCh38
rs760833509 Chr.14: 23561980-23561980 on Build GRCh38
rs764307065 Chr.14: 23561988-23561988 on Build GRCh38
rs934440194 Chr.14: 23561914-23561914 on Build GRCh38
rs767564556 Chr.14: 23562117-23562117 on Build GRCh38
rs76920030 Chr.14: 23561989-23561989 on Build GRCh38
rs369638005 Chr.14: 23561921-23561921 on Build GRCh38
rs542059161 Chr.14: 23562213-23562213 on Build GRCh38
rs748335436 Chr.14: 23562230-23562230 on Build GRCh38
rs375677387 Chr.14: 23562064-23562064 on Build GRCh38
rs200780955 Chr.14: 23562058-23562058 on Build GRCh38
rs747512295 Chr.14: 23562356-23562356 on Build GRCh38
rs751633676 Chr.14: 23561911-23561911 on Build GRCh38
rs376968278 Chr.14: 23562023-23562023 on Build GRCh38
rs778432854 Chr.14: 23562296-23562296 on Build GRCh38
rs781121124 Chr.14: 23562007-23562007 on Build GRCh38
rs924502802 Chr.14: 23562035-23562035 on Build GRCh38
rs753591811 Chr.14: 23562336-23562336 on Build GRCh38
rs777598503 Chr.14: 23562027-23562027 on Build GRCh38
rs942149105 Chr.14: 23562203-23562203 on Build GRCh38
rs750526449 Chr.14: 23562082-23562082 on Build GRCh38
rs756869444 Chr.14: 23562338-23562338 on Build GRCh38
rs767888618 Chr.14: 23561975-23561975 on Build GRCh38
rs369820949 Chr.14: 23562299-23562299 on Build GRCh38
rs550111210 Chr.14: 23562367-23562367 on Build GRCh38

View Full Product Details